Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2537907-2538426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAT1
Entrez Gene ID
221927
Gene Name
BRCA1 associated ATM activator 1
Gene Aliases
BAAT1, C7orf27, RMFSL
UniGene
Hs.520623
Gene Chromosome Location
Chr.7: 2537810-2555758 on Build GRCh38
Transcripts
NM_152743.3 XM_005249643.4 XM_011515177.2 XM_011515178.1 XM_011515179.2 XM_011515181.2 XM_011515182.2 XM_011515183.1 XM_011515184.2 XM_011515186.2 XM_017011833.1 XM_017011834.1 XM_017011835.1 XM_017011836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1196144 c.2325G>A p.M775I Chr.7: 2538210-2538210 on Build GRCh38
4651608 c.2241C>T p.S747S Chr.7: 2538294-2538294 on Build GRCh38
4666005 c.2258C>A p.P753Q Chr.7: 2538277-2538277 on Build GRCh38
746354 c.2147C>T p.A716V Chr.7: 2538388-2538388 on Build GRCh38
6177277 c.2212G>A p.E738K Chr.7: 2538323-2538323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369152767 Chr.7: 2538291-2538291 on Build GRCh38
rs763541403 Chr.7: 2538054-2538054 on Build GRCh38
rs772474094 Chr.7: 2538143-2538143 on Build GRCh38
rs369064014 Chr.7: 2538265-2538265 on Build GRCh38
rs765459628 Chr.7: 2538224-2538224 on Build GRCh38
rs760489828 Chr.7: 2538108-2538108 on Build GRCh38
rs561027633 Chr.7: 2537944-2537944 on Build GRCh38
rs780417945 Chr.7: 2538398-2538398 on Build GRCh38
rs773514268 Chr.7: 2538146-2538146 on Build GRCh38
rs1043294 Chr.7: 2538057-2538057 on Build GRCh38
rs541075552 Chr.7: 2537943-2537943 on Build GRCh38
rs375916445 Chr.7: 2538175-2538175 on Build GRCh38
rs751042897 Chr.7: 2538365-2538365 on Build GRCh38
rs781049787 Chr.7: 2538030-2538030 on Build GRCh38
rs865782498 Chr.7: 2538348-2538348 on Build GRCh38
rs528088936 Chr.7: 2538045-2538045 on Build GRCh38
rs959001317 Chr.7: 2538246-2538246 on Build GRCh38
rs557088612 Chr.7: 2538081-2538081 on Build GRCh38
rs762488461 Chr.7: 2538364-2538366 on Build GRCh38
rs948833526 Chr.7: 2538368-2538368 on Build GRCh38
rs762276921 Chr.7: 2538140-2538140 on Build GRCh38
rs975996696 Chr.7: 2538311-2538311 on Build GRCh38
rs757243199 Chr.7: 2538174-2538174 on Build GRCh38
rs766108481 Chr.7: 2538283-2538283 on Build GRCh38
rs756583395 Chr.7: 2538387-2538387 on Build GRCh38
rs923807425 Chr.7: 2538295-2538295 on Build GRCh38
rs61755262 Chr.7: 2538065-2538065 on Build GRCh38
rs866016750 Chr.7: 2538014-2538014 on Build GRCh38
rs556303347 Chr.7: 2537964-2537964 on Build GRCh38
rs200892967 Chr.7: 2538168-2538168 on Build GRCh38
rs368044267 Chr.7: 2538231-2538231 on Build GRCh38
rs777656966 Chr.7: 2538023-2538023 on Build GRCh38
rs190232808 Chr.7: 2538059-2538059 on Build GRCh38
rs759592506 Chr.7: 2538165-2538165 on Build GRCh38
rs750148903 Chr.7: 2538177-2538177 on Build GRCh38
rs761588914 Chr.7: 2538086-2538086 on Build GRCh38
rs903154998 Chr.7: 2537972-2537972 on Build GRCh38
rs1043297 Chr.7: 2538011-2538011 on Build GRCh38
rs759775503 Chr.7: 2538221-2538221 on Build GRCh38
rs546466299 Chr.7: 2538330-2538330 on Build GRCh38
rs753480934 Chr.7: 2538242-2538242 on Build GRCh38
rs372332387 Chr.7: 2538369-2538369 on Build GRCh38
rs921871387 Chr.7: 2538316-2538316 on Build GRCh38
rs773060720 Chr.7: 2538278-2538278 on Build GRCh38
rs770105500 Chr.7: 2538269-2538269 on Build GRCh38
rs376888751 Chr.7: 2538234-2538234 on Build GRCh38
rs774902699 Chr.7: 2538138-2538138 on Build GRCh38
rs376271902 Chr.7: 2538276-2538276 on Build GRCh38
rs35099363 Chr.7: 2538136-2538137 on Build GRCh38
rs144085416 Chr.7: 2538237-2538237 on Build GRCh38
rs755802114 Chr.7: 2538181-2538181 on Build GRCh38
rs757188244 Chr.7: 2538101-2538101 on Build GRCh38
rs200784213 Chr.7: 2538087-2538087 on Build GRCh38
rs767862918 Chr.7: 2538064-2538064 on Build GRCh38
rs373302503 Chr.7: 2538272-2538272 on Build GRCh38
rs201045158 Chr.7: 2538195-2538195 on Build GRCh38
rs747280032 Chr.7: 2538210-2538210 on Build GRCh38
rs60152725 Chr.7: 2538326-2538326 on Build GRCh38
rs752390091 Chr.7: 2538351-2538351 on Build GRCh38
rs759384531 Chr.7: 2538340-2538340 on Build GRCh38
rs759036645 Chr.7: 2538158-2538158 on Build GRCh38
rs778372510 Chr.7: 2538197-2538197 on Build GRCh38
rs766445982 Chr.7: 2538152-2538152 on Build GRCh38
rs148220310 Chr.7: 2537973-2537973 on Build GRCh38
rs765404910 Chr.7: 2538167-2538167 on Build GRCh38
rs758920429 Chr.7: 2538252-2538252 on Build GRCh38
rs763537863 Chr.7: 2538364-2538364 on Build GRCh38
rs61753093 Chr.7: 2538327-2538327 on Build GRCh38
rs746172037 Chr.7: 2538309-2538309 on Build GRCh38
rs770720943 Chr.7: 2538027-2538027 on Build GRCh38
rs747589994 Chr.7: 2538248-2538248 on Build GRCh38
rs773571503 Chr.7: 2538206-2538206 on Build GRCh38
rs775667282 Chr.7: 2538225-2538225 on Build GRCh38
rs757972461 Chr.7: 2538294-2538294 on Build GRCh38
rs1043291 Chr.7: 2538147-2538147 on Build GRCh38
rs761877409 Chr.7: 2538229-2538229 on Build GRCh38
rs372230882 Chr.7: 2538214-2538214 on Build GRCh38
rs771313289 Chr.7: 2538211-2538211 on Build GRCh38
rs760637806 Chr.7: 2538279-2538279 on Build GRCh38
rs768195140 Chr.7: 2538126-2538126 on Build GRCh38
rs149690979 Chr.7: 2538402-2538402 on Build GRCh38
rs139111440 Chr.7: 2538336-2538336 on Build GRCh38
rs1013387094 Chr.7: 2538198-2538198 on Build GRCh38
rs868110270 Chr.7: 2538308-2538308 on Build GRCh38
rs140802292 Chr.7: 2538388-2538388 on Build GRCh38
rs753289498 Chr.7: 2538153-2538153 on Build GRCh38
rs932050999 Chr.7: 2538159-2538159 on Build GRCh38
rs140062589 Chr.7: 2538055-2538055 on Build GRCh38
rs781107087 Chr.7: 2538107-2538107 on Build GRCh38
rs140546659 Chr.7: 2538281-2538281 on Build GRCh38
rs184247341 Chr.7: 2537962-2537962 on Build GRCh38
rs746736482 Chr.7: 2538026-2538026 on Build GRCh38
rs766164248 Chr.7: 2538239-2538239 on Build GRCh38
rs143963658 Chr.7: 2538346-2538346 on Build GRCh38
rs148661872 Chr.7: 2538307-2538307 on Build GRCh38
rs554560289 Chr.7: 2538264-2538264 on Build GRCh38
rs764766013 Chr.7: 2538163-2538163 on Build GRCh38
rs1040384383 Chr.7: 2537957-2537957 on Build GRCh38
rs895676842 Chr.7: 2538002-2538002 on Build GRCh38
rs770266976 Chr.7: 2538313-2538313 on Build GRCh38
rs780485844 Chr.7: 2538325-2538325 on Build GRCh38
rs760784792 Chr.7: 2538076-2538076 on Build GRCh38
rs373538456 Chr.7: 2538293-2538293 on Build GRCh38
rs777308516 Chr.7: 2538301-2538301 on Build GRCh38
rs959097537 Chr.7: 2537978-2537978 on Build GRCh38
rs776302978 Chr.7: 2538339-2538339 on Build GRCh38
rs61733815 Chr.7: 2538110-2538110 on Build GRCh38
rs150300694 Chr.7: 2538366-2538366 on Build GRCh38
rs900069743 Chr.7: 2537959-2537959 on Build GRCh38
rs748961243 Chr.7: 2538192-2538192 on Build GRCh38
rs767501044 Chr.7: 2538233-2538233 on Build GRCh38
rs779616647 Chr.7: 2538109-2538109 on Build GRCh38
rs199914857 Chr.7: 2538277-2538277 on Build GRCh38
rs757736829 Chr.7: 2538304-2538304 on Build GRCh38
rs552913746 Chr.7: 2538007-2538007 on Build GRCh38
rs748686129 Chr.7: 2538119-2538119 on Build GRCh38
rs35955968 Chr.7: 2538268-2538268 on Build GRCh38
rs748675181 Chr.7: 2538202-2538202 on Build GRCh38
rs752058292 Chr.7: 2538302-2538302 on Build GRCh38
rs61729932 Chr.7: 2538182-2538182 on Build GRCh38
rs766501904 Chr.7: 2538080-2538080 on Build GRCh38
rs532794940 Chr.7: 2537990-2537990 on Build GRCh38
rs543555790 Chr.7: 2538285-2538285 on Build GRCh38
rs972064942 Chr.7: 2537967-2537967 on Build GRCh38
rs754574676 Chr.7: 2538245-2538245 on Build GRCh38
rs769390713 Chr.7: 2538040-2538040 on Build GRCh38
rs369607597 Chr.7: 2538204-2538204 on Build GRCh38

View Full Product Details