Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 7245757-7246339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1GALT1
Entrez Gene ID
56913
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1
Gene Aliases
C1GALT, T-synthase
UniGene
Hs.239666
Gene Chromosome Location
Chr.7: 7182547-7248651 on Build GRCh38
Transcripts
NM_020156.4 XM_005249812.1 XM_011515453.2 XM_011515455.2 XM_011515456.2 XM_017012442.1 XM_017012443.1 XM_017012444.1 XM_017012445.1 XM_017012446.1 XM_017012447.1 XM_017012448.1 XM_017012449.1

SNP Detail

SNP ID Chromosome Location
rs892641258 Chr.7: 7246156-7246156 on Build GRCh38
rs1045721948 Chr.7: 7245858-7245858 on Build GRCh38
rs978487613 Chr.7: 7246014-7246014 on Build GRCh38
rs750593557 Chr.7: 7246094-7246094 on Build GRCh38
rs1003818847 Chr.7: 7246148-7246148 on Build GRCh38
rs140130002 Chr.7: 7246089-7246089 on Build GRCh38
rs758906255 Chr.7: 7246173-7246173 on Build GRCh38
rs867545734 Chr.7: 7245788-7245788 on Build GRCh38
rs976563301 Chr.7: 7246036-7246036 on Build GRCh38
rs930068344 Chr.7: 7245838-7245838 on Build GRCh38
rs143762091 Chr.7: 7246114-7246114 on Build GRCh38
rs73049985 Chr.7: 7246077-7246077 on Build GRCh38
rs964023176 Chr.7: 7246034-7246034 on Build GRCh38
rs1035177320 Chr.7: 7245895-7245895 on Build GRCh38
rs760377019 Chr.7: 7245897-7245897 on Build GRCh38
rs377402192 Chr.7: 7246226-7246226 on Build GRCh38
rs750466496 Chr.7: 7246104-7246104 on Build GRCh38
rs202194376 Chr.7: 7246225-7246225 on Build GRCh38
rs146883698 Chr.7: 7246285-7246285 on Build GRCh38
rs930014704 Chr.7: 7246066-7246066 on Build GRCh38
rs200338497 Chr.7: 7246226-7246227 on Build GRCh38
rs572541196 Chr.7: 7246225-7246226 on Build GRCh38
rs201453874 Chr.7: 7245850-7245851 on Build GRCh38
rs536635959 Chr.7: 7245814-7245814 on Build GRCh38
rs187314505 Chr.7: 7245869-7245869 on Build GRCh38
rs1056530921 Chr.7: 7246150-7246150 on Build GRCh38
rs1017962256 Chr.7: 7246301-7246302 on Build GRCh38
rs182414750 Chr.7: 7246242-7246242 on Build GRCh38
rs879273797 Chr.7: 7245844-7245844 on Build GRCh38
rs559152361 Chr.7: 7245968-7245968 on Build GRCh38
rs533802836 Chr.7: 7246009-7246011 on Build GRCh38
rs577293354 Chr.7: 7246018-7246019 on Build GRCh38
rs573287231 Chr.7: 7245883-7245883 on Build GRCh38
rs1045102127 Chr.7: 7246142-7246145 on Build GRCh38
rs1047071279 Chr.7: 7245840-7245840 on Build GRCh38
rs1019987628 Chr.7: 7246255-7246255 on Build GRCh38
rs537833842 Chr.7: 7246051-7246052 on Build GRCh38
rs61047106 Chr.7: 7246226-7246226 on Build GRCh38
rs12536138 Chr.7: 7246238-7246238 on Build GRCh38
rs922667646 Chr.7: 7245824-7245824 on Build GRCh38
rs540766548 Chr.7: 7245969-7245969 on Build GRCh38
rs369570776 Chr.7: 7245855-7245856 on Build GRCh38
rs904304308 Chr.7: 7246280-7246280 on Build GRCh38
rs905427322 Chr.7: 7246147-7246147 on Build GRCh38
rs1019872437 Chr.7: 7246000-7246000 on Build GRCh38
rs555342651 Chr.7: 7245865-7245865 on Build GRCh38
rs532037474 Chr.7: 7246283-7246283 on Build GRCh38
rs73049983 Chr.7: 7245976-7245976 on Build GRCh38
rs114287739 Chr.7: 7245947-7245947 on Build GRCh38
rs758552135 Chr.7: 7246146-7246146 on Build GRCh38

View Full Product Details