Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 7246398-7246976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1GALT1
Entrez Gene ID
56913
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1
Gene Aliases
C1GALT, T-synthase
UniGene
Hs.239666
Gene Chromosome Location
Chr.7: 7182547-7248651 on Build GRCh38
Transcripts
NM_020156.4 XM_005249812.1 XM_011515453.2 XM_011515455.2 XM_011515456.2 XM_017012442.1 XM_017012443.1 XM_017012444.1 XM_017012445.1 XM_017012446.1 XM_017012447.1 XM_017012448.1 XM_017012449.1

SNP Detail

SNP ID Chromosome Location
rs963090391 Chr.7: 7246644-7246644 on Build GRCh38
rs939350044 Chr.7: 7246678-7246678 on Build GRCh38
rs561777155 Chr.7: 7246825-7246825 on Build GRCh38
rs997891281 Chr.7: 7246621-7246621 on Build GRCh38
rs992152555 Chr.7: 7246679-7246679 on Build GRCh38
rs5010183 Chr.7: 7246567-7246567 on Build GRCh38
rs999965137 Chr.7: 7246590-7246590 on Build GRCh38
rs945539587 Chr.7: 7246538-7246538 on Build GRCh38
rs531420374 Chr.7: 7246501-7246501 on Build GRCh38
rs892757345 Chr.7: 7246514-7246514 on Build GRCh38
rs887002165 Chr.7: 7246914-7246914 on Build GRCh38
rs1016787698 Chr.7: 7246638-7246642 on Build GRCh38
rs563823803 Chr.7: 7246760-7246760 on Build GRCh38
rs899504773 Chr.7: 7246612-7246612 on Build GRCh38
rs926575009 Chr.7: 7246668-7246673 on Build GRCh38
rs899618064 Chr.7: 7246859-7246859 on Build GRCh38
rs568325437 Chr.7: 7246525-7246525 on Build GRCh38
rs768822764 Chr.7: 7246421-7246421 on Build GRCh38
rs1056642734 Chr.7: 7246471-7246471 on Build GRCh38
rs941480017 Chr.7: 7246431-7246431 on Build GRCh38
rs759392581 Chr.7: 7246670-7246670 on Build GRCh38
rs560047866 Chr.7: 7246717-7246717 on Build GRCh38
rs529697615 Chr.7: 7246473-7246473 on Build GRCh38
rs982594836 Chr.7: 7246629-7246629 on Build GRCh38
rs139744919 Chr.7: 7246522-7246522 on Build GRCh38
rs115595378 Chr.7: 7246718-7246718 on Build GRCh38
rs563912033 Chr.7: 7246456-7246456 on Build GRCh38
rs5010184 Chr.7: 7246627-7246627 on Build GRCh38
rs140931201 Chr.7: 7246750-7246750 on Build GRCh38
rs997839120 Chr.7: 7246865-7246865 on Build GRCh38
rs981158334 Chr.7: 7246437-7246437 on Build GRCh38
rs114210531 Chr.7: 7246765-7246765 on Build GRCh38
rs777520071 Chr.7: 7246607-7246607 on Build GRCh38
rs575074182 Chr.7: 7246656-7246656 on Build GRCh38
rs556506766 Chr.7: 7246632-7246632 on Build GRCh38
rs1044258213 Chr.7: 7246747-7246747 on Build GRCh38
rs939573019 Chr.7: 7246470-7246470 on Build GRCh38
rs10246303 Chr.7: 7246814-7246814 on Build GRCh38
rs980889933 Chr.7: 7246663-7246663 on Build GRCh38
rs778596952 Chr.7: 7246688-7246688 on Build GRCh38
rs150231701 Chr.7: 7246907-7246907 on Build GRCh38
rs5010182 Chr.7: 7246562-7246562 on Build GRCh38
rs186116553 Chr.7: 7246697-7246697 on Build GRCh38
rs577477187 Chr.7: 7246626-7246626 on Build GRCh38
rs142964556 Chr.7: 7246536-7246536 on Build GRCh38
rs151046065 Chr.7: 7246603-7246603 on Build GRCh38
rs760070624 Chr.7: 7246757-7246757 on Build GRCh38
rs548557324 Chr.7: 7246547-7246547 on Build GRCh38

View Full Product Details