Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 25120636-25121171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYCS
Entrez Gene ID
54205
Gene Name
cytochrome c, somatic
Gene Aliases
CYC, HCS, THC4
UniGene
Hs.437060
Gene Chromosome Location
Chr.7: 25118651-25125361 on Build GRCh38
Transcripts
NM_018947.5

SNP Detail

SNP ID Chromosome Location
rs1054450768 Chr.7: 25121065-25121065 on Build GRCh38
rs973309398 Chr.7: 25121104-25121104 on Build GRCh38
rs989069541 Chr.7: 25120893-25120893 on Build GRCh38
rs770325574 Chr.7: 25120850-25120850 on Build GRCh38
rs965945877 Chr.7: 25121001-25121001 on Build GRCh38
rs1052838666 Chr.7: 25120683-25120683 on Build GRCh38
rs766404372 Chr.7: 25121078-25121078 on Build GRCh38
rs760845205 Chr.7: 25121074-25121074 on Build GRCh38
rs1050006204 Chr.7: 25120736-25120736 on Build GRCh38
rs963546023 Chr.7: 25121102-25121102 on Build GRCh38
rs1037324193 Chr.7: 25121105-25121105 on Build GRCh38
rs546129201 Chr.7: 25120900-25120900 on Build GRCh38
rs543248679 Chr.7: 25120992-25120992 on Build GRCh38
rs1002254692 Chr.7: 25121081-25121081 on Build GRCh38
rs562633520 Chr.7: 25120993-25120993 on Build GRCh38
rs776182103 Chr.7: 25120873-25120873 on Build GRCh38
rs556271252 Chr.7: 25120773-25120773 on Build GRCh38
rs369688580 Chr.7: 25121056-25121056 on Build GRCh38
rs918930406 Chr.7: 25120814-25120814 on Build GRCh38
rs886062240 Chr.7: 25120897-25120897 on Build GRCh38
rs988644734 Chr.7: 25120684-25120684 on Build GRCh38
rs929046840 Chr.7: 25120815-25120815 on Build GRCh38
rs1049935020 Chr.7: 25120915-25120915 on Build GRCh38
rs572009993 Chr.7: 25120914-25120914 on Build GRCh38
rs921694755 Chr.7: 25121026-25121026 on Build GRCh38
rs867547682 Chr.7: 25120729-25120729 on Build GRCh38
rs144018608 Chr.7: 25121111-25121111 on Build GRCh38
rs892434058 Chr.7: 25121068-25121068 on Build GRCh38
rs1001548451 Chr.7: 25120970-25120970 on Build GRCh38
rs1000873227 Chr.7: 25120666-25120666 on Build GRCh38
rs886062238 Chr.7: 25120677-25120677 on Build GRCh38
rs963296978 Chr.7: 25120926-25120926 on Build GRCh38
rs199887192 Chr.7: 25120767-25120767 on Build GRCh38
rs73086238 Chr.7: 25120750-25120750 on Build GRCh38
rs964324837 Chr.7: 25120665-25120665 on Build GRCh38
rs987170585 Chr.7: 25120984-25120984 on Build GRCh38
rs906611983 Chr.7: 25121080-25121080 on Build GRCh38
rs548081671 Chr.7: 25120728-25120728 on Build GRCh38
rs970084956 Chr.7: 25120831-25120831 on Build GRCh38
rs1024397290 Chr.7: 25120951-25120951 on Build GRCh38
rs886062239 Chr.7: 25120863-25120863 on Build GRCh38
rs1020529555 Chr.7: 25120995-25120995 on Build GRCh38
rs771686993 Chr.7: 25120675-25120675 on Build GRCh38
rs139509288 Chr.7: 25120925-25120925 on Build GRCh38
rs1001553120 Chr.7: 25120874-25120874 on Build GRCh38
rs891264584 Chr.7: 25120841-25120841 on Build GRCh38
rs1032318705 Chr.7: 25120875-25120875 on Build GRCh38
rs1037591350 Chr.7: 25120936-25120936 on Build GRCh38
rs980605108 Chr.7: 25120725-25120725 on Build GRCh38
rs773280177 Chr.7: 25121052-25121052 on Build GRCh38
rs753792536 Chr.7: 25121085-25121085 on Build GRCh38
rs973746714 Chr.7: 25120937-25120937 on Build GRCh38
rs752396666 Chr.7: 25120718-25120718 on Build GRCh38
rs191350311 Chr.7: 25120686-25120686 on Build GRCh38
rs928971514 Chr.7: 25120966-25120966 on Build GRCh38
rs944180777 Chr.7: 25120851-25120851 on Build GRCh38
rs905827556 Chr.7: 25120961-25120961 on Build GRCh38
rs936917659 Chr.7: 25121046-25121046 on Build GRCh38
rs569587122 Chr.7: 25120837-25120837 on Build GRCh38
rs576178443 Chr.7: 25121010-25121010 on Build GRCh38
rs1039882267 Chr.7: 25121015-25121015 on Build GRCh38
rs1032905267 Chr.7: 25120979-25120979 on Build GRCh38
rs1025023699 Chr.7: 25120903-25120903 on Build GRCh38
rs956800961 Chr.7: 25120679-25120679 on Build GRCh38
rs900064706 Chr.7: 25120869-25120869 on Build GRCh38
rs1026640443 Chr.7: 25121108-25121108 on Build GRCh38
rs183671878 Chr.7: 25120759-25120759 on Build GRCh38
rs774802082 Chr.7: 25120950-25120950 on Build GRCh38
rs558630511 Chr.7: 25120902-25120902 on Build GRCh38
rs190557217 Chr.7: 25120986-25120986 on Build GRCh38
rs900014524 Chr.7: 25121034-25121034 on Build GRCh38
rs886062241 Chr.7: 25120938-25120938 on Build GRCh38
rs980552657 Chr.7: 25120917-25120917 on Build GRCh38
rs188208626 Chr.7: 25120954-25120954 on Build GRCh38
rs551405526 Chr.7: 25120667-25120667 on Build GRCh38
rs886062242 Chr.7: 25121137-25121138 on Build GRCh38
rs1033494082 Chr.7: 25121100-25121100 on Build GRCh38
rs147290519 Chr.7: 25121112-25121112 on Build GRCh38
rs769168847 Chr.7: 25120911-25120911 on Build GRCh38
rs917726649 Chr.7: 25120693-25120693 on Build GRCh38
rs527935971 Chr.7: 25120697-25120697 on Build GRCh38
rs918890073 Chr.7: 25120946-25120946 on Build GRCh38
rs973079726 Chr.7: 25120804-25120804 on Build GRCh38
rs1039935947 Chr.7: 25120864-25120864 on Build GRCh38

View Full Product Details