Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 32988515-32989074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP9
Entrez Gene ID
11328
Gene Name
FK506 binding protein 9
Gene Aliases
FKBP60, FKBP63, PPIase
UniGene
Hs.103934
Gene Chromosome Location
Chr.7: 32957404-33006931 on Build GRCh38
Transcripts
NM_001284341.1 NM_001284343.1 NM_007270.4 XM_011515115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1313046 c.921G>A p.T307T Chr.7: 32988534-32988534 on Build GRCh38
1313047 c.225G>A p.T75T Chr.7: 32988534-32988534 on Build GRCh38
5158642 c.1173C>T p.H391H Chr.7: 32988627-32988627 on Build GRCh38
42968 c.1008G>C p.P336P Chr.7: 32988621-32988621 on Build GRCh38
3228242 c.341G>A p.R114K Chr.7: 32988650-32988650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039857506 Chr.7: 32988575-32988575 on Build GRCh38
rs762321128 Chr.7: 32988559-32988559 on Build GRCh38
rs200601982 Chr.7: 32988621-32988621 on Build GRCh38
rs59428480 Chr.7: 32988954-32988954 on Build GRCh38
rs141653893 Chr.7: 32988639-32988639 on Build GRCh38
rs773623765 Chr.7: 32988702-32988702 on Build GRCh38
rs529018899 Chr.7: 32988677-32988677 on Build GRCh38
rs1038385297 Chr.7: 32988947-32988947 on Build GRCh38
rs561555064 Chr.7: 32988625-32988625 on Build GRCh38
rs935579196 Chr.7: 32988548-32988548 on Build GRCh38
rs549062172 Chr.7: 32989036-32989036 on Build GRCh38
rs756924304 Chr.7: 32988665-32988665 on Build GRCh38
rs887114923 Chr.7: 32988876-32988876 on Build GRCh38
rs150348129 Chr.7: 32988553-32988553 on Build GRCh38
rs745430914 Chr.7: 32988620-32988620 on Build GRCh38
rs780746238 Chr.7: 32988608-32988608 on Build GRCh38
rs1044886865 Chr.7: 32988970-32988970 on Build GRCh38
rs1012187730 Chr.7: 32988956-32988956 on Build GRCh38
rs975584453 Chr.7: 32988767-32988767 on Build GRCh38
rs772587807 Chr.7: 32988628-32988628 on Build GRCh38
rs773250761 Chr.7: 32988630-32988630 on Build GRCh38
rs190191486 Chr.7: 32988748-32988748 on Build GRCh38
rs576449965 Chr.7: 32988579-32988579 on Build GRCh38
rs759574063 Chr.7: 32988645-32988645 on Build GRCh38
rs778869322 Chr.7: 32988584-32988584 on Build GRCh38
rs13246887 Chr.7: 32988571-32988571 on Build GRCh38
rs752457854 Chr.7: 32988587-32988587 on Build GRCh38
rs377462158 Chr.7: 32988546-32988546 on Build GRCh38
rs777441645 Chr.7: 32988538-32988538 on Build GRCh38
rs748678484 Chr.7: 32988700-32988700 on Build GRCh38
rs970479116 Chr.7: 32988714-32988714 on Build GRCh38
rs956407122 Chr.7: 32988735-32988735 on Build GRCh38
rs899083059 Chr.7: 32988671-32988671 on Build GRCh38
rs765119101 Chr.7: 32988649-32988649 on Build GRCh38
rs915461022 Chr.7: 32988752-32988752 on Build GRCh38
rs760957917 Chr.7: 32988631-32988631 on Build GRCh38
rs763878973 Chr.7: 32988659-32988659 on Build GRCh38
rs776224928 Chr.7: 32988870-32988870 on Build GRCh38
rs752649846 Chr.7: 32988652-32988652 on Build GRCh38
rs942968732 Chr.7: 32988754-32988754 on Build GRCh38
rs1030652645 Chr.7: 32988729-32988729 on Build GRCh38
rs377325070 Chr.7: 32988664-32988664 on Build GRCh38
rs941300250 Chr.7: 32988929-32988929 on Build GRCh38
rs368581521 Chr.7: 32988566-32988566 on Build GRCh38
rs755719633 Chr.7: 32988691-32988691 on Build GRCh38
rs200951500 Chr.7: 32988813-32988813 on Build GRCh38
rs148054453 Chr.7: 32988593-32988593 on Build GRCh38
rs758616448 Chr.7: 32988843-32988843 on Build GRCh38
rs551077687 Chr.7: 32988911-32988911 on Build GRCh38
rs758127608 Chr.7: 32988655-32988655 on Build GRCh38
rs561070504 Chr.7: 32988600-32988600 on Build GRCh38
rs746459254 Chr.7: 32988543-32988543 on Build GRCh38
rs538273126 Chr.7: 32989045-32989045 on Build GRCh38
rs35727868 Chr.7: 32988875-32988875 on Build GRCh38
rs763738337 Chr.7: 32988833-32988833 on Build GRCh38
rs112980758 Chr.7: 32988716-32988716 on Build GRCh38
rs775799980 Chr.7: 32988549-32988549 on Build GRCh38
rs372039546 Chr.7: 32988683-32988683 on Build GRCh38
rs922787486 Chr.7: 32988799-32988799 on Build GRCh38
rs530628804 Chr.7: 32989022-32989022 on Build GRCh38
rs746683922 Chr.7: 32988604-32988604 on Build GRCh38
rs776022690 Chr.7: 32988659-32988660 on Build GRCh38
rs906387161 Chr.7: 32988996-32988996 on Build GRCh38
rs763516998 Chr.7: 32988552-32988552 on Build GRCh38
rs779513875 Chr.7: 32988697-32988697 on Build GRCh38
rs766495959 Chr.7: 32988577-32988577 on Build GRCh38
rs770278279 Chr.7: 32988605-32988605 on Build GRCh38
rs776748124 Chr.7: 32988640-32988640 on Build GRCh38
rs1003285882 Chr.7: 32989000-32989000 on Build GRCh38
rs545857626 Chr.7: 32988565-32988565 on Build GRCh38
rs772499871 Chr.7: 32988701-32988701 on Build GRCh38

View Full Product Details