Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144138786-144139301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGH1
Entrez Gene ID
51236
Gene Name
HGH1 homolog
Gene Aliases
BRP16, BRP16L, C8orf30A, C8orf30B, FAM203A, FAM203B
UniGene
Hs.300224
Gene Chromosome Location
Chr.8: 144137721-144140843 on Build GRCh38
Transcripts
NM_016458.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4806945 c.793+1G>A p.? Chr.8: 144138814-144138814 on Build GRCh38
7178162 c.778G>A p.E260K Chr.8: 144138798-144138798 on Build GRCh38
1097191 c.793C>A p.R265R Chr.8: 144138813-144138813 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1056919343 Chr.8: 144139024-144139025 on Build GRCh38
rs1017735686 Chr.8: 144138810-144138810 on Build GRCh38
rs938848721 Chr.8: 144138987-144138987 on Build GRCh38
rs998409451 Chr.8: 144139137-144139138 on Build GRCh38
rs559665428 Chr.8: 144138867-144138867 on Build GRCh38
rs775398801 Chr.8: 144138830-144138830 on Build GRCh38
rs779027043 Chr.8: 144138813-144138813 on Build GRCh38
rs951001726 Chr.8: 144138850-144138850 on Build GRCh38
rs946387036 Chr.8: 144138961-144138961 on Build GRCh38
rs758479800 Chr.8: 144138820-144138820 on Build GRCh38
rs983840343 Chr.8: 144138941-144138942 on Build GRCh38
rs902190903 Chr.8: 144139015-144139015 on Build GRCh38
rs541382806 Chr.8: 144138866-144138866 on Build GRCh38
rs368046928 Chr.8: 144138827-144138827 on Build GRCh38
rs963312493 Chr.8: 144139222-144139222 on Build GRCh38
rs950876607 Chr.8: 144139163-144139163 on Build GRCh38
rs1043492668 Chr.8: 144139274-144139274 on Build GRCh38
rs994943225 Chr.8: 144139071-144139071 on Build GRCh38
rs577082937 Chr.8: 144138862-144138862 on Build GRCh38
rs1043763600 Chr.8: 144138976-144138976 on Build GRCh38
rs1017681716 Chr.8: 144139053-144139053 on Build GRCh38
rs1054797041 Chr.8: 144138946-144138946 on Build GRCh38
rs904151101 Chr.8: 144139049-144139049 on Build GRCh38
rs939563133 Chr.8: 144138905-144138905 on Build GRCh38
rs755276640 Chr.8: 144138811-144138811 on Build GRCh38
rs963613306 Chr.8: 144138953-144138953 on Build GRCh38
rs773876745 Chr.8: 144138846-144138846 on Build GRCh38
rs768454477 Chr.8: 144138845-144138845 on Build GRCh38
rs960850132 Chr.8: 144139172-144139172 on Build GRCh38
rs1006821665 Chr.8: 144139073-144139073 on Build GRCh38
rs201604127 Chr.8: 144138818-144138818 on Build GRCh38
rs950748228 Chr.8: 144139095-144139095 on Build GRCh38
rs923563911 Chr.8: 144139279-144139279 on Build GRCh38
rs1042764445 Chr.8: 144139046-144139046 on Build GRCh38
rs1039635497 Chr.8: 144139080-144139080 on Build GRCh38
rs777774810 Chr.8: 144138821-144138821 on Build GRCh38
rs1030824017 Chr.8: 144138922-144138922 on Build GRCh38
rs990972461 Chr.8: 144139215-144139215 on Build GRCh38
rs766775226 Chr.8: 144139059-144139059 on Build GRCh38
rs950776343 Chr.8: 144138923-144138923 on Build GRCh38
rs928256062 Chr.8: 144139159-144139159 on Build GRCh38
rs999815298 Chr.8: 144139022-144139022 on Build GRCh38
rs916766370 Chr.8: 144139219-144139219 on Build GRCh38
rs916362380 Chr.8: 144138954-144138954 on Build GRCh38
rs946430620 Chr.8: 144139250-144139250 on Build GRCh38
rs548639665 Chr.8: 144138892-144138892 on Build GRCh38
rs1006756351 Chr.8: 144139051-144139051 on Build GRCh38
rs563676070 Chr.8: 144138970-144138970 on Build GRCh38
rs983726506 Chr.8: 144138872-144138872 on Build GRCh38
rs374735521 Chr.8: 144138825-144138825 on Build GRCh38
rs890956048 Chr.8: 144139036-144139036 on Build GRCh38
rs530073393 Chr.8: 144138889-144138889 on Build GRCh38
rs928094409 Chr.8: 144138880-144138880 on Build GRCh38
rs1028032893 Chr.8: 144139075-144139075 on Build GRCh38
rs761497189 Chr.8: 144138847-144138847 on Build GRCh38
rs749055818 Chr.8: 144138840-144138840 on Build GRCh38
rs901045858 Chr.8: 144139085-144139085 on Build GRCh38
rs964693872 Chr.8: 144138822-144138822 on Build GRCh38
rs984108911 Chr.8: 144139107-144139107 on Build GRCh38
rs997649372 Chr.8: 144138913-144138913 on Build GRCh38
rs909194336 Chr.8: 144138949-144138949 on Build GRCh38
rs761305022 Chr.8: 144139031-144139031 on Build GRCh38
rs935046316 Chr.8: 144139282-144139282 on Build GRCh38
rs771763459 Chr.8: 144138856-144138856 on Build GRCh38

View Full Product Details