Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 115400266-115400818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEC1
Entrez Gene ID
50514
Gene Name
deleted in esophageal cancer 1
Gene Aliases
CTS9
UniGene
Hs.148841
Gene Chromosome Location
Chr.9: 115141818-115402644 on Build GRCh38
Transcripts
NM_017418.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3653446 c.32G>A p.W11* Chr.9: 115400377-115400377 on Build GRCh38
3413297 c.67C>G p.L23V Chr.9: 115400412-115400412 on Build GRCh38
7275605 c.71C>T p.A24V Chr.9: 115400416-115400416 on Build GRCh38
1739557 c.79C>T p.H27Y Chr.9: 115400424-115400424 on Build GRCh38
366740 c.63C>A p.G21G Chr.9: 115400408-115400408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057423854 Chr.9: 115400602-115400602 on Build GRCh38
rs1005210241 Chr.9: 115400655-115400655 on Build GRCh38
rs758846325 Chr.9: 115400395-115400395 on Build GRCh38
rs759734810 Chr.9: 115400461-115400461 on Build GRCh38
rs373918275 Chr.9: 115400510-115400510 on Build GRCh38
rs955246619 Chr.9: 115400619-115400619 on Build GRCh38
rs1043066245 Chr.9: 115400528-115400528 on Build GRCh38
rs988970619 Chr.9: 115400784-115400784 on Build GRCh38
rs201857798 Chr.9: 115400413-115400413 on Build GRCh38
rs770920402 Chr.9: 115400443-115400443 on Build GRCh38
rs964069452 Chr.9: 115400721-115400721 on Build GRCh38
rs959005945 Chr.9: 115400726-115400726 on Build GRCh38
rs112022194 Chr.9: 115400511-115400511 on Build GRCh38
rs777907264 Chr.9: 115400485-115400485 on Build GRCh38
rs926397084 Chr.9: 115400496-115400497 on Build GRCh38
rs770335256 Chr.9: 115400418-115400418 on Build GRCh38
rs755775515 Chr.9: 115400640-115400640 on Build GRCh38
rs371535859 Chr.9: 115400369-115400369 on Build GRCh38
rs1054378340 Chr.9: 115400556-115400556 on Build GRCh38
rs905769378 Chr.9: 115400546-115400546 on Build GRCh38
rs535342387 Chr.9: 115400581-115400581 on Build GRCh38
rs148421882 Chr.9: 115400681-115400681 on Build GRCh38
rs576370153 Chr.9: 115400632-115400632 on Build GRCh38
rs759738377 Chr.9: 115400346-115400346 on Build GRCh38
rs774468204 Chr.9: 115400334-115400334 on Build GRCh38
rs982743748 Chr.9: 115400470-115400470 on Build GRCh38
rs952318261 Chr.9: 115400316-115400316 on Build GRCh38
rs758363246 Chr.9: 115400451-115400452 on Build GRCh38
rs756735513 Chr.9: 115400397-115400397 on Build GRCh38
rs899956022 Chr.9: 115400583-115400583 on Build GRCh38
rs777447934 Chr.9: 115400691-115400691 on Build GRCh38
rs749762889 Chr.9: 115400427-115400427 on Build GRCh38
rs753445101 Chr.9: 115400391-115400391 on Build GRCh38
rs374778210 Chr.9: 115400474-115400474 on Build GRCh38
rs763931047 Chr.9: 115400479-115400479 on Build GRCh38
rs1042277226 Chr.9: 115400785-115400785 on Build GRCh38
rs185135152 Chr.9: 115400489-115400489 on Build GRCh38
rs368577373 Chr.9: 115400348-115400348 on Build GRCh38
rs937249399 Chr.9: 115400560-115400560 on Build GRCh38
rs748485010 Chr.9: 115400417-115400417 on Build GRCh38
rs778443081 Chr.9: 115400398-115400398 on Build GRCh38
rs753539296 Chr.9: 115400554-115400554 on Build GRCh38
rs973733344 Chr.9: 115400495-115400495 on Build GRCh38
rs778276599 Chr.9: 115400420-115400420 on Build GRCh38
rs762921454 Chr.9: 115400328-115400328 on Build GRCh38
rs199739666 Chr.9: 115400727-115400730 on Build GRCh38
rs1000193594 Chr.9: 115400555-115400555 on Build GRCh38
rs933500108 Chr.9: 115400504-115400504 on Build GRCh38
rs566409899 Chr.9: 115400563-115400563 on Build GRCh38
rs78802664 Chr.9: 115400737-115400737 on Build GRCh38
rs200753855 Chr.9: 115400728-115400732 on Build GRCh38
rs937733306 Chr.9: 115400505-115400505 on Build GRCh38
rs750060380 Chr.9: 115400409-115400409 on Build GRCh38
rs750912529 Chr.9: 115400311-115400311 on Build GRCh38
rs760759160 Chr.9: 115400358-115400358 on Build GRCh38
rs146919515 Chr.9: 115400357-115400357 on Build GRCh38
rs913356243 Chr.9: 115400515-115400515 on Build GRCh38
rs550258789 Chr.9: 115400472-115400472 on Build GRCh38
rs752259957 Chr.9: 115400345-115400346 on Build GRCh38
rs542250807 Chr.9: 115400756-115400756 on Build GRCh38
rs201473510 Chr.9: 115400729-115400733 on Build GRCh38
rs774429975 Chr.9: 115400457-115400457 on Build GRCh38
rs556676378 Chr.9: 115400722-115400722 on Build GRCh38
rs558038493 Chr.9: 115400598-115400598 on Build GRCh38
rs955491127 Chr.9: 115400466-115400466 on Build GRCh38
rs895615860 Chr.9: 115400607-115400607 on Build GRCh38
rs200068429 Chr.9: 115400330-115400330 on Build GRCh38
rs111776574 Chr.9: 115400520-115400520 on Build GRCh38
rs551255525 Chr.9: 115400300-115400300 on Build GRCh38
rs371259513 Chr.9: 115400631-115400631 on Build GRCh38
rs773323886 Chr.9: 115400317-115400317 on Build GRCh38
rs996868446 Chr.9: 115400614-115400614 on Build GRCh38
rs994409973 Chr.9: 115400615-115400615 on Build GRCh38
rs201064588 Chr.9: 115400416-115400416 on Build GRCh38

View Full Product Details