Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41973383-41973880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNP
Entrez Gene ID
1267
Gene Name
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases
CNP1
UniGene
Hs.273621
Gene Chromosome Location
Chr.17: 41966741-41977736 on Build GRCh38
Transcripts
NM_033133.4 XM_006721701.3 XM_011524340.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4066490 c.924C>T p.R308R Chr.17: 41973642-41973642 on Build GRCh38
4130075 c.971C>T p.T324M Chr.17: 41973689-41973689 on Build GRCh38
4884037 c.1081G>A p.G361S Chr.17: 41973739-41973739 on Build GRCh38
7203158 c.775T>C p.S259P Chr.17: 41973493-41973493 on Build GRCh38
5749851 c.998delG p.Q334fs*14 Chr.17: 41973716-41973716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781807639 Chr.17: 41973657-41973657 on Build GRCh38
rs565841044 Chr.17: 41973630-41973630 on Build GRCh38
rs782532165 Chr.17: 41973673-41973673 on Build GRCh38
rs782770728 Chr.17: 41973681-41973681 on Build GRCh38
rs201557422 Chr.17: 41973516-41973516 on Build GRCh38
rs782764243 Chr.17: 41973641-41973641 on Build GRCh38
rs782721891 Chr.17: 41973651-41973651 on Build GRCh38
rs782368828 Chr.17: 41973477-41973477 on Build GRCh38
rs939596473 Chr.17: 41973458-41973458 on Build GRCh38
rs782480365 Chr.17: 41973724-41973724 on Build GRCh38
rs781848378 Chr.17: 41973452-41973452 on Build GRCh38
rs781975778 Chr.17: 41973843-41973843 on Build GRCh38
rs782261384 Chr.17: 41973613-41973613 on Build GRCh38
rs782540990 Chr.17: 41973788-41973788 on Build GRCh38
rs940104617 Chr.17: 41973653-41973653 on Build GRCh38
rs570548839 Chr.17: 41973632-41973632 on Build GRCh38
rs370234532 Chr.17: 41973849-41973849 on Build GRCh38
rs782600136 Chr.17: 41973456-41973456 on Build GRCh38
rs781940101 Chr.17: 41973682-41973682 on Build GRCh38
rs782315979 Chr.17: 41973742-41973742 on Build GRCh38
rs368335643 Chr.17: 41973555-41973555 on Build GRCh38
rs371108522 Chr.17: 41973567-41973567 on Build GRCh38
rs2229931 Chr.17: 41973753-41973753 on Build GRCh38
rs782378608 Chr.17: 41973842-41973842 on Build GRCh38
rs367704141 Chr.17: 41973852-41973852 on Build GRCh38
rs372778050 Chr.17: 41973745-41973745 on Build GRCh38
rs782630033 Chr.17: 41973674-41973674 on Build GRCh38
rs113618186 Chr.17: 41973474-41973474 on Build GRCh38
rs370644988 Chr.17: 41973766-41973766 on Build GRCh38
rs782166990 Chr.17: 41973859-41973859 on Build GRCh38
rs782252776 Chr.17: 41973616-41973616 on Build GRCh38
rs575459583 Chr.17: 41973832-41973832 on Build GRCh38
rs781850469 Chr.17: 41973642-41973642 on Build GRCh38
rs534388905 Chr.17: 41973689-41973689 on Build GRCh38
rs781807577 Chr.17: 41973725-41973726 on Build GRCh38
rs782611282 Chr.17: 41973726-41973726 on Build GRCh38
rs782661998 Chr.17: 41973829-41973829 on Build GRCh38
rs62076903 Chr.17: 41973557-41973557 on Build GRCh38
rs782284945 Chr.17: 41973731-41973731 on Build GRCh38
rs2070106 Chr.17: 41973846-41973846 on Build GRCh38
rs34044046 Chr.17: 41973629-41973629 on Build GRCh38
rs1016790393 Chr.17: 41973805-41973805 on Build GRCh38
rs782225791 Chr.17: 41973581-41973581 on Build GRCh38
rs782752987 Chr.17: 41973700-41973700 on Build GRCh38
rs781997085 Chr.17: 41973690-41973690 on Build GRCh38
rs1055667270 Chr.17: 41973492-41973492 on Build GRCh38
rs199535375 Chr.17: 41973568-41973568 on Build GRCh38
rs568335984 Chr.17: 41973478-41973478 on Build GRCh38
rs367771051 Chr.17: 41973591-41973591 on Build GRCh38
rs782657028 Chr.17: 41973793-41973793 on Build GRCh38
rs536184459 Chr.17: 41973749-41973749 on Build GRCh38
rs782067807 Chr.17: 41973776-41973776 on Build GRCh38
rs552934479 Chr.17: 41973705-41973705 on Build GRCh38
rs202171457 Chr.17: 41973763-41973763 on Build GRCh38
rs369051487 Chr.17: 41973723-41973723 on Build GRCh38
rs372905630 Chr.17: 41973454-41973454 on Build GRCh38
rs1043897984 Chr.17: 41973735-41973735 on Build GRCh38
rs781913833 Chr.17: 41973785-41973785 on Build GRCh38
rs778605376 Chr.17: 41973750-41973750 on Build GRCh38
rs782472388 Chr.17: 41973590-41973590 on Build GRCh38
rs376149897 Chr.17: 41973737-41973737 on Build GRCh38
rs782021529 Chr.17: 41973603-41973603 on Build GRCh38
rs376643712 Chr.17: 41973715-41973715 on Build GRCh38
rs782433845 Chr.17: 41973823-41973823 on Build GRCh38
rs782189743 Chr.17: 41973685-41973685 on Build GRCh38
rs782324779 Chr.17: 41973507-41973507 on Build GRCh38
rs370800761 Chr.17: 41973550-41973550 on Build GRCh38
rs781860039 Chr.17: 41973432-41973432 on Build GRCh38
rs782699148 Chr.17: 41973714-41973714 on Build GRCh38
rs782444637 Chr.17: 41973728-41973728 on Build GRCh38
rs782375707 Chr.17: 41973625-41973625 on Build GRCh38
rs749007400 Chr.17: 41973542-41973542 on Build GRCh38
rs199970993 Chr.17: 41973631-41973631 on Build GRCh38
rs998291648 Chr.17: 41973764-41973764 on Build GRCh38
rs782698086 Chr.17: 41973778-41973778 on Build GRCh38
rs781944543 Chr.17: 41973702-41973702 on Build GRCh38
rs782146999 Chr.17: 41973716-41973716 on Build GRCh38
rs782003027 Chr.17: 41973755-41973755 on Build GRCh38
rs200122334 Chr.17: 41973543-41973543 on Build GRCh38
rs370930907 Chr.17: 41973741-41973741 on Build GRCh38
rs528500610 Chr.17: 41973459-41973459 on Build GRCh38
rs782587919 Chr.17: 41973814-41973814 on Build GRCh38
rs782456858 Chr.17: 41973812-41973812 on Build GRCh38
rs200742095 Chr.17: 41973571-41973571 on Build GRCh38
rs782157834 Chr.17: 41973539-41973539 on Build GRCh38
rs201266482 Chr.17: 41973736-41973736 on Build GRCh38
rs546987699 Chr.17: 41973465-41973465 on Build GRCh38
rs782687743 Chr.17: 41973730-41973730 on Build GRCh38
rs200914994 Chr.17: 41973457-41973457 on Build GRCh38
rs189232763 Chr.17: 41973423-41973423 on Build GRCh38
rs782772375 Chr.17: 41973722-41973722 on Build GRCh38
rs745329861 Chr.17: 41973768-41973768 on Build GRCh38
rs782652904 Chr.17: 41973431-41973431 on Build GRCh38
rs782259540 Chr.17: 41973467-41973467 on Build GRCh38
rs782439120 Chr.17: 41973658-41973658 on Build GRCh38
rs372024910 Chr.17: 41973640-41973640 on Build GRCh38
rs11871638 Chr.17: 41973409-41973409 on Build GRCh38
rs781916228 Chr.17: 41973525-41973525 on Build GRCh38
rs369504990 Chr.17: 41973679-41973679 on Build GRCh38
rs374583295 Chr.17: 41973808-41973808 on Build GRCh38
rs201242451 Chr.17: 41973834-41973834 on Build GRCh38
rs782414384 Chr.17: 41973595-41973595 on Build GRCh38
rs782481884 Chr.17: 41973643-41973643 on Build GRCh38
rs781812372 Chr.17: 41973727-41973727 on Build GRCh38
rs782066235 Chr.17: 41973703-41973703 on Build GRCh38
rs781822198 Chr.17: 41973772-41973772 on Build GRCh38
rs782166517 Chr.17: 41973746-41973746 on Build GRCh38
rs782078035 Chr.17: 41973506-41973506 on Build GRCh38

View Full Product Details