Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41976427-41976932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNP
Entrez Gene ID
1267
Gene Name
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases
CNP1
UniGene
Hs.273621
Gene Chromosome Location
Chr.17: 41966741-41977736 on Build GRCh38
Transcripts
NM_033133.4 NM_001144766.2 NM_003315.3 NR_029431.1 XM_011525167.2 XM_011525168.2 XM_011525169.2 XM_017024994.1

Gene Symbol
DNAJC7
Entrez Gene ID
7266
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
Gene Aliases
DJ11, DJC7, TPR2, TTC2
UniGene
Hs.500156
Gene Chromosome Location
Chr.17: 41976421-42021371 on Build GRCh38
Transcripts
NM_033133.4 NM_001144766.2 NM_003315.3 NR_029431.1 XM_011525167.2 XM_011525168.2 XM_011525169.2 XM_017024994.1

SNP Detail

SNP ID Chromosome Location
rs893579798 Chr.17: 41976522-41976522 on Build GRCh38
rs781956517 Chr.17: 41976793-41976793 on Build GRCh38
rs782201238 Chr.17: 41976717-41976717 on Build GRCh38
rs139687730 Chr.17: 41976461-41976462 on Build GRCh38
rs782403406 Chr.17: 41976759-41976759 on Build GRCh38
rs1009220506 Chr.17: 41976532-41976532 on Build GRCh38
rs782795947 Chr.17: 41976644-41976644 on Build GRCh38
rs532470444 Chr.17: 41976565-41976565 on Build GRCh38
rs945125252 Chr.17: 41976791-41976791 on Build GRCh38
rs782688530 Chr.17: 41976691-41976691 on Build GRCh38
rs551097996 Chr.17: 41976612-41976612 on Build GRCh38
rs781890591 Chr.17: 41976584-41976584 on Build GRCh38
rs890341446 Chr.17: 41976548-41976548 on Build GRCh38
rs1017557160 Chr.17: 41976550-41976550 on Build GRCh38
rs373424749 Chr.17: 41976789-41976789 on Build GRCh38
rs983448360 Chr.17: 41976604-41976604 on Build GRCh38
rs782278404 Chr.17: 41976697-41976697 on Build GRCh38
rs1008605042 Chr.17: 41976885-41976885 on Build GRCh38
rs532804087 Chr.17: 41976602-41976602 on Build GRCh38
rs782520057 Chr.17: 41976709-41976709 on Build GRCh38
rs375855857 Chr.17: 41976731-41976731 on Build GRCh38
rs538440665 Chr.17: 41976893-41976893 on Build GRCh38
rs71705091 Chr.17: 41976468-41976469 on Build GRCh38
rs112594389 Chr.17: 41976558-41976558 on Build GRCh38
rs782811259 Chr.17: 41976474-41976474 on Build GRCh38
rs11383892 Chr.17: 41976472-41976472 on Build GRCh38
rs371726846 Chr.17: 41976471-41976472 on Build GRCh38
rs1053463438 Chr.17: 41976513-41976513 on Build GRCh38
rs782319088 Chr.17: 41976722-41976722 on Build GRCh38
rs782506550 Chr.17: 41976780-41976780 on Build GRCh38
rs782582576 Chr.17: 41976873-41976873 on Build GRCh38
rs577962906 Chr.17: 41976639-41976639 on Build GRCh38
rs551479926 Chr.17: 41976796-41976796 on Build GRCh38
rs782610218 Chr.17: 41976779-41976780 on Build GRCh38
rs950791349 Chr.17: 41976603-41976603 on Build GRCh38
rs781832564 Chr.17: 41976825-41976825 on Build GRCh38
rs566144086 Chr.17: 41976549-41976549 on Build GRCh38
rs556413850 Chr.17: 41976569-41976569 on Build GRCh38
rs781907045 Chr.17: 41976814-41976814 on Build GRCh38
rs782633518 Chr.17: 41976718-41976718 on Build GRCh38
rs534841809 Chr.17: 41976620-41976620 on Build GRCh38
rs748657312 Chr.17: 41976728-41976728 on Build GRCh38
rs782068251 Chr.17: 41976805-41976805 on Build GRCh38
rs370498974 Chr.17: 41976619-41976619 on Build GRCh38
rs12950546 Chr.17: 41976473-41976473 on Build GRCh38
rs781995338 Chr.17: 41976769-41976769 on Build GRCh38
rs527995417 Chr.17: 41976645-41976645 on Build GRCh38
rs1054578328 Chr.17: 41976682-41976682 on Build GRCh38
rs782475516 Chr.17: 41976853-41976853 on Build GRCh38
rs529038216 Chr.17: 41976533-41976533 on Build GRCh38
rs567890037 Chr.17: 41976806-41976806 on Build GRCh38
rs782650996 Chr.17: 41976715-41976715 on Build GRCh38
rs782112991 Chr.17: 41976779-41976779 on Build GRCh38
rs151305433 Chr.17: 41976742-41976742 on Build GRCh38
rs1038951853 Chr.17: 41976901-41976901 on Build GRCh38
rs561748623 Chr.17: 41976491-41976491 on Build GRCh38

View Full Product Details