Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41989192-41989706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC7
Entrez Gene ID
7266
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
Gene Aliases
DJ11, DJC7, TPR2, TTC2
UniGene
Hs.500156
Gene Chromosome Location
Chr.17: 41976421-42021371 on Build GRCh38
Transcripts
NM_001144766.2 NM_003315.3 NR_029431.1 XM_011525167.2 XM_011525168.2 XM_011525169.2 XM_017024994.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6647418 c.580C>T p.R194* Chr.17: 41989547-41989547 on Build GRCh38
3937388 c.700G>A p.E234K Chr.17: 41989427-41989427 on Build GRCh38
6647416 c.637G>A p.E213K Chr.17: 41989490-41989490 on Build GRCh38
5164580 c.661_665delCAGTT p.Q221fs*10 Chr.17: 41989462-41989466 on Build GRCh38
4442784 c.623C>T p.T208I Chr.17: 41989534-41989534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782005741 Chr.17: 41989539-41989539 on Build GRCh38
rs782810453 Chr.17: 41989490-41989490 on Build GRCh38
rs782603115 Chr.17: 41989597-41989597 on Build GRCh38
rs992639622 Chr.17: 41989291-41989291 on Build GRCh38
rs969770223 Chr.17: 41989311-41989311 on Build GRCh38
rs1025539823 Chr.17: 41989675-41989675 on Build GRCh38
rs782008110 Chr.17: 41989428-41989428 on Build GRCh38
rs767065210 Chr.17: 41989586-41989586 on Build GRCh38
rs377703181 Chr.17: 41989488-41989488 on Build GRCh38
rs1025070817 Chr.17: 41989309-41989309 on Build GRCh38
rs782776979 Chr.17: 41989563-41989563 on Build GRCh38
rs752171755 Chr.17: 41989665-41989665 on Build GRCh38
rs781904749 Chr.17: 41989510-41989510 on Build GRCh38
rs548344455 Chr.17: 41989376-41989376 on Build GRCh38
rs142865844 Chr.17: 41989410-41989410 on Build GRCh38
rs200650446 Chr.17: 41989505-41989505 on Build GRCh38
rs782688087 Chr.17: 41989401-41989402 on Build GRCh38
rs942322955 Chr.17: 41989531-41989531 on Build GRCh38
rs545274496 Chr.17: 41989588-41989588 on Build GRCh38
rs551905449 Chr.17: 41989612-41989612 on Build GRCh38
rs782222229 Chr.17: 41989370-41989370 on Build GRCh38
rs782348244 Chr.17: 41989491-41989491 on Build GRCh38
rs782559875 Chr.17: 41989364-41989364 on Build GRCh38
rs148515578 Chr.17: 41989244-41989244 on Build GRCh38
rs782607233 Chr.17: 41989368-41989368 on Build GRCh38
rs782118074 Chr.17: 41989546-41989546 on Build GRCh38
rs781923026 Chr.17: 41989458-41989458 on Build GRCh38
rs73309739 Chr.17: 41989248-41989248 on Build GRCh38
rs199823135 Chr.17: 41989473-41989473 on Build GRCh38
rs980769751 Chr.17: 41989338-41989338 on Build GRCh38
rs377382456 Chr.17: 41989371-41989371 on Build GRCh38

View Full Product Details