Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41990178-41990584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC7
Entrez Gene ID
7266
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
Gene Aliases
DJ11, DJC7, TPR2, TTC2
UniGene
Hs.500156
Gene Chromosome Location
Chr.17: 41976421-42021371 on Build GRCh38
Transcripts
NM_001144766.2 NM_003315.3 NR_029431.1 XM_011525167.2 XM_011525168.2 XM_011525169.2 XM_017024994.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5386375 c.486C>T p.A162A Chr.17: 41990347-41990347 on Build GRCh38
5386374 c.516C>T p.A172A Chr.17: 41990347-41990347 on Build GRCh38
6840718 c.481-1G>T p.? Chr.17: 41990383-41990383 on Build GRCh38
4550453 c.470G>A p.R157H Chr.17: 41990363-41990363 on Build GRCh38
4499375 c.540C>T p.I180I Chr.17: 41990323-41990323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009591292 Chr.17: 41990226-41990226 on Build GRCh38
rs141300090 Chr.17: 41990410-41990410 on Build GRCh38
rs782591045 Chr.17: 41990407-41990407 on Build GRCh38
rs782547751 Chr.17: 41990423-41990423 on Build GRCh38
rs782626490 Chr.17: 41990291-41990291 on Build GRCh38
rs201143722 Chr.17: 41990339-41990339 on Build GRCh38
rs550910039 Chr.17: 41990372-41990372 on Build GRCh38
rs930940578 Chr.17: 41990526-41990526 on Build GRCh38
rs782180594 Chr.17: 41990304-41990304 on Build GRCh38
rs781895803 Chr.17: 41990364-41990364 on Build GRCh38
rs201948683 Chr.17: 41990213-41990213 on Build GRCh38
rs782556213 Chr.17: 41990216-41990216 on Build GRCh38
rs534192780 Chr.17: 41990562-41990562 on Build GRCh38
rs144747060 Chr.17: 41990529-41990529 on Build GRCh38
rs782138444 Chr.17: 41990374-41990375 on Build GRCh38
rs372160315 Chr.17: 41990224-41990224 on Build GRCh38
rs781914909 Chr.17: 41990349-41990349 on Build GRCh38
rs782739081 Chr.17: 41990389-41990389 on Build GRCh38
rs781798680 Chr.17: 41990221-41990221 on Build GRCh38
rs919695418 Chr.17: 41990445-41990445 on Build GRCh38
rs781849909 Chr.17: 41990399-41990399 on Build GRCh38
rs528069693 Chr.17: 41990327-41990327 on Build GRCh38
rs782376786 Chr.17: 41990334-41990334 on Build GRCh38
rs530533014 Chr.17: 41990204-41990204 on Build GRCh38
rs782082498 Chr.17: 41990346-41990346 on Build GRCh38
rs552231658 Chr.17: 41990276-41990276 on Build GRCh38
rs781804742 Chr.17: 41990505-41990505 on Build GRCh38
rs573314305 Chr.17: 41990533-41990533 on Build GRCh38
rs367980872 Chr.17: 41990510-41990510 on Build GRCh38
rs199951187 Chr.17: 41990359-41990359 on Build GRCh38
rs568420019 Chr.17: 41990351-41990351 on Build GRCh38
rs782387263 Chr.17: 41990205-41990205 on Build GRCh38
rs539578542 Chr.17: 41990504-41990504 on Build GRCh38
rs1127182 Chr.17: 41990284-41990284 on Build GRCh38
rs546150146 Chr.17: 41990338-41990338 on Build GRCh38
rs782351222 Chr.17: 41990285-41990285 on Build GRCh38
rs1021587551 Chr.17: 41990264-41990264 on Build GRCh38
rs782644045 Chr.17: 41990426-41990426 on Build GRCh38
rs145343720 Chr.17: 41990223-41990223 on Build GRCh38
rs782328990 Chr.17: 41990314-41990314 on Build GRCh38
rs201096021 Chr.17: 41990386-41990386 on Build GRCh38
rs781991604 Chr.17: 41990274-41990274 on Build GRCh38
rs370305071 Chr.17: 41990259-41990259 on Build GRCh38
rs782442751 Chr.17: 41990404-41990404 on Build GRCh38

View Full Product Details