Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42101833-42102326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX58
Entrez Gene ID
79132
Gene Name
DEXH-box helicase 58
Gene Aliases
D11LGP2, D11lgp2e, LGP2, RLR-3
UniGene
Hs.55918
Gene Chromosome Location
Chr.17: 42101404-42112733 on Build GRCh38
Transcripts
NM_024119.2 XM_017025059.1 XM_017025060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6303683 c.1880C>T p.S627L Chr.17: 42101918-42101918 on Build GRCh38
5021944 c.1852-10T>C p.? Chr.17: 42101956-42101956 on Build GRCh38
979401 c.1937G>A p.R646Q Chr.17: 42101861-42101861 on Build GRCh38
6280362 c.1906C>T p.R636C Chr.17: 42101892-42101892 on Build GRCh38
4532988 c.1938G>A p.R646R Chr.17: 42101860-42101860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145445108 Chr.17: 42102221-42102221 on Build GRCh38
rs368171285 Chr.17: 42102199-42102199 on Build GRCh38
rs782331940 Chr.17: 42102230-42102230 on Build GRCh38
rs908567097 Chr.17: 42102134-42102134 on Build GRCh38
rs9807056 Chr.17: 42102234-42102234 on Build GRCh38
rs188176098 Chr.17: 42102161-42102161 on Build GRCh38
rs369552739 Chr.17: 42102159-42102159 on Build GRCh38
rs575052653 Chr.17: 42101891-42101891 on Build GRCh38
rs368613039 Chr.17: 42101906-42101906 on Build GRCh38
rs892878434 Chr.17: 42101862-42101862 on Build GRCh38
rs781885173 Chr.17: 42102297-42102297 on Build GRCh38
rs139963886 Chr.17: 42102244-42102244 on Build GRCh38
rs782664534 Chr.17: 42101892-42101892 on Build GRCh38
rs146616923 Chr.17: 42102114-42102114 on Build GRCh38
rs149287727 Chr.17: 42102219-42102219 on Build GRCh38
rs781839679 Chr.17: 42101991-42101991 on Build GRCh38
rs374555157 Chr.17: 42101878-42101878 on Build GRCh38
rs782431551 Chr.17: 42101996-42101996 on Build GRCh38
rs782265248 Chr.17: 42102215-42102215 on Build GRCh38
rs542067143 Chr.17: 42101915-42101915 on Build GRCh38
rs982330070 Chr.17: 42102142-42102142 on Build GRCh38
rs563957116 Chr.17: 42101923-42101923 on Build GRCh38
rs782186420 Chr.17: 42101888-42101888 on Build GRCh38
rs531106041 Chr.17: 42101949-42101949 on Build GRCh38
rs782702878 Chr.17: 42102260-42102260 on Build GRCh38
rs963883153 Chr.17: 42102055-42102055 on Build GRCh38
rs781953225 Chr.17: 42102243-42102243 on Build GRCh38
rs782362099 Chr.17: 42101920-42101920 on Build GRCh38
rs781917445 Chr.17: 42101939-42101939 on Build GRCh38
rs782148354 Chr.17: 42101870-42101870 on Build GRCh38
rs77876788 Chr.17: 42102047-42102047 on Build GRCh38
rs1027270789 Chr.17: 42102294-42102294 on Build GRCh38
rs952940327 Chr.17: 42102121-42102121 on Build GRCh38
rs782080168 Chr.17: 42101931-42101931 on Build GRCh38
rs144364815 Chr.17: 42101974-42101974 on Build GRCh38
rs996683567 Chr.17: 42102270-42102270 on Build GRCh38
rs781996971 Chr.17: 42101930-42101930 on Build GRCh38
rs563033779 Chr.17: 42102012-42102012 on Build GRCh38
rs782723489 Chr.17: 42101978-42101978 on Build GRCh38
rs781876679 Chr.17: 42101852-42101852 on Build GRCh38
rs142641565 Chr.17: 42102267-42102267 on Build GRCh38
rs899751053 Chr.17: 42102249-42102249 on Build GRCh38
rs782383661 Chr.17: 42102235-42102235 on Build GRCh38
rs111241943 Chr.17: 42101921-42101921 on Build GRCh38
rs1027032874 Chr.17: 42102080-42102080 on Build GRCh38
rs782357714 Chr.17: 42101936-42101936 on Build GRCh38
rs782145726 Chr.17: 42102290-42102290 on Build GRCh38
rs782712379 Chr.17: 42102196-42102196 on Build GRCh38
rs768087126 Chr.17: 42102057-42102057 on Build GRCh38
rs982777511 Chr.17: 42102133-42102133 on Build GRCh38
rs139876577 Chr.17: 42101956-42101956 on Build GRCh38
rs970955287 Chr.17: 42102139-42102139 on Build GRCh38
rs947180513 Chr.17: 42102218-42102218 on Build GRCh38
rs782471953 Chr.17: 42101857-42101857 on Build GRCh38
rs782807092 Chr.17: 42101952-42101952 on Build GRCh38
rs369043122 Chr.17: 42102003-42102005 on Build GRCh38
rs782621199 Chr.17: 42102220-42102220 on Build GRCh38
rs113308191 Chr.17: 42102198-42102198 on Build GRCh38

View Full Product Details