Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42188987-42189481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHDC
Entrez Gene ID
84514
Gene Name
GH3 domain containing
Gene Aliases
D11LGP1, LGP1
UniGene
Hs.38039
Gene Chromosome Location
Chr.17: 42189087-42194532 on Build GRCh38
Transcripts
NM_001142623.1 NM_032484.4

SNP Detail

SNP ID Chromosome Location
rs1047299 Chr.17: 42189170-42189170 on Build GRCh38
rs116050394 Chr.17: 42189134-42189134 on Build GRCh38
rs923804901 Chr.17: 42189444-42189444 on Build GRCh38
rs999036996 Chr.17: 42189411-42189411 on Build GRCh38
rs1017070986 Chr.17: 42189461-42189461 on Build GRCh38
rs556458375 Chr.17: 42189196-42189196 on Build GRCh38
rs916490209 Chr.17: 42189101-42189101 on Build GRCh38
rs372195675 Chr.17: 42189317-42189317 on Build GRCh38
rs909955698 Chr.17: 42189160-42189160 on Build GRCh38
rs71377267 Chr.17: 42189435-42189435 on Build GRCh38
rs1028563554 Chr.17: 42189443-42189443 on Build GRCh38
rs576792887 Chr.17: 42189081-42189081 on Build GRCh38
rs1056817349 Chr.17: 42189255-42189255 on Build GRCh38
rs533658781 Chr.17: 42189007-42189007 on Build GRCh38
rs879618908 Chr.17: 42189414-42189414 on Build GRCh38
rs368316764 Chr.17: 42189026-42189026 on Build GRCh38
rs868708567 Chr.17: 42189209-42189209 on Build GRCh38
rs768133241 Chr.17: 42189164-42189164 on Build GRCh38
rs972267850 Chr.17: 42189185-42189185 on Build GRCh38
rs190197518 Chr.17: 42189146-42189146 on Build GRCh38
rs527375198 Chr.17: 42189434-42189434 on Build GRCh38
rs1034759537 Chr.17: 42189301-42189301 on Build GRCh38
rs956523504 Chr.17: 42189459-42189459 on Build GRCh38
rs1016651388 Chr.17: 42189044-42189044 on Build GRCh38
rs1035062791 Chr.17: 42189060-42189060 on Build GRCh38
rs753457110 Chr.17: 42189237-42189237 on Build GRCh38
rs1053663962 Chr.17: 42189422-42189422 on Build GRCh38
rs931982940 Chr.17: 42189153-42189153 on Build GRCh38
rs557792316 Chr.17: 42189341-42189342 on Build GRCh38
rs749667667 Chr.17: 42189423-42189423 on Build GRCh38
rs566963294 Chr.17: 42189120-42189120 on Build GRCh38
rs79081642 Chr.17: 42189066-42189066 on Build GRCh38
rs762449625 Chr.17: 42189162-42189162 on Build GRCh38
rs867901643 Chr.17: 42189077-42189077 on Build GRCh38
rs544490077 Chr.17: 42189167-42189167 on Build GRCh38
rs1005550161 Chr.17: 42189447-42189447 on Build GRCh38
rs763786453 Chr.17: 42189135-42189135 on Build GRCh38
rs544608507 Chr.17: 42189330-42189330 on Build GRCh38
rs533110626 Chr.17: 42189316-42189316 on Build GRCh38
rs368356722 Chr.17: 42189141-42189141 on Build GRCh38
rs960683650 Chr.17: 42189310-42189310 on Build GRCh38
rs967920051 Chr.17: 42189380-42189380 on Build GRCh38
rs1042819311 Chr.17: 42189378-42189378 on Build GRCh38
rs1006361492 Chr.17: 42189200-42189200 on Build GRCh38
rs961087684 Chr.17: 42189047-42189047 on Build GRCh38
rs887999712 Chr.17: 42189193-42189193 on Build GRCh38
rs559776586 Chr.17: 42189336-42189336 on Build GRCh38
rs754450861 Chr.17: 42189263-42189263 on Build GRCh38
rs972426929 Chr.17: 42189056-42189056 on Build GRCh38
rs3210456 Chr.17: 42189264-42189264 on Build GRCh38
rs946547751 Chr.17: 42189107-42189107 on Build GRCh38
rs558233857 Chr.17: 42189117-42189117 on Build GRCh38
rs374301009 Chr.17: 42189347-42189348 on Build GRCh38

View Full Product Details