Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42189811-42190316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHDC
Entrez Gene ID
84514
Gene Name
GH3 domain containing
Gene Aliases
D11LGP1, LGP1
UniGene
Hs.38039
Gene Chromosome Location
Chr.17: 42189087-42194532 on Build GRCh38
Transcripts
NM_001142623.1 NM_032484.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4878943 c.1337C>A p.P446Q Chr.17: 42190311-42190311 on Build GRCh38
5605742 c.1344G>A p.L448L Chr.17: 42190215-42190215 on Build GRCh38
1640696 c.1298C>T p.A433V Chr.17: 42190261-42190261 on Build GRCh38
3188508 c.1345G>A p.G449R Chr.17: 42190303-42190303 on Build GRCh38
6670825 c.1332G>A p.A444A Chr.17: 42190227-42190227 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919506860 Chr.17: 42190090-42190090 on Build GRCh38
rs878959306 Chr.17: 42190058-42190058 on Build GRCh38
rs972018260 Chr.17: 42190071-42190071 on Build GRCh38
rs762253755 Chr.17: 42190227-42190227 on Build GRCh38
rs948882539 Chr.17: 42190164-42190164 on Build GRCh38
rs200095635 Chr.17: 42189876-42189876 on Build GRCh38
rs745361716 Chr.17: 42190246-42190246 on Build GRCh38
rs760780876 Chr.17: 42190174-42190174 on Build GRCh38
rs750320133 Chr.17: 42190200-42190201 on Build GRCh38
rs766677663 Chr.17: 42190188-42190188 on Build GRCh38
rs542670641 Chr.17: 42189840-42189840 on Build GRCh38
rs762879640 Chr.17: 42190150-42190150 on Build GRCh38
rs758277837 Chr.17: 42190224-42190235 on Build GRCh38
rs763830135 Chr.17: 42190156-42190156 on Build GRCh38
rs773999315 Chr.17: 42190287-42190287 on Build GRCh38
rs571976574 Chr.17: 42190245-42190245 on Build GRCh38
rs993480688 Chr.17: 42190108-42190108 on Build GRCh38
rs756870241 Chr.17: 42190085-42190085 on Build GRCh38
rs780110374 Chr.17: 42190230-42190231 on Build GRCh38
rs767087592 Chr.17: 42190135-42190135 on Build GRCh38
rs780950850 Chr.17: 42190243-42190243 on Build GRCh38
rs963362809 Chr.17: 42190055-42190055 on Build GRCh38
rs765259473 Chr.17: 42189846-42189846 on Build GRCh38
rs180989188 Chr.17: 42190206-42190206 on Build GRCh38
rs769224655 Chr.17: 42190258-42190258 on Build GRCh38
rs753200015 Chr.17: 42190220-42190220 on Build GRCh38
rs768889950 Chr.17: 42189956-42189956 on Build GRCh38
rs768623749 Chr.17: 42190268-42190268 on Build GRCh38
rs753848898 Chr.17: 42190192-42190192 on Build GRCh38
rs567660637 Chr.17: 42190136-42190136 on Build GRCh38
rs897173930 Chr.17: 42190086-42190086 on Build GRCh38
rs145507063 Chr.17: 42190229-42190229 on Build GRCh38
rs1049885367 Chr.17: 42189893-42189893 on Build GRCh38
rs79309130 Chr.17: 42189891-42189891 on Build GRCh38
rs770286745 Chr.17: 42189835-42189835 on Build GRCh38
rs1014049986 Chr.17: 42190151-42190151 on Build GRCh38
rs986215636 Chr.17: 42190008-42190008 on Build GRCh38
rs140351409 Chr.17: 42190193-42190193 on Build GRCh38
rs755422497 Chr.17: 42189905-42189905 on Build GRCh38
rs896232916 Chr.17: 42189852-42189852 on Build GRCh38
rs774165636 Chr.17: 42190160-42190160 on Build GRCh38
rs200047467 Chr.17: 42189937-42189937 on Build GRCh38
rs756837467 Chr.17: 42190242-42190242 on Build GRCh38
rs953325004 Chr.17: 42189980-42189980 on Build GRCh38
rs565115379 Chr.17: 42189924-42189924 on Build GRCh38
rs369509799 Chr.17: 42190241-42190241 on Build GRCh38
rs577208365 Chr.17: 42189862-42189862 on Build GRCh38
rs748519132 Chr.17: 42189935-42189935 on Build GRCh38
rs778298347 Chr.17: 42189952-42189952 on Build GRCh38
rs767719177 Chr.17: 42189879-42189879 on Build GRCh38
rs560524000 Chr.17: 42189973-42189973 on Build GRCh38
rs138138681 Chr.17: 42190261-42190261 on Build GRCh38
rs553271264 Chr.17: 42190148-42190148 on Build GRCh38
rs1030658692 Chr.17: 42190253-42190253 on Build GRCh38
rs527701340 Chr.17: 42190033-42190033 on Build GRCh38
rs867910983 Chr.17: 42189903-42189903 on Build GRCh38
rs780130784 Chr.17: 42189872-42189872 on Build GRCh38
rs554513459 Chr.17: 42190212-42190212 on Build GRCh38
rs758769861 Chr.17: 42189942-42189942 on Build GRCh38
rs1032918421 Chr.17: 42190260-42190260 on Build GRCh38
rs377225863 Chr.17: 42190134-42190134 on Build GRCh38
rs115952579 Chr.17: 42189890-42189890 on Build GRCh38
rs774992745 Chr.17: 42190259-42190259 on Build GRCh38
rs141532903 Chr.17: 42190093-42190093 on Build GRCh38
rs753994218 Chr.17: 42189896-42189896 on Build GRCh38
rs1016282366 Chr.17: 42190019-42190019 on Build GRCh38
rs941568001 Chr.17: 42189965-42189965 on Build GRCh38
rs201134987 Chr.17: 42190228-42190228 on Build GRCh38
rs138550464 Chr.17: 42189971-42189971 on Build GRCh38

View Full Product Details