Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42192579-42193082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHDC
Entrez Gene ID
84514
Gene Name
GH3 domain containing
Gene Aliases
D11LGP1, LGP1
UniGene
Hs.38039
Gene Chromosome Location
Chr.17: 42189087-42194532 on Build GRCh38
Transcripts
NM_001142623.1 NM_032484.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1383331 c.386A>G p.Q129R Chr.17: 42192907-42192907 on Build GRCh38
6867255 c.399G>T p.L133L Chr.17: 42192731-42192731 on Build GRCh38
4426245 c.351G>A p.P117P Chr.17: 42192942-42192942 on Build GRCh38
6670829 c.449G>A p.R150H Chr.17: 42192681-42192681 on Build GRCh38
4066530 c.357A>G p.S119S Chr.17: 42192936-42192936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747532592 Chr.17: 42192993-42192993 on Build GRCh38
rs778785883 Chr.17: 42192606-42192606 on Build GRCh38
rs780206014 Chr.17: 42192718-42192718 on Build GRCh38
rs749384218 Chr.17: 42192654-42192654 on Build GRCh38
rs766006094 Chr.17: 42192666-42192666 on Build GRCh38
rs778188818 Chr.17: 42192973-42192973 on Build GRCh38
rs376526384 Chr.17: 42192684-42192684 on Build GRCh38
rs774654444 Chr.17: 42192662-42192662 on Build GRCh38
rs374619710 Chr.17: 42192651-42192651 on Build GRCh38
rs764182574 Chr.17: 42192946-42192946 on Build GRCh38
rs140008451 Chr.17: 42193039-42193039 on Build GRCh38
rs777729298 Chr.17: 42192643-42192643 on Build GRCh38
rs772619124 Chr.17: 42192858-42192858 on Build GRCh38
rs777856639 Chr.17: 42193048-42193048 on Build GRCh38
rs150176372 Chr.17: 42192664-42192664 on Build GRCh38
rs945538598 Chr.17: 42192610-42192610 on Build GRCh38
rs748624084 Chr.17: 42192772-42192772 on Build GRCh38
rs906898416 Chr.17: 42193001-42193001 on Build GRCh38
rs754725963 Chr.17: 42192766-42192770 on Build GRCh38
rs374551993 Chr.17: 42193030-42193030 on Build GRCh38
rs755112603 Chr.17: 42192855-42192855 on Build GRCh38
rs199638076 Chr.17: 42192964-42192964 on Build GRCh38
rs765512106 Chr.17: 42193040-42193040 on Build GRCh38
rs780027517 Chr.17: 42192693-42192693 on Build GRCh38
rs539395657 Chr.17: 42192609-42192609 on Build GRCh38
rs61729323 Chr.17: 42192625-42192625 on Build GRCh38
rs772564297 Chr.17: 42192968-42192968 on Build GRCh38
rs777678073 Chr.17: 42192688-42192688 on Build GRCh38
rs564400023 Chr.17: 42192672-42192672 on Build GRCh38
rs779082754 Chr.17: 42192771-42192771 on Build GRCh38
rs748287081 Chr.17: 42192966-42192966 on Build GRCh38
rs182734817 Chr.17: 42193013-42193013 on Build GRCh38
rs751446435 Chr.17: 42192706-42192706 on Build GRCh38
rs756389635 Chr.17: 42192711-42192711 on Build GRCh38
rs753387151 Chr.17: 42192671-42192671 on Build GRCh38
rs912603257 Chr.17: 42192646-42192646 on Build GRCh38
rs1015357478 Chr.17: 42192837-42192837 on Build GRCh38
rs756933638 Chr.17: 42192952-42192952 on Build GRCh38
rs375798442 Chr.17: 42193045-42193045 on Build GRCh38
rs559821717 Chr.17: 42192795-42192795 on Build GRCh38
rs763699771 Chr.17: 42193020-42193020 on Build GRCh38
rs757818828 Chr.17: 42193053-42193053 on Build GRCh38
rs767320668 Chr.17: 42192872-42192888 on Build GRCh38
rs577201246 Chr.17: 42192665-42192665 on Build GRCh38
rs750281504 Chr.17: 42193037-42193037 on Build GRCh38
rs762959016 Chr.17: 42192943-42192943 on Build GRCh38
rs202045196 Chr.17: 42192940-42192940 on Build GRCh38
rs752923813 Chr.17: 42193042-42193042 on Build GRCh38
rs373811508 Chr.17: 42192785-42192785 on Build GRCh38
rs149568450 Chr.17: 42193012-42193012 on Build GRCh38
rs959337898 Chr.17: 42193017-42193017 on Build GRCh38
rs775123287 Chr.17: 42193014-42193014 on Build GRCh38
rs774372969 Chr.17: 42193022-42193022 on Build GRCh38
rs542452469 Chr.17: 42192903-42192903 on Build GRCh38
rs371516105 Chr.17: 42192942-42192942 on Build GRCh38
rs989121562 Chr.17: 42193030-42193033 on Build GRCh38
rs138123618 Chr.17: 42192682-42192682 on Build GRCh38
rs781054734 Chr.17: 42192956-42192956 on Build GRCh38
rs769200489 Chr.17: 42192656-42192656 on Build GRCh38
rs773554817 Chr.17: 42192873-42192873 on Build GRCh38
rs376784170 Chr.17: 42192631-42192631 on Build GRCh38
rs755268459 Chr.17: 42192962-42192962 on Build GRCh38
rs747437313 Chr.17: 42193052-42193052 on Build GRCh38
rs200716113 Chr.17: 42192832-42192832 on Build GRCh38
rs370935274 Chr.17: 42192645-42192645 on Build GRCh38
rs1047186842 Chr.17: 42192787-42192787 on Build GRCh38
rs771333746 Chr.17: 42192881-42192881 on Build GRCh38
rs761097305 Chr.17: 42192877-42192877 on Build GRCh38
rs768561172 Chr.17: 42192760-42192760 on Build GRCh38
rs751395178 Chr.17: 42192949-42192949 on Build GRCh38
rs761831641 Chr.17: 42193025-42193025 on Build GRCh38
rs762616981 Chr.17: 42193018-42193018 on Build GRCh38
rs200346068 Chr.17: 42192907-42192907 on Build GRCh38
rs749530572 Chr.17: 42192757-42192757 on Build GRCh38
rs199625401 Chr.17: 42192681-42192681 on Build GRCh38
rs778837039 Chr.17: 42192963-42192963 on Build GRCh38
rs776094138 Chr.17: 42192886-42192886 on Build GRCh38
rs994843481 Chr.17: 42192708-42192708 on Build GRCh38
rs1049613370 Chr.17: 42192751-42192751 on Build GRCh38
rs373251396 Chr.17: 42192602-42192602 on Build GRCh38
rs753957466 Chr.17: 42192957-42192957 on Build GRCh38

View Full Product Details