Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42493894-42494437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0A1
Entrez Gene ID
535
Gene Name
ATPase H+ transporting V0 subunit a1
Gene Aliases
ATP6N1, ATP6N1A, Stv1, VPP1, Vph1, a1
UniGene
Hs.463074
Gene Chromosome Location
Chr.17: 42458844-42522579 on Build GRCh38
Transcripts
NM_001130020.1 NM_001130021.1 NM_005177.3 XM_005257459.1 XM_005257461.1 XM_005257463.2 XM_011524908.1 XM_017024766.1 XM_017024767.1 XM_017024768.1 XM_017024769.1 XM_017024770.1 XM_017024771.1 XM_017024772.1 XM_017024773.1 XM_017024774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1383375 c.1175C>T p.A392V Chr.17: 42494334-42494334 on Build GRCh38
5678904 c.1265G>T p.W422L Chr.17: 42494424-42494424 on Build GRCh38
385866 c.1267A>G p.M423V Chr.17: 42494426-42494426 on Build GRCh38
5828532 c.1196C>T p.A399V Chr.17: 42494334-42494334 on Build GRCh38
3402907 c.1179G>A p.P393P Chr.17: 42494338-42494338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039212784 Chr.17: 42493918-42493918 on Build GRCh38
rs758672070 Chr.17: 42494353-42494353 on Build GRCh38
rs111923187 Chr.17: 42493981-42493981 on Build GRCh38
rs955976019 Chr.17: 42494086-42494086 on Build GRCh38
rs777658942 Chr.17: 42494365-42494365 on Build GRCh38
rs548912123 Chr.17: 42494010-42494010 on Build GRCh38
rs62075826 Chr.17: 42493941-42493941 on Build GRCh38
rs959445968 Chr.17: 42494069-42494069 on Build GRCh38
rs200532770 Chr.17: 42494411-42494411 on Build GRCh38
rs778893425 Chr.17: 42494246-42494246 on Build GRCh38
rs755938027 Chr.17: 42493991-42493991 on Build GRCh38
rs186620688 Chr.17: 42493979-42493979 on Build GRCh38
rs773589122 Chr.17: 42494299-42494299 on Build GRCh38
rs941015310 Chr.17: 42494227-42494227 on Build GRCh38
rs1012292207 Chr.17: 42493983-42493983 on Build GRCh38
rs751148740 Chr.17: 42494308-42494308 on Build GRCh38
rs559930661 Chr.17: 42493971-42493971 on Build GRCh38
rs764696555 Chr.17: 42494352-42494352 on Build GRCh38
rs897438350 Chr.17: 42493964-42493964 on Build GRCh38
rs142629560 Chr.17: 42494338-42494338 on Build GRCh38
rs966655201 Chr.17: 42494137-42494137 on Build GRCh38
rs989049876 Chr.17: 42494088-42494088 on Build GRCh38
rs55666448 Chr.17: 42494344-42494344 on Build GRCh38
rs1001606521 Chr.17: 42494048-42494048 on Build GRCh38
rs1022598344 Chr.17: 42494129-42494129 on Build GRCh38
rs570187892 Chr.17: 42494117-42494117 on Build GRCh38
rs754550279 Chr.17: 42494345-42494345 on Build GRCh38
rs369271937 Chr.17: 42494360-42494360 on Build GRCh38
rs529908817 Chr.17: 42494020-42494020 on Build GRCh38
rs4796591 Chr.17: 42494039-42494039 on Build GRCh38
rs995080049 Chr.17: 42493956-42493956 on Build GRCh38
rs919744749 Chr.17: 42494251-42494251 on Build GRCh38
rs368502073 Chr.17: 42494389-42494389 on Build GRCh38
rs761481524 Chr.17: 42493924-42493924 on Build GRCh38
rs552868127 Chr.17: 42494244-42494244 on Build GRCh38
rs896558366 Chr.17: 42493982-42493982 on Build GRCh38
rs200117905 Chr.17: 42494302-42494302 on Build GRCh38
rs753885814 Chr.17: 42494205-42494205 on Build GRCh38
rs775061772 Chr.17: 42494286-42494286 on Build GRCh38
rs754798607 Chr.17: 42494239-42494239 on Build GRCh38
rs765960354 Chr.17: 42494319-42494319 on Build GRCh38
rs551760051 Chr.17: 42494116-42494116 on Build GRCh38
rs200130950 Chr.17: 42494288-42494288 on Build GRCh38
rs1012841623 Chr.17: 42494250-42494250 on Build GRCh38
rs536611226 Chr.17: 42494082-42494082 on Build GRCh38
rs777458553 Chr.17: 42494371-42494371 on Build GRCh38
rs767798503 Chr.17: 42494291-42494291 on Build GRCh38
rs149445720 Chr.17: 42494240-42494240 on Build GRCh38
rs910982703 Chr.17: 42494156-42494156 on Build GRCh38
rs1033457223 Chr.17: 42494051-42494051 on Build GRCh38
rs930265311 Chr.17: 42494244-42494245 on Build GRCh38
rs368586952 Chr.17: 42494234-42494234 on Build GRCh38

View Full Product Details