Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42561976-42562501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COASY
Entrez Gene ID
80347
Gene Name
Coenzyme A synthase
Gene Aliases
DPCK, NBIA6, NBP, PPAT, UKR1, pOV-2
UniGene
Hs.296422
Gene Chromosome Location
Chr.17: 42562074-42566281 on Build GRCh38
Transcripts
NM_001042529.2 NM_001042532.3 NM_025233.6 XM_006722116.3 XM_011525300.1 XM_017025167.1 XM_017025168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5893915 c.70C>T p.P24S Chr.17: 42562481-42562481 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370095840 Chr.17: 42562450-42562450 on Build GRCh38
rs763770736 Chr.17: 42562362-42562362 on Build GRCh38
rs1024942105 Chr.17: 42562157-42562157 on Build GRCh38
rs763334155 Chr.17: 42562403-42562403 on Build GRCh38
rs548663586 Chr.17: 42562071-42562071 on Build GRCh38
rs995662211 Chr.17: 42562087-42562087 on Build GRCh38
rs1025668508 Chr.17: 42562364-42562364 on Build GRCh38
rs887287458 Chr.17: 42562354-42562354 on Build GRCh38
rs1013800831 Chr.17: 42562359-42562359 on Build GRCh38
rs761758708 Chr.17: 42562028-42562028 on Build GRCh38
rs756607674 Chr.17: 42562247-42562247 on Build GRCh38
rs771314312 Chr.17: 42562040-42562040 on Build GRCh38
rs565751461 Chr.17: 42562049-42562049 on Build GRCh38
rs776636915 Chr.17: 42562389-42562389 on Build GRCh38
rs986258719 Chr.17: 42562153-42562153 on Build GRCh38
rs980857213 Chr.17: 42562204-42562204 on Build GRCh38
rs780081141 Chr.17: 42562375-42562375 on Build GRCh38
rs563192501 Chr.17: 42561995-42561995 on Build GRCh38
rs1047254585 Chr.17: 42562352-42562352 on Build GRCh38
rs912190276 Chr.17: 42562249-42562249 on Build GRCh38
rs754940764 Chr.17: 42562442-42562442 on Build GRCh38
rs927109998 Chr.17: 42562009-42562009 on Build GRCh38
rs956962789 Chr.17: 42562290-42562290 on Build GRCh38
rs749835853 Chr.17: 42562463-42562463 on Build GRCh38
rs767186759 Chr.17: 42562422-42562422 on Build GRCh38
rs1056718856 Chr.17: 42562171-42562171 on Build GRCh38
rs1023793398 Chr.17: 42562101-42562101 on Build GRCh38
rs749946116 Chr.17: 42562370-42562370 on Build GRCh38
rs191068578 Chr.17: 42562460-42562460 on Build GRCh38
rs1038272540 Chr.17: 42562070-42562070 on Build GRCh38
rs1031294805 Chr.17: 42562133-42562133 on Build GRCh38
rs1054343625 Chr.17: 42562035-42562035 on Build GRCh38
rs745969255 Chr.17: 42562461-42562461 on Build GRCh38
rs629861 Chr.17: 42561996-42561996 on Build GRCh38
rs764492939 Chr.17: 42562414-42562414 on Build GRCh38
rs183626516 Chr.17: 42562152-42562152 on Build GRCh38
rs753779026 Chr.17: 42562441-42562441 on Build GRCh38
rs997751193 Chr.17: 42562316-42562316 on Build GRCh38
rs537541340 Chr.17: 42562228-42562228 on Build GRCh38
rs1000543807 Chr.17: 42562252-42562252 on Build GRCh38
rs373319332 Chr.17: 42562118-42562118 on Build GRCh38
rs887749218 Chr.17: 42562044-42562044 on Build GRCh38
rs749098593 Chr.17: 42562385-42562385 on Build GRCh38
rs538122894 Chr.17: 42562013-42562013 on Build GRCh38
rs749914439 Chr.17: 42562425-42562425 on Build GRCh38
rs571030856 Chr.17: 42562233-42562233 on Build GRCh38
rs545863395 Chr.17: 42562394-42562394 on Build GRCh38
rs979090663 Chr.17: 42562114-42562114 on Build GRCh38
rs771627280 Chr.17: 42562472-42562472 on Build GRCh38
rs529996015 Chr.17: 42562066-42562066 on Build GRCh38
rs541507808 Chr.17: 42562332-42562332 on Build GRCh38
rs745907150 Chr.17: 42562392-42562392 on Build GRCh38
rs376677692 Chr.17: 42562374-42562374 on Build GRCh38
rs942073703 Chr.17: 42562156-42562156 on Build GRCh38
rs1032408401 Chr.17: 42562397-42562397 on Build GRCh38
rs1025835785 Chr.17: 42562138-42562138 on Build GRCh38
rs986699620 Chr.17: 42562363-42562363 on Build GRCh38
rs953593832 Chr.17: 42562357-42562357 on Build GRCh38
rs1036312813 Chr.17: 42562160-42562160 on Build GRCh38
rs528235676 Chr.17: 42562447-42562447 on Build GRCh38
rs898275264 Chr.17: 42562330-42562330 on Build GRCh38
rs1039861377 Chr.17: 42562079-42562079 on Build GRCh38
rs945676227 Chr.17: 42562041-42562041 on Build GRCh38
rs923441494 Chr.17: 42562142-42562142 on Build GRCh38
rs919235639 Chr.17: 42562424-42562424 on Build GRCh38
rs769149059 Chr.17: 42562465-42562465 on Build GRCh38
rs990004872 Chr.17: 42562229-42562229 on Build GRCh38
rs967730364 Chr.17: 42562102-42562102 on Build GRCh38
rs925374023 Chr.17: 42562206-42562206 on Build GRCh38
rs774636354 Chr.17: 42562467-42562467 on Build GRCh38
rs766034518 Chr.17: 42562476-42562476 on Build GRCh38
rs1012476599 Chr.17: 42562208-42562208 on Build GRCh38
rs986711026 Chr.17: 42562479-42562479 on Build GRCh38
rs913180163 Chr.17: 42562036-42562036 on Build GRCh38
rs898643107 Chr.17: 42562086-42562086 on Build GRCh38
rs772880365 Chr.17: 42562474-42562474 on Build GRCh38
rs545720982 Chr.17: 42561997-42561997 on Build GRCh38
rs767648742 Chr.17: 42562167-42562167 on Build GRCh38
rs760282616 Chr.17: 42562475-42562475 on Build GRCh38
rs574491277 Chr.17: 42562327-42562327 on Build GRCh38
rs563959731 Chr.17: 42562400-42562400 on Build GRCh38
rs1039426008 Chr.17: 42562312-42562312 on Build GRCh38
rs191362307 Chr.17: 42562043-42562043 on Build GRCh38
rs112423989 Chr.17: 42562286-42562286 on Build GRCh38
rs750526480 Chr.17: 42562182-42562182 on Build GRCh38
rs766448525 Chr.17: 42562440-42562440 on Build GRCh38
rs1050821401 Chr.17: 42562046-42562046 on Build GRCh38
rs932046994 Chr.17: 42562003-42562003 on Build GRCh38
rs758442645 Chr.17: 42562472-42562472 on Build GRCh38
rs563924473 Chr.17: 42562002-42562002 on Build GRCh38
rs575280023 Chr.17: 42562366-42562366 on Build GRCh38
rs762180459 Chr.17: 42562471-42562471 on Build GRCh38
rs377332971 Chr.17: 42562230-42562230 on Build GRCh38
rs945978440 Chr.17: 42562236-42562236 on Build GRCh38
rs778161660 Chr.17: 42562387-42562387 on Build GRCh38
rs1013535115 Chr.17: 42562154-42562154 on Build GRCh38
rs188473326 Chr.17: 42562338-42562338 on Build GRCh38
rs761439725 Chr.17: 42562419-42562419 on Build GRCh38
rs1006605203 Chr.17: 42562055-42562055 on Build GRCh38
rs774748328 Chr.17: 42562417-42562417 on Build GRCh38
rs553664611 Chr.17: 42562314-42562314 on Build GRCh38
rs753365072 Chr.17: 42562478-42562478 on Build GRCh38
rs766977029 Chr.17: 42562368-42562368 on Build GRCh38
rs749726420 Chr.17: 42562019-42562019 on Build GRCh38
rs932070277 Chr.17: 42562148-42562148 on Build GRCh38
rs755619184 Chr.17: 42562429-42562429 on Build GRCh38
rs769818125 Chr.17: 42562462-42562462 on Build GRCh38
rs747242800 Chr.17: 42562388-42562388 on Build GRCh38
rs754778428 Chr.17: 42562386-42562386 on Build GRCh38
rs570094654 Chr.17: 42562165-42562165 on Build GRCh38
rs993995584 Chr.17: 42562073-42562073 on Build GRCh38

View Full Product Details