Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31641992-31642486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG6
Entrez Gene ID
7917
Gene Name
BCL2 associated athanogene 6
Gene Aliases
BAG-6, BAT3, D6S52E, G3
UniGene
Hs.440900
Gene Chromosome Location
Chr.6: 31639028-31660900 on Build GRCh38
Transcripts
NM_001098534.1 NM_001199697.1 NM_001199698.1 NM_004639.3 NM_080702.2 NM_080703.2 XM_011514892.2 XM_017011275.1 XM_017011276.1 XM_017011277.1 XM_017011278.1 XM_017011279.1 XM_017011280.1 XM_017011281.1 XM_017011282.1 XM_017011283.1 XM_017011284.1 XM_017011285.1 XM_017011286.1 XM_017011287.1 XM_017011288.1 XM_017011289.1 XM_017011290.1 XM_017011291.1 XM_017011292.1 XM_017011293.1 XM_017011294.1 XM_017011295.1 XM_017011296.1 XM_017011297.1 XM_017011298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1723861 c.2330_2331CC>TT p.A777V Chr.6: 31642116-31642117 on Build GRCh38
7448967 c.2089C>T p.P697S Chr.6: 31642358-31642358 on Build GRCh38
4918204 c.1998G>T p.Q666H Chr.6: 31642341-31642341 on Build GRCh38
6586731 c.2234C>T p.S745F Chr.6: 31642213-31642213 on Build GRCh38
5835180 c.2127_2128insT p.L710fs*9 Chr.6: 31642211-31642212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016803218 Chr.6: 31642229-31642229 on Build GRCh38
rs776039684 Chr.6: 31642212-31642212 on Build GRCh38
rs766613142 Chr.6: 31642383-31642383 on Build GRCh38
rs757298004 Chr.6: 31642096-31642096 on Build GRCh38
rs148186069 Chr.6: 31642305-31642305 on Build GRCh38
rs753986543 Chr.6: 31642386-31642386 on Build GRCh38
rs3034254 Chr.6: 31642416-31642418 on Build GRCh38
rs747942520 Chr.6: 31642140-31642140 on Build GRCh38
rs764484869 Chr.6: 31642227-31642227 on Build GRCh38
rs767765376 Chr.6: 31642349-31642349 on Build GRCh38
rs887039270 Chr.6: 31642141-31642141 on Build GRCh38
rs745957031 Chr.6: 31642115-31642115 on Build GRCh38
rs770282825 Chr.6: 31642208-31642208 on Build GRCh38
rs1046316000 Chr.6: 31642105-31642105 on Build GRCh38
rs781383370 Chr.6: 31642099-31642099 on Build GRCh38
rs949007956 Chr.6: 31642412-31642412 on Build GRCh38
rs758970230 Chr.6: 31642207-31642207 on Build GRCh38
rs536599930 Chr.6: 31642026-31642026 on Build GRCh38
rs534596230 Chr.6: 31642263-31642263 on Build GRCh38
rs1040906155 Chr.6: 31642421-31642421 on Build GRCh38
rs770509349 Chr.6: 31642202-31642202 on Build GRCh38
rs910789449 Chr.6: 31642344-31642344 on Build GRCh38
rs775947992 Chr.6: 31642331-31642331 on Build GRCh38
rs1046076945 Chr.6: 31642458-31642458 on Build GRCh38
rs1013263770 Chr.6: 31642104-31642104 on Build GRCh38
rs756204588 Chr.6: 31642271-31642271 on Build GRCh38
rs746828364 Chr.6: 31642307-31642307 on Build GRCh38
rs369959235 Chr.6: 31642245-31642245 on Build GRCh38
rs780725341 Chr.6: 31642315-31642315 on Build GRCh38
rs542084968 Chr.6: 31642335-31642335 on Build GRCh38
rs920101732 Chr.6: 31642324-31642324 on Build GRCh38
rs878917781 Chr.6: 31642138-31642138 on Build GRCh38
rs931068875 Chr.6: 31642029-31642029 on Build GRCh38
rs778728908 Chr.6: 31642119-31642119 on Build GRCh38
rs200073459 Chr.6: 31642332-31642332 on Build GRCh38
rs770546772 Chr.6: 31642312-31642312 on Build GRCh38
rs191917970 Chr.6: 31642459-31642459 on Build GRCh38
rs747605981 Chr.6: 31642328-31642328 on Build GRCh38
rs961050537 Chr.6: 31642264-31642264 on Build GRCh38
rs763305970 Chr.6: 31642223-31642223 on Build GRCh38
rs922395346 Chr.6: 31642449-31642449 on Build GRCh38
rs752719899 Chr.6: 31642278-31642278 on Build GRCh38
rs570688060 Chr.6: 31642239-31642239 on Build GRCh38
rs772057792 Chr.6: 31642160-31642160 on Build GRCh38
rs777534985 Chr.6: 31642175-31642175 on Build GRCh38
rs767665553 Chr.6: 31642254-31642254 on Build GRCh38
rs1002411823 Chr.6: 31642252-31642252 on Build GRCh38
rs557448963 Chr.6: 31642353-31642353 on Build GRCh38
rs758779293 Chr.6: 31642061-31642061 on Build GRCh38
rs745374375 Chr.6: 31642320-31642320 on Build GRCh38
rs758242824 Chr.6: 31642282-31642282 on Build GRCh38
rs762089651 Chr.6: 31642345-31642345 on Build GRCh38
rs372412369 Chr.6: 31642419-31642422 on Build GRCh38
rs774659500 Chr.6: 31642341-31642341 on Build GRCh38
rs112889368 Chr.6: 31642416-31642416 on Build GRCh38
rs755714 Chr.6: 31642036-31642036 on Build GRCh38
rs760775657 Chr.6: 31642382-31642382 on Build GRCh38
rs750677124 Chr.6: 31642374-31642374 on Build GRCh38
rs764290386 Chr.6: 31642065-31642065 on Build GRCh38
rs11548856 Chr.6: 31642174-31642174 on Build GRCh38
rs371338269 Chr.6: 31642071-31642071 on Build GRCh38
rs986776721 Chr.6: 31642277-31642277 on Build GRCh38
rs36173461 Chr.6: 31642413-31642416 on Build GRCh38
rs778903271 Chr.6: 31642273-31642273 on Build GRCh38
rs375303723 Chr.6: 31642201-31642201 on Build GRCh38

View Full Product Details