Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31656840-31657371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOM
Entrez Gene ID
55937
Gene Name
apolipoprotein M
Gene Aliases
G3a, HSPC336, NG20, apo-M
UniGene
Hs.534468
Gene Chromosome Location
Chr.6: 31652410-31658210 on Build GRCh38
Transcripts
NM_001256169.1 NM_019101.2 NR_045828.1 XM_006715150.3 XM_017011279.1

Gene Symbol
BAG6
Entrez Gene ID
7917
Gene Name
BCL2 associated athanogene 6
Gene Aliases
BAG-6, BAT3, D6S52E, G3
UniGene
Hs.440900
Gene Chromosome Location
Chr.6: 31639028-31660900 on Build GRCh38
Transcripts
NM_001256169.1 NM_019101.2 NR_045828.1 XM_006715150.3 XM_017011279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3624668 c.54G>A p.M18I Chr.6: 31657225-31657225 on Build GRCh38
6445053 c.292C>A p.R98R Chr.6: 31657247-31657247 on Build GRCh38
3662386 c.119G>C p.R40T Chr.6: 31657290-31657290 on Build GRCh38
1621433 c.335G>C p.R112T Chr.6: 31657290-31657290 on Build GRCh38
3994838 c.282C>G p.L94L Chr.6: 31657237-31657237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34490746 Chr.6: 31657231-31657231 on Build GRCh38
rs745413531 Chr.6: 31657299-31657299 on Build GRCh38
rs377060558 Chr.6: 31657145-31657146 on Build GRCh38
rs1042578831 Chr.6: 31657158-31657160 on Build GRCh38
rs778859096 Chr.6: 31657247-31657247 on Build GRCh38
rs982686331 Chr.6: 31657050-31657050 on Build GRCh38
rs557308811 Chr.6: 31656997-31656997 on Build GRCh38
rs774809381 Chr.6: 31657342-31657342 on Build GRCh38
rs553459028 Chr.6: 31656929-31656929 on Build GRCh38
rs777632761 Chr.6: 31657283-31657283 on Build GRCh38
rs764298278 Chr.6: 31657341-31657341 on Build GRCh38
rs932788914 Chr.6: 31657076-31657076 on Build GRCh38
rs754885489 Chr.6: 31657243-31657243 on Build GRCh38
rs761891194 Chr.6: 31657345-31657345 on Build GRCh38
rs1003806338 Chr.6: 31656885-31656885 on Build GRCh38
rs758181825 Chr.6: 31657257-31657257 on Build GRCh38
rs1036652651 Chr.6: 31657165-31657165 on Build GRCh38
rs750522968 Chr.6: 31657235-31657235 on Build GRCh38
rs187611888 Chr.6: 31656973-31656973 on Build GRCh38
rs775990589 Chr.6: 31657175-31657175 on Build GRCh38
rs1054228230 Chr.6: 31657298-31657298 on Build GRCh38
rs977439499 Chr.6: 31657063-31657063 on Build GRCh38
rs575596385 Chr.6: 31657002-31657002 on Build GRCh38
rs576528095 Chr.6: 31657094-31657094 on Build GRCh38
rs756714758 Chr.6: 31657325-31657328 on Build GRCh38
rs921425095 Chr.6: 31657072-31657072 on Build GRCh38
rs757530676 Chr.6: 31657217-31657217 on Build GRCh38
rs568592057 Chr.6: 31656939-31656939 on Build GRCh38
rs146017759 Chr.6: 31657041-31657041 on Build GRCh38
rs888160366 Chr.6: 31657095-31657095 on Build GRCh38
rs751683577 Chr.6: 31657199-31657199 on Build GRCh38
rs977381752 Chr.6: 31657040-31657040 on Build GRCh38
rs991568945 Chr.6: 31656999-31656999 on Build GRCh38
rs776197459 Chr.6: 31657315-31657315 on Build GRCh38
rs960397723 Chr.6: 31657111-31657111 on Build GRCh38
rs999904139 Chr.6: 31656942-31656942 on Build GRCh38
rs747967562 Chr.6: 31657248-31657248 on Build GRCh38
rs1015220370 Chr.6: 31656912-31656912 on Build GRCh38
rs1003454247 Chr.6: 31656934-31656934 on Build GRCh38
rs543557924 Chr.6: 31657301-31657301 on Build GRCh38
rs558987938 Chr.6: 31657170-31657170 on Build GRCh38
rs939775028 Chr.6: 31657101-31657101 on Build GRCh38
rs944267193 Chr.6: 31657045-31657045 on Build GRCh38
rs945672108 Chr.6: 31657156-31657156 on Build GRCh38
rs3117581 Chr.6: 31657087-31657087 on Build GRCh38
rs748550789 Chr.6: 31657216-31657216 on Build GRCh38
rs763529245 Chr.6: 31657320-31657320 on Build GRCh38
rs764354826 Chr.6: 31657197-31657197 on Build GRCh38
rs763266789 Chr.6: 31657184-31657184 on Build GRCh38
rs540446691 Chr.6: 31657141-31657141 on Build GRCh38
rs113765873 Chr.6: 31657014-31657014 on Build GRCh38
rs746605413 Chr.6: 31657285-31657285 on Build GRCh38
rs908379795 Chr.6: 31657056-31657056 on Build GRCh38
rs534822736 Chr.6: 31656884-31656884 on Build GRCh38
rs776260881 Chr.6: 31657291-31657291 on Build GRCh38
rs577192457 Chr.6: 31656872-31656872 on Build GRCh38
rs770733076 Chr.6: 31657286-31657286 on Build GRCh38
rs879856798 Chr.6: 31657079-31657079 on Build GRCh38
rs974199274 Chr.6: 31657073-31657073 on Build GRCh38
rs34518823 Chr.6: 31656983-31656983 on Build GRCh38
rs968801571 Chr.6: 31657025-31657025 on Build GRCh38
rs558037993 Chr.6: 31657071-31657071 on Build GRCh38
rs770422280 Chr.6: 31657311-31657311 on Build GRCh38
rs572449926 Chr.6: 31656974-31656974 on Build GRCh38
rs1011126480 Chr.6: 31657279-31657279 on Build GRCh38

View Full Product Details