Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31669012-31669512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK2B
Entrez Gene ID
1460
Gene Name
casein kinase 2 beta
Gene Aliases
CK2B, CK2N, CSK2B, G5A
UniGene
Hs.73527
Gene Chromosome Location
Chr.6: 31665880-31670070 on Build GRCh38
Transcripts
NM_001282385.1 NM_001320.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3861768 c.547T>C p.F183L Chr.6: 31669498-31669498 on Build GRCh38
4153253 c.329C>G p.P110R Chr.6: 31669134-31669134 on Build GRCh38
4675158 c.317T>G p.F106C Chr.6: 31669122-31669122 on Build GRCh38
4654085 c.296A>G p.E99G Chr.6: 31669101-31669101 on Build GRCh38
5424277 c.522C>G p.Y174* Chr.6: 31669473-31669473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778563436 Chr.6: 31669277-31669277 on Build GRCh38
rs141636476 Chr.6: 31669382-31669382 on Build GRCh38
rs182936888 Chr.6: 31669064-31669064 on Build GRCh38
rs1041406256 Chr.6: 31669233-31669233 on Build GRCh38
rs369412604 Chr.6: 31669300-31669300 on Build GRCh38
rs770377932 Chr.6: 31669326-31669326 on Build GRCh38
rs549636828 Chr.6: 31669075-31669075 on Build GRCh38
rs758488928 Chr.6: 31669385-31669386 on Build GRCh38
rs201663166 Chr.6: 31669280-31669280 on Build GRCh38
rs143237802 Chr.6: 31669413-31669413 on Build GRCh38
rs778849848 Chr.6: 31669429-31669429 on Build GRCh38
rs527376842 Chr.6: 31669040-31669040 on Build GRCh38
rs200632979 Chr.6: 31669303-31669303 on Build GRCh38
rs777337858 Chr.6: 31669289-31669289 on Build GRCh38
rs772692956 Chr.6: 31669152-31669152 on Build GRCh38
rs139853188 Chr.6: 31669179-31669179 on Build GRCh38
rs771774143 Chr.6: 31669141-31669141 on Build GRCh38
rs548844893 Chr.6: 31669044-31669044 on Build GRCh38
rs899642200 Chr.6: 31669302-31669302 on Build GRCh38
rs898804504 Chr.6: 31669310-31669310 on Build GRCh38
rs142505258 Chr.6: 31669037-31669037 on Build GRCh38
rs1040839559 Chr.6: 31669297-31669297 on Build GRCh38
rs772868074 Chr.6: 31669309-31669309 on Build GRCh38
rs760110505 Chr.6: 31669198-31669198 on Build GRCh38
rs551070915 Chr.6: 31669249-31669249 on Build GRCh38
rs553912850 Chr.6: 31669190-31669190 on Build GRCh38
rs760116345 Chr.6: 31669160-31669160 on Build GRCh38
rs754963798 Chr.6: 31669050-31669052 on Build GRCh38
rs753700894 Chr.6: 31669271-31669271 on Build GRCh38
rs752538843 Chr.6: 31669280-31669280 on Build GRCh38
rs996300120 Chr.6: 31669036-31669036 on Build GRCh38
rs780559137 Chr.6: 31669216-31669217 on Build GRCh38
rs767467400 Chr.6: 31669419-31669419 on Build GRCh38
rs764585534 Chr.6: 31669412-31669412 on Build GRCh38
rs777997021 Chr.6: 31669063-31669063 on Build GRCh38
rs901401479 Chr.6: 31669263-31669263 on Build GRCh38
rs372025688 Chr.6: 31669221-31669221 on Build GRCh38
rs947036942 Chr.6: 31669224-31669224 on Build GRCh38
rs146463773 Chr.6: 31669062-31669062 on Build GRCh38
rs747720776 Chr.6: 31669133-31669133 on Build GRCh38
rs754988023 Chr.6: 31669276-31669276 on Build GRCh38
rs752166195 Chr.6: 31669262-31669262 on Build GRCh38
rs373542967 Chr.6: 31669372-31669372 on Build GRCh38
rs934202288 Chr.6: 31669267-31669267 on Build GRCh38
rs376794955 Chr.6: 31669401-31669401 on Build GRCh38
rs369875517 Chr.6: 31669301-31669301 on Build GRCh38
rs758681622 Chr.6: 31669206-31669206 on Build GRCh38
rs767812124 Chr.6: 31669182-31669184 on Build GRCh38
rs571224228 Chr.6: 31669097-31669097 on Build GRCh38
rs763344088 Chr.6: 31669407-31669407 on Build GRCh38
rs376337507 Chr.6: 31669283-31669283 on Build GRCh38
rs752579315 Chr.6: 31669467-31669467 on Build GRCh38
rs879013932 Chr.6: 31669200-31669200 on Build GRCh38
rs776252994 Chr.6: 31669345-31669345 on Build GRCh38
rs959877906 Chr.6: 31669038-31669038 on Build GRCh38
rs755230275 Chr.6: 31669054-31669054 on Build GRCh38
rs565785708 Chr.6: 31669226-31669226 on Build GRCh38
rs757375419 Chr.6: 31669220-31669220 on Build GRCh38
rs768289582 Chr.6: 31669091-31669091 on Build GRCh38
rs368195940 Chr.6: 31669204-31669204 on Build GRCh38
rs937522735 Chr.6: 31669196-31669196 on Build GRCh38
rs769320958 Chr.6: 31669094-31669094 on Build GRCh38
rs747587877 Chr.6: 31669089-31669089 on Build GRCh38
rs375923827 Chr.6: 31669168-31669168 on Build GRCh38
rs369625950 Chr.6: 31669218-31669218 on Build GRCh38
rs754855184 Chr.6: 31669428-31669428 on Build GRCh38
rs537452358 Chr.6: 31669065-31669065 on Build GRCh38
rs780072145 Chr.6: 31669072-31669072 on Build GRCh38
rs765496721 Chr.6: 31669203-31669203 on Build GRCh38

View Full Product Details