Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31734065-31734562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC1
Entrez Gene ID
1192
Gene Name
chloride intracellular channel 1
Gene Aliases
G6, NCC27
UniGene
Hs.414565
Gene Chromosome Location
Chr.6: 31730581-31737318 on Build GRCh38
Transcripts
NM_001287593.1 NM_001287594.1 NM_001288.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1621439 c.149G>A p.R50K Chr.6: 31734154-31734154 on Build GRCh38
7170096 c.108C>T p.L36L Chr.6: 31734195-31734195 on Build GRCh38
3674659 c.124A>C p.N42H Chr.6: 31734179-31734179 on Build GRCh38
451213 c.123C>A p.F41L Chr.6: 31734180-31734180 on Build GRCh38
1077651 c.100C>A p.L34M Chr.6: 31734203-31734203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025172024 Chr.6: 31734349-31734349 on Build GRCh38
rs549670224 Chr.6: 31734111-31734112 on Build GRCh38
rs892877847 Chr.6: 31734222-31734222 on Build GRCh38
rs763914555 Chr.6: 31734278-31734278 on Build GRCh38
rs957917326 Chr.6: 31734440-31734440 on Build GRCh38
rs188156453 Chr.6: 31734339-31734339 on Build GRCh38
rs772234744 Chr.6: 31734234-31734234 on Build GRCh38
rs1039975474 Chr.6: 31734161-31734161 on Build GRCh38
rs538098551 Chr.6: 31734357-31734358 on Build GRCh38
rs774609759 Chr.6: 31734206-31734206 on Build GRCh38
rs11549121 Chr.6: 31734159-31734159 on Build GRCh38
rs368078159 Chr.6: 31734115-31734115 on Build GRCh38
rs993978629 Chr.6: 31734430-31734430 on Build GRCh38
rs117944423 Chr.6: 31734124-31734124 on Build GRCh38
rs879145176 Chr.6: 31734231-31734231 on Build GRCh38
rs780642956 Chr.6: 31734173-31734173 on Build GRCh38
rs776237460 Chr.6: 31734400-31734400 on Build GRCh38
rs887290987 Chr.6: 31734317-31734317 on Build GRCh38
rs995765939 Chr.6: 31734237-31734237 on Build GRCh38
rs762068573 Chr.6: 31734213-31734213 on Build GRCh38
rs1027520165 Chr.6: 31734445-31734448 on Build GRCh38
rs760814992 Chr.6: 31734251-31734251 on Build GRCh38
rs376901386 Chr.6: 31734109-31734109 on Build GRCh38
rs961195469 Chr.6: 31734420-31734420 on Build GRCh38
rs566358114 Chr.6: 31734235-31734235 on Build GRCh38
rs1025777559 Chr.6: 31734263-31734263 on Build GRCh38
rs762685409 Chr.6: 31734315-31734315 on Build GRCh38
rs558077604 Chr.6: 31734151-31734151 on Build GRCh38
rs761696662 Chr.6: 31734500-31734500 on Build GRCh38
rs771023111 Chr.6: 31734122-31734122 on Build GRCh38
rs570496971 Chr.6: 31734203-31734203 on Build GRCh38
rs980446120 Chr.6: 31734377-31734377 on Build GRCh38
rs988021423 Chr.6: 31734452-31734452 on Build GRCh38
rs933164446 Chr.6: 31734533-31734533 on Build GRCh38
rs780731721 Chr.6: 31734156-31734156 on Build GRCh38
rs949599512 Chr.6: 31734094-31734094 on Build GRCh38
rs766457611 Chr.6: 31734264-31734264 on Build GRCh38
rs754891567 Chr.6: 31734111-31734111 on Build GRCh38
rs772640544 Chr.6: 31734328-31734328 on Build GRCh38
rs1011697299 Chr.6: 31734316-31734316 on Build GRCh38
rs752580211 Chr.6: 31734266-31734266 on Build GRCh38
rs760181052 Chr.6: 31734324-31734324 on Build GRCh38
rs969063982 Chr.6: 31734355-31734355 on Build GRCh38
rs569618430 Chr.6: 31734091-31734091 on Build GRCh38
rs924120648 Chr.6: 31734167-31734167 on Build GRCh38
rs751359929 Chr.6: 31734307-31734307 on Build GRCh38
rs772430760 Chr.6: 31734122-31734123 on Build GRCh38
rs763824365 Chr.6: 31734325-31734325 on Build GRCh38
rs192913816 Chr.6: 31734504-31734504 on Build GRCh38
rs770418303 Chr.6: 31734172-31734172 on Build GRCh38
rs762825524 Chr.6: 31734275-31734275 on Build GRCh38
rs777448313 Chr.6: 31734125-31734125 on Build GRCh38
rs756896652 Chr.6: 31734152-31734152 on Build GRCh38
rs749827308 Chr.6: 31734181-31734181 on Build GRCh38
rs373292411 Chr.6: 31734514-31734514 on Build GRCh38
rs752488228 Chr.6: 31734121-31734121 on Build GRCh38
rs1019465936 Chr.6: 31734415-31734415 on Build GRCh38
rs878994628 Chr.6: 31734149-31734149 on Build GRCh38
rs141106730 Chr.6: 31734168-31734168 on Build GRCh38

View Full Product Details