Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32028347-32028853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4B
Entrez Gene ID
721
Gene Name
complement component 4B (Chido blood group)
Gene Aliases
C4B1, C4B12, C4B2, C4B3, C4B5, C4BD, C4B_2, C4F, CH, CO4, CPAMD3
UniGene
Hs.534847
Gene Chromosome Location
Chr.6: 32014795-32035418 on Build GRCh38
Transcripts
NM_001002029.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3921426 c.3364C>T p.P1122S Chr.6: 32028826-32028826 on Build GRCh38
4824964 c.3218G>A p.G1073D Chr.6: 32028520-32028520 on Build GRCh38
6051676 c.3373C>T p.H1125Y Chr.6: 32028835-32028835 on Build GRCh38
5079858 c.3219C>A p.G1073G Chr.6: 32028521-32028521 on Build GRCh38
3624973 c.3369G>A p.V1123V Chr.6: 32028831-32028831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370548864 Chr.6: 32028806-32028806 on Build GRCh38
rs750118429 Chr.6: 32028487-32028487 on Build GRCh38
rs772562861 Chr.6: 32028434-32028434 on Build GRCh38
rs778115809 Chr.6: 32028737-32028737 on Build GRCh38
rs558404699 Chr.6: 32028707-32028707 on Build GRCh38
rs1044841490 Chr.6: 32028627-32028627 on Build GRCh38
rs769935919 Chr.6: 32028779-32028779 on Build GRCh38
rs776681958 Chr.6: 32028536-32028536 on Build GRCh38
rs773367670 Chr.6: 32028436-32028436 on Build GRCh38
rs765284935 Chr.6: 32028546-32028546 on Build GRCh38
rs776894309 Chr.6: 32028761-32028761 on Build GRCh38
rs796137642 Chr.6: 32028486-32028486 on Build GRCh38
rs779524856 Chr.6: 32028492-32028492 on Build GRCh38
rs200897112 Chr.6: 32028820-32028820 on Build GRCh38
rs753631255 Chr.6: 32028828-32028828 on Build GRCh38
rs199623227 Chr.6: 32028508-32028508 on Build GRCh38
rs748856364 Chr.6: 32028497-32028497 on Build GRCh38
rs139543260 Chr.6: 32028516-32028516 on Build GRCh38
rs758980099 Chr.6: 32028727-32028727 on Build GRCh38
rs760969836 Chr.6: 32028437-32028437 on Build GRCh38
rs761741343 Chr.6: 32028556-32028556 on Build GRCh38
rs369928228 Chr.6: 32028502-32028502 on Build GRCh38
rs764056735 Chr.6: 32028464-32028464 on Build GRCh38
rs368701430 Chr.6: 32028829-32028829 on Build GRCh38
rs753971112 Chr.6: 32028444-32028444 on Build GRCh38
rs762310787 Chr.6: 32028414-32028414 on Build GRCh38
rs759584925 Chr.6: 32028447-32028447 on Build GRCh38
rs377662798 Chr.6: 32028541-32028541 on Build GRCh38
rs2734319 Chr.6: 32028821-32028821 on Build GRCh38
rs2229399 Chr.6: 32028832-32028832 on Build GRCh38
rs751491740 Chr.6: 32028473-32028473 on Build GRCh38
rs2258218 Chr.6: 32028520-32028520 on Build GRCh38
rs55704392 Chr.6: 32028408-32028408 on Build GRCh38
rs1048557718 Chr.6: 32028390-32028390 on Build GRCh38
rs772403639 Chr.6: 32028517-32028517 on Build GRCh38
rs762892849 Chr.6: 32028805-32028805 on Build GRCh38
rs753537210 Chr.6: 32028658-32028658 on Build GRCh38
rs750254719 Chr.6: 32028558-32028558 on Build GRCh38
rs370516666 Chr.6: 32028750-32028750 on Build GRCh38
rs145045914 Chr.6: 32028521-32028521 on Build GRCh38
rs371168662 Chr.6: 32028831-32028831 on Build GRCh38
rs755842558 Chr.6: 32028569-32028569 on Build GRCh38
rs375317133 Chr.6: 32028638-32028638 on Build GRCh38
rs754596083 Chr.6: 32028661-32028661 on Build GRCh38
rs767540386 Chr.6: 32028809-32028809 on Build GRCh38
rs757099827 Chr.6: 32028480-32028480 on Build GRCh38
rs373921185 Chr.6: 32028766-32028766 on Build GRCh38
rs767316218 Chr.6: 32028557-32028557 on Build GRCh38
rs2734318 Chr.6: 32028824-32028824 on Build GRCh38
rs574421495 Chr.6: 32028810-32028810 on Build GRCh38
rs775488185 Chr.6: 32028553-32028553 on Build GRCh38
rs767356444 Chr.6: 32028481-32028481 on Build GRCh38
rs540001873 Chr.6: 32028700-32028700 on Build GRCh38
rs377359889 Chr.6: 32028804-32028804 on Build GRCh38
rs771229399 Chr.6: 32028533-32028533 on Build GRCh38
rs766096753 Chr.6: 32028584-32028584 on Build GRCh38
rs755820820 Chr.6: 32028488-32028488 on Build GRCh38

View Full Product Details