Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35776482-35777014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPSL2
Entrez Gene ID
389383
Gene Name
colipase like 2
Gene Aliases
AAAL3045, C6orf126, UNQ3045, dJ510O8.5
UniGene
Hs.259563
Gene Chromosome Location
Chr.6: 35776594-35779552 on Build GRCh38
Transcripts
NM_001286550.1 NM_207409.3 NR_104467.1 NR_104469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6381736 c.8C>T p.A3V Chr.6: 35776626-35776626 on Build GRCh38
5690813 c.6C>G p.A2A Chr.6: 35776624-35776624 on Build GRCh38
5570619 c.32T>G p.V11G Chr.6: 35776650-35776650 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs902247358 Chr.6: 35776879-35776879 on Build GRCh38
rs999874326 Chr.6: 35776627-35776627 on Build GRCh38
rs967574946 Chr.6: 35776581-35776581 on Build GRCh38
rs111305117 Chr.6: 35776755-35776755 on Build GRCh38
rs972729199 Chr.6: 35776770-35776770 on Build GRCh38
rs370833443 Chr.6: 35776584-35776584 on Build GRCh38
rs987323297 Chr.6: 35776702-35776702 on Build GRCh38
rs529766282 Chr.6: 35776944-35776944 on Build GRCh38
rs547386035 Chr.6: 35776895-35776895 on Build GRCh38
rs188591349 Chr.6: 35776528-35776528 on Build GRCh38
rs1052017970 Chr.6: 35776501-35776501 on Build GRCh38
rs766309715 Chr.6: 35776645-35776645 on Build GRCh38
rs934099089 Chr.6: 35776673-35776673 on Build GRCh38
rs904291652 Chr.6: 35776948-35776948 on Build GRCh38
rs892024465 Chr.6: 35776644-35776644 on Build GRCh38
rs1028566056 Chr.6: 35776936-35776936 on Build GRCh38
rs904154313 Chr.6: 35776620-35776620 on Build GRCh38
rs565013285 Chr.6: 35776859-35776859 on Build GRCh38
rs768211863 Chr.6: 35776730-35776730 on Build GRCh38
rs921667666 Chr.6: 35776567-35776567 on Build GRCh38
rs1024487629 Chr.6: 35776576-35776576 on Build GRCh38
rs753158423 Chr.6: 35776973-35776973 on Build GRCh38
rs896252081 Chr.6: 35776541-35776541 on Build GRCh38
rs144080170 Chr.6: 35776883-35776883 on Build GRCh38
rs939850118 Chr.6: 35776500-35776500 on Build GRCh38
rs2296662 Chr.6: 35776992-35776992 on Build GRCh38
rs115808575 Chr.6: 35776789-35776789 on Build GRCh38
rs1038290738 Chr.6: 35776504-35776504 on Build GRCh38
rs760971197 Chr.6: 35776985-35776985 on Build GRCh38
rs565799601 Chr.6: 35776931-35776931 on Build GRCh38
rs1033736688 Chr.6: 35776534-35776534 on Build GRCh38
rs1033150744 Chr.6: 35776911-35776911 on Build GRCh38
rs970292321 Chr.6: 35776751-35776751 on Build GRCh38
rs975424856 Chr.6: 35776641-35776641 on Build GRCh38
rs985624222 Chr.6: 35776650-35776650 on Build GRCh38
rs896108128 Chr.6: 35776791-35776791 on Build GRCh38
rs542418070 Chr.6: 35776619-35776619 on Build GRCh38
rs955597142 Chr.6: 35776954-35776954 on Build GRCh38
rs753624440 Chr.6: 35776707-35776707 on Build GRCh38
rs908621989 Chr.6: 35776721-35776721 on Build GRCh38
rs2766578 Chr.6: 35776526-35776526 on Build GRCh38
rs1031904519 Chr.6: 35776637-35776637 on Build GRCh38
rs759325966 Chr.6: 35776736-35776736 on Build GRCh38
rs1038153987 Chr.6: 35776745-35776745 on Build GRCh38
rs1013246098 Chr.6: 35776847-35776847 on Build GRCh38
rs560710029 Chr.6: 35776741-35776741 on Build GRCh38
rs1033286346 Chr.6: 35776608-35776608 on Build GRCh38
rs962658855 Chr.6: 35776767-35776767 on Build GRCh38
rs1013715619 Chr.6: 35776558-35776558 on Build GRCh38
rs943108358 Chr.6: 35776527-35776527 on Build GRCh38
rs937904418 Chr.6: 35776765-35776765 on Build GRCh38
rs943223932 Chr.6: 35776906-35776906 on Build GRCh38
rs957628357 Chr.6: 35776928-35776928 on Build GRCh38
rs2296663 Chr.6: 35776969-35776969 on Build GRCh38
rs1038773007 Chr.6: 35776947-35776947 on Build GRCh38
rs931753778 Chr.6: 35776590-35776590 on Build GRCh38
rs112977978 Chr.6: 35776990-35776990 on Build GRCh38

View Full Product Details