Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36196617-36197115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRPF3
Entrez Gene ID
27154
Gene Name
bromodomain and PHD finger containing 3
Gene Aliases
-
UniGene
Hs.520096
Gene Chromosome Location
Chr.6: 36196773-36232790 on Build GRCh38
Transcripts
NM_015695.2

SNP Detail

SNP ID Chromosome Location
rs936483885 Chr.6: 36196845-36196845 on Build GRCh38
rs958815069 Chr.6: 36197056-36197056 on Build GRCh38
rs1020676578 Chr.6: 36196925-36196925 on Build GRCh38
rs1039490148 Chr.6: 36196836-36196836 on Build GRCh38
rs1039520740 Chr.6: 36196722-36196722 on Build GRCh38
rs1034458186 Chr.6: 36196756-36196756 on Build GRCh38
rs1000759224 Chr.6: 36196731-36196731 on Build GRCh38
rs943738508 Chr.6: 36196832-36196832 on Build GRCh38
rs1010653213 Chr.6: 36196744-36196744 on Build GRCh38
rs552871656 Chr.6: 36196930-36196930 on Build GRCh38
rs912237286 Chr.6: 36196827-36196827 on Build GRCh38
rs892419147 Chr.6: 36196734-36196734 on Build GRCh38
rs1055783427 Chr.6: 36196980-36196980 on Build GRCh38
rs982816281 Chr.6: 36196823-36196823 on Build GRCh38
rs894397864 Chr.6: 36197065-36197065 on Build GRCh38
rs971421169 Chr.6: 36196746-36196746 on Build GRCh38
rs1002853748 Chr.6: 36196939-36196939 on Build GRCh38
rs974644160 Chr.6: 36197057-36197057 on Build GRCh38
rs929618039 Chr.6: 36196673-36196673 on Build GRCh38
rs540893408 Chr.6: 36196909-36196909 on Build GRCh38
rs890866429 Chr.6: 36196714-36196714 on Build GRCh38
rs1009497372 Chr.6: 36196918-36196918 on Build GRCh38
rs949926566 Chr.6: 36197089-36197089 on Build GRCh38
rs1046759260 Chr.6: 36196694-36196694 on Build GRCh38
rs974295777 Chr.6: 36196793-36196794 on Build GRCh38
rs892340054 Chr.6: 36196903-36196903 on Build GRCh38
rs899678645 Chr.6: 36196730-36196730 on Build GRCh38
rs920214430 Chr.6: 36196642-36196642 on Build GRCh38
rs867778634 Chr.6: 36196659-36196659 on Build GRCh38
rs930280951 Chr.6: 36196805-36196805 on Build GRCh38
rs1020289308 Chr.6: 36196745-36196745 on Build GRCh38
rs576861710 Chr.6: 36196843-36196843 on Build GRCh38
rs1009105773 Chr.6: 36196781-36196781 on Build GRCh38
rs899606488 Chr.6: 36196838-36196838 on Build GRCh38
rs773364411 Chr.6: 36196764-36196764 on Build GRCh38
rs558660360 Chr.6: 36196716-36196716 on Build GRCh38
rs920163997 Chr.6: 36196799-36196799 on Build GRCh38
rs907232277 Chr.6: 36196927-36196927 on Build GRCh38
rs1040535711 Chr.6: 36196972-36196972 on Build GRCh38
rs1053667406 Chr.6: 36196875-36196875 on Build GRCh38
rs1032662681 Chr.6: 36196732-36196732 on Build GRCh38
rs981504893 Chr.6: 36196754-36196754 on Build GRCh38

View Full Product Details