Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36201362-36201872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRPF3
Entrez Gene ID
27154
Gene Name
bromodomain and PHD finger containing 3
Gene Aliases
-
UniGene
Hs.520096
Gene Chromosome Location
Chr.6: 36196773-36232790 on Build GRCh38
Transcripts
NM_015695.2 XM_005249010.2 XM_005249011.3 XM_011514489.1 XM_011514490.1 XM_011514491.1 XM_011514492.1 XM_017010742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1078487 c.1347G>T p.K449N Chr.6: 36201669-36201669 on Build GRCh38
6836808 c.1048G>C p.V350L Chr.6: 36201370-36201370 on Build GRCh38
3745209 c.1063C>T p.R355W Chr.6: 36201385-36201385 on Build GRCh38
5720583 c.1092C>T p.P364P Chr.6: 36201414-36201414 on Build GRCh38
3765276 c.1255G>A p.E419K Chr.6: 36201577-36201577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375502327 Chr.6: 36201546-36201546 on Build GRCh38
rs755747786 Chr.6: 36201683-36201683 on Build GRCh38
rs201472164 Chr.6: 36201418-36201418 on Build GRCh38
rs35085365 Chr.6: 36201444-36201445 on Build GRCh38
rs774157573 Chr.6: 36201487-36201487 on Build GRCh38
rs766466169 Chr.6: 36201412-36201412 on Build GRCh38
rs200505383 Chr.6: 36201602-36201602 on Build GRCh38
rs146148846 Chr.6: 36201627-36201627 on Build GRCh38
rs143027666 Chr.6: 36201528-36201528 on Build GRCh38
rs1001736296 Chr.6: 36201621-36201621 on Build GRCh38
rs188995700 Chr.6: 36201547-36201547 on Build GRCh38
rs373444225 Chr.6: 36201564-36201564 on Build GRCh38
rs773051200 Chr.6: 36201632-36201632 on Build GRCh38
rs752002255 Chr.6: 36201554-36201554 on Build GRCh38
rs755067208 Chr.6: 36201809-36201811 on Build GRCh38
rs778378713 Chr.6: 36201719-36201719 on Build GRCh38
rs766711409 Chr.6: 36201818-36201818 on Build GRCh38
rs755050538 Chr.6: 36201416-36201416 on Build GRCh38
rs771975195 Chr.6: 36201727-36201727 on Build GRCh38
rs543987259 Chr.6: 36201592-36201592 on Build GRCh38
rs950685173 Chr.6: 36201837-36201837 on Build GRCh38
rs146699190 Chr.6: 36201846-36201846 on Build GRCh38
rs775734689 Chr.6: 36201636-36201636 on Build GRCh38
rs753460033 Chr.6: 36201714-36201714 on Build GRCh38
rs749228433 Chr.6: 36201482-36201482 on Build GRCh38
rs779083604 Chr.6: 36201740-36201740 on Build GRCh38
rs774507949 Chr.6: 36201775-36201775 on Build GRCh38
rs149353065 Chr.6: 36201443-36201443 on Build GRCh38
rs973054745 Chr.6: 36201705-36201705 on Build GRCh38
rs777439878 Chr.6: 36201728-36201728 on Build GRCh38
rs369451160 Chr.6: 36201768-36201768 on Build GRCh38
rs771817686 Chr.6: 36201631-36201631 on Build GRCh38
rs764237669 Chr.6: 36201648-36201648 on Build GRCh38
rs747319225 Chr.6: 36201425-36201425 on Build GRCh38
rs142522369 Chr.6: 36201580-36201580 on Build GRCh38
rs538064790 Chr.6: 36201764-36201764 on Build GRCh38
rs745575376 Chr.6: 36201563-36201563 on Build GRCh38
rs140145706 Chr.6: 36201667-36201667 on Build GRCh38
rs775892441 Chr.6: 36201736-36201736 on Build GRCh38
rs754535785 Chr.6: 36201718-36201718 on Build GRCh38
rs561103333 Chr.6: 36201389-36201389 on Build GRCh38
rs867021451 Chr.6: 36201598-36201598 on Build GRCh38
rs531428048 Chr.6: 36201420-36201420 on Build GRCh38
rs963400328 Chr.6: 36201669-36201669 on Build GRCh38
rs747521501 Chr.6: 36201588-36201588 on Build GRCh38
rs781608347 Chr.6: 36201561-36201561 on Build GRCh38
rs768023291 Chr.6: 36201796-36201796 on Build GRCh38
rs759678310 Chr.6: 36201634-36201634 on Build GRCh38
rs201805258 Chr.6: 36201577-36201577 on Build GRCh38
rs367822135 Chr.6: 36201581-36201581 on Build GRCh38
rs369791645 Chr.6: 36201614-36201614 on Build GRCh38
rs769248096 Chr.6: 36201454-36201454 on Build GRCh38
rs905645857 Chr.6: 36201618-36201618 on Build GRCh38
rs776763624 Chr.6: 36201510-36201510 on Build GRCh38
rs775060976 Chr.6: 36201476-36201476 on Build GRCh38
rs375532065 Chr.6: 36201647-36201647 on Build GRCh38
rs1055306226 Chr.6: 36201776-36201776 on Build GRCh38
rs145368816 Chr.6: 36201517-36201517 on Build GRCh38
rs752171996 Chr.6: 36201652-36201652 on Build GRCh38
rs749349987 Chr.6: 36201784-36201784 on Build GRCh38
rs745512954 Chr.6: 36201448-36201448 on Build GRCh38
rs577867309 Chr.6: 36201628-36201628 on Build GRCh38
rs753985752 Chr.6: 36201414-36201414 on Build GRCh38
rs367591401 Chr.6: 36201411-36201411 on Build GRCh38
rs772065988 Chr.6: 36201815-36201815 on Build GRCh38
rs920158164 Chr.6: 36201406-36201406 on Build GRCh38
rs769940575 Chr.6: 36201733-36201733 on Build GRCh38
rs748052118 Chr.6: 36201629-36201629 on Build GRCh38
rs767777978 Chr.6: 36201674-36201674 on Build GRCh38
rs750743676 Chr.6: 36201675-36201675 on Build GRCh38
rs372570612 Chr.6: 36201635-36201635 on Build GRCh38
rs762865271 Chr.6: 36201385-36201385 on Build GRCh38
rs983641139 Chr.6: 36201651-36201651 on Build GRCh38
rs774069589 Chr.6: 36201394-36201394 on Build GRCh38
rs779645176 Chr.6: 36201704-36201704 on Build GRCh38
rs757601395 Chr.6: 36201560-36201560 on Build GRCh38
rs780551785 Chr.6: 36201580-36201588 on Build GRCh38
rs764646708 Chr.6: 36201552-36201552 on Build GRCh38
rs778286287 Chr.6: 36201421-36201421 on Build GRCh38
rs748148054 Chr.6: 36201724-36201724 on Build GRCh38
rs545548021 Chr.6: 36201847-36201847 on Build GRCh38
rs964570729 Chr.6: 36201494-36201494 on Build GRCh38
rs943527164 Chr.6: 36201488-36201488 on Build GRCh38
rs1000853912 Chr.6: 36201489-36201489 on Build GRCh38
rs936262595 Chr.6: 36201579-36201579 on Build GRCh38
rs918112521 Chr.6: 36201799-36201799 on Build GRCh38
rs761559040 Chr.6: 36201490-36201490 on Build GRCh38
rs750789148 Chr.6: 36201812-36201812 on Build GRCh38
rs200192995 Chr.6: 36201770-36201770 on Build GRCh38
rs143631852 Chr.6: 36201483-36201483 on Build GRCh38
rs755726547 Chr.6: 36201567-36201567 on Build GRCh38
rs756990414 Chr.6: 36201435-36201435 on Build GRCh38
rs571016734 Chr.6: 36201500-36201500 on Build GRCh38
rs762240891 Chr.6: 36201785-36201785 on Build GRCh38
rs748751219 Chr.6: 36201606-36201606 on Build GRCh38
rs753198591 Chr.6: 36201419-36201419 on Build GRCh38
rs1042465505 Chr.6: 36201450-36201450 on Build GRCh38

View Full Product Details