Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36204569-36205066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRPF3
Entrez Gene ID
27154
Gene Name
bromodomain and PHD finger containing 3
Gene Aliases
-
UniGene
Hs.520096
Gene Chromosome Location
Chr.6: 36196773-36232790 on Build GRCh38
Transcripts
NM_015695.2 XM_005249010.2 XM_005249011.3 XM_011514489.1 XM_011514490.1 XM_011514491.1 XM_011514492.1 XM_017010742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3626163 c.1476C>T p.S492S Chr.6: 36204685-36204685 on Build GRCh38
7290570 c.1596C>T p.N532N Chr.6: 36204805-36204805 on Build GRCh38
3626165 c.1551C>T p.V517V Chr.6: 36204760-36204760 on Build GRCh38
6612918 c.1561C>T p.R521W Chr.6: 36204770-36204770 on Build GRCh38
3626164 c.1484G>A p.R495K Chr.6: 36204693-36204693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529294626 Chr.6: 36204915-36204916 on Build GRCh38
rs977699473 Chr.6: 36204916-36204916 on Build GRCh38
rs750358421 Chr.6: 36204789-36204789 on Build GRCh38
rs750742278 Chr.6: 36204714-36204714 on Build GRCh38
rs761867097 Chr.6: 36204784-36204784 on Build GRCh38
rs767523670 Chr.6: 36204788-36204788 on Build GRCh38
rs202026755 Chr.6: 36204648-36204648 on Build GRCh38
rs368933034 Chr.6: 36204651-36204651 on Build GRCh38
rs1002871195 Chr.6: 36204930-36204930 on Build GRCh38
rs778878085 Chr.6: 36204638-36204639 on Build GRCh38
rs907219553 Chr.6: 36204914-36204914 on Build GRCh38
rs747514574 Chr.6: 36204745-36204745 on Build GRCh38
rs200416863 Chr.6: 36204781-36204781 on Build GRCh38
rs771455384 Chr.6: 36204856-36204856 on Build GRCh38
rs868571253 Chr.6: 36204796-36204796 on Build GRCh38
rs1011383988 Chr.6: 36205000-36205000 on Build GRCh38
rs530369080 Chr.6: 36204895-36204895 on Build GRCh38
rs193190287 Chr.6: 36204596-36204596 on Build GRCh38
rs771367448 Chr.6: 36204747-36204747 on Build GRCh38
rs2245972 Chr.6: 36204597-36204597 on Build GRCh38
rs200992622 Chr.6: 36204676-36204676 on Build GRCh38
rs923649260 Chr.6: 36204942-36204942 on Build GRCh38
rs915456116 Chr.6: 36204906-36204906 on Build GRCh38
rs774595904 Chr.6: 36204608-36204608 on Build GRCh38
rs116373261 Chr.6: 36205007-36205007 on Build GRCh38
rs1030187454 Chr.6: 36204891-36204891 on Build GRCh38
rs747039619 Chr.6: 36204828-36204828 on Build GRCh38
rs985638149 Chr.6: 36205042-36205042 on Build GRCh38
rs763666019 Chr.6: 36204783-36204783 on Build GRCh38
rs745611343 Chr.6: 36204724-36204724 on Build GRCh38
rs985666314 Chr.6: 36204619-36204619 on Build GRCh38
rs779239085 Chr.6: 36204733-36204733 on Build GRCh38
rs779705162 Chr.6: 36204805-36204805 on Build GRCh38
rs377760778 Chr.6: 36204727-36204727 on Build GRCh38
rs759196312 Chr.6: 36204654-36204654 on Build GRCh38
rs541317631 Chr.6: 36204793-36204793 on Build GRCh38
rs1056207912 Chr.6: 36204966-36204966 on Build GRCh38
rs746349490 Chr.6: 36204859-36204859 on Build GRCh38
rs757196169 Chr.6: 36204713-36204713 on Build GRCh38
rs759852544 Chr.6: 36204769-36204769 on Build GRCh38
rs879194094 Chr.6: 36204708-36204708 on Build GRCh38
rs955920070 Chr.6: 36204955-36204955 on Build GRCh38
rs1012975969 Chr.6: 36204843-36204843 on Build GRCh38
rs772017988 Chr.6: 36204640-36204640 on Build GRCh38
rs763488575 Chr.6: 36204674-36204674 on Build GRCh38
rs752178915 Chr.6: 36204663-36204663 on Build GRCh38
rs368808053 Chr.6: 36204712-36204712 on Build GRCh38
rs557769148 Chr.6: 36204741-36204741 on Build GRCh38
rs1005520876 Chr.6: 36204824-36204824 on Build GRCh38
rs772255575 Chr.6: 36204736-36204736 on Build GRCh38
rs762458025 Chr.6: 36205033-36205033 on Build GRCh38
rs753033654 Chr.6: 36204806-36204806 on Build GRCh38
rs373180513 Chr.6: 36204659-36204659 on Build GRCh38
rs542408683 Chr.6: 36204939-36204939 on Build GRCh38
rs200249910 Chr.6: 36204774-36204774 on Build GRCh38
rs985635177 Chr.6: 36204905-36204905 on Build GRCh38
rs375182851 Chr.6: 36204818-36204818 on Build GRCh38
rs757965976 Chr.6: 36204670-36204670 on Build GRCh38
rs961734878 Chr.6: 36204857-36204857 on Build GRCh38
rs879113642 Chr.6: 36204889-36204889 on Build GRCh38
rs55855551 Chr.6: 36205006-36205006 on Build GRCh38
rs777000359 Chr.6: 36204750-36204750 on Build GRCh38
rs777263159 Chr.6: 36204915-36204915 on Build GRCh38
rs140994826 Chr.6: 36204944-36204944 on Build GRCh38
rs781692679 Chr.6: 36204858-36204858 on Build GRCh38
rs777917899 Chr.6: 36204825-36204825 on Build GRCh38
rs74365270 Chr.6: 36204894-36204894 on Build GRCh38
rs773613723 Chr.6: 36204759-36204759 on Build GRCh38
rs372776155 Chr.6: 36204622-36204622 on Build GRCh38
rs748223037 Chr.6: 36204635-36204635 on Build GRCh38
rs765553189 Chr.6: 36204770-36204770 on Build GRCh38
rs773820315 Chr.6: 36204641-36204641 on Build GRCh38
rs748299995 Chr.6: 36204734-36204734 on Build GRCh38
rs530897294 Chr.6: 36204946-36204946 on Build GRCh38

View Full Product Details