Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36365777-36366277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV7
Entrez Gene ID
51513
Gene Name
ETS variant 7
Gene Aliases
TEL-2, TEL2, TELB
UniGene
Hs.272398
Gene Chromosome Location
Chr.6: 36354221-36387800 on Build GRCh38
Transcripts
NM_001207035.1 NM_001207036.1 NM_001207037.1 NM_001207038.1 NM_001207039.1 NM_001207040.1 NM_001207041.1 NM_016135.3 XM_011514659.1

SNP Detail

SNP ID Chromosome Location
rs1043283079 Chr.6: 36366152-36366152 on Build GRCh38
rs892018842 Chr.6: 36365952-36365952 on Build GRCh38
rs560121118 Chr.6: 36365889-36365889 on Build GRCh38
rs1010454976 Chr.6: 36365961-36365961 on Build GRCh38
rs147873925 Chr.6: 36365982-36365982 on Build GRCh38
rs779628434 Chr.6: 36365943-36365943 on Build GRCh38
rs909214477 Chr.6: 36365897-36365897 on Build GRCh38
rs974354362 Chr.6: 36366100-36366100 on Build GRCh38
rs967739197 Chr.6: 36365801-36365801 on Build GRCh38
rs903517245 Chr.6: 36365970-36365970 on Build GRCh38
rs912326046 Chr.6: 36366219-36366219 on Build GRCh38
rs369934376 Chr.6: 36366238-36366246 on Build GRCh38
rs563007008 Chr.6: 36365890-36365890 on Build GRCh38
rs6457903 Chr.6: 36365807-36365807 on Build GRCh38
rs144666850 Chr.6: 36365962-36365962 on Build GRCh38
rs1020755446 Chr.6: 36365811-36365811 on Build GRCh38
rs899650272 Chr.6: 36365966-36365966 on Build GRCh38
rs747130273 Chr.6: 36365909-36365909 on Build GRCh38
rs565720432 Chr.6: 36366122-36366122 on Build GRCh38
rs982829471 Chr.6: 36366180-36366183 on Build GRCh38
rs538747276 Chr.6: 36366067-36366067 on Build GRCh38
rs771466740 Chr.6: 36366064-36366064 on Build GRCh38
rs370587577 Chr.6: 36366128-36366128 on Build GRCh38
rs1034955098 Chr.6: 36366078-36366078 on Build GRCh38
rs920141472 Chr.6: 36366105-36366105 on Build GRCh38
rs7748318 Chr.6: 36365849-36365849 on Build GRCh38
rs904803500 Chr.6: 36366164-36366164 on Build GRCh38
rs1053624165 Chr.6: 36365986-36365986 on Build GRCh38
rs140423881 Chr.6: 36366090-36366090 on Build GRCh38
rs775666577 Chr.6: 36366236-36366236 on Build GRCh38
rs111953269 Chr.6: 36366031-36366031 on Build GRCh38
rs181895750 Chr.6: 36366056-36366056 on Build GRCh38
rs943812349 Chr.6: 36366221-36366221 on Build GRCh38
rs983364501 Chr.6: 36366039-36366039 on Build GRCh38
rs1003435859 Chr.6: 36365921-36365921 on Build GRCh38
rs1030175756 Chr.6: 36366030-36366030 on Build GRCh38
rs1057195453 Chr.6: 36366123-36366123 on Build GRCh38
rs964510011 Chr.6: 36365988-36365988 on Build GRCh38
rs975963457 Chr.6: 36365827-36365827 on Build GRCh38
rs746162518 Chr.6: 36366120-36366120 on Build GRCh38
rs927583263 Chr.6: 36365861-36365861 on Build GRCh38
rs750156652 Chr.6: 36365898-36365898 on Build GRCh38
rs1017281785 Chr.6: 36366246-36366246 on Build GRCh38
rs971591998 Chr.6: 36365818-36365818 on Build GRCh38
rs946259295 Chr.6: 36366131-36366131 on Build GRCh38
rs971858923 Chr.6: 36366033-36366033 on Build GRCh38
rs929620281 Chr.6: 36365925-36365925 on Build GRCh38
rs912257652 Chr.6: 36365955-36365955 on Build GRCh38
rs6930836 Chr.6: 36365802-36365802 on Build GRCh38
rs539338162 Chr.6: 36365969-36365969 on Build GRCh38
rs757089304 Chr.6: 36365867-36365867 on Build GRCh38
rs1000572679 Chr.6: 36365977-36365977 on Build GRCh38
rs756721896 Chr.6: 36366025-36366025 on Build GRCh38
rs900214018 Chr.6: 36366254-36366254 on Build GRCh38
rs964895591 Chr.6: 36365812-36365812 on Build GRCh38
rs185490823 Chr.6: 36366106-36366106 on Build GRCh38
rs80096334 Chr.6: 36365907-36365907 on Build GRCh38
rs373381390 Chr.6: 36365910-36365910 on Build GRCh38

View Full Product Details