Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 144377028-144377530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF5
Entrez Gene ID
7984
Gene Name
Rho guanine nucleotide exchange factor 5
Gene Aliases
GEF5, P60, TIM, TIM1
UniGene
Hs.334
Gene Chromosome Location
Chr.7: 144355341-144380632 on Build GRCh38
Transcripts
NM_005435.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4602800 c.4478G>T p.W1493L Chr.7: 144377137-144377137 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs144782851 Chr.7: 144377358-144377358 on Build GRCh38
rs1038704493 Chr.7: 144377433-144377433 on Build GRCh38
rs552701969 Chr.7: 144377371-144377371 on Build GRCh38
rs1046368976 Chr.7: 144377309-144377309 on Build GRCh38
rs543421284 Chr.7: 144377499-144377499 on Build GRCh38
rs986029513 Chr.7: 144377463-144377463 on Build GRCh38
rs982148475 Chr.7: 144377342-144377342 on Build GRCh38
rs905214730 Chr.7: 144377346-144377346 on Build GRCh38
rs549280702 Chr.7: 144377310-144377310 on Build GRCh38
rs1052428312 Chr.7: 144377299-144377299 on Build GRCh38
rs766168734 Chr.7: 144377112-144377112 on Build GRCh38
rs1032829395 Chr.7: 144377294-144377294 on Build GRCh38
rs891640755 Chr.7: 144377295-144377295 on Build GRCh38
rs983680649 Chr.7: 144377389-144377389 on Build GRCh38
rs961829523 Chr.7: 144377060-144377060 on Build GRCh38
rs776296252 Chr.7: 144377236-144377236 on Build GRCh38
rs753838118 Chr.7: 144377133-144377133 on Build GRCh38
rs147927791 Chr.7: 144377492-144377492 on Build GRCh38
rs750210889 Chr.7: 144377086-144377086 on Build GRCh38
rs777881963 Chr.7: 144377156-144377156 on Build GRCh38
rs551180188 Chr.7: 144377062-144377062 on Build GRCh38
rs751741570 Chr.7: 144377171-144377171 on Build GRCh38
rs867591068 Chr.7: 144377413-144377413 on Build GRCh38
rs949504660 Chr.7: 144377305-144377305 on Build GRCh38
rs367816677 Chr.7: 144377355-144377355 on Build GRCh38
rs920738869 Chr.7: 144377372-144377372 on Build GRCh38
rs1010125915 Chr.7: 144377315-144377315 on Build GRCh38
rs959475088 Chr.7: 144377357-144377357 on Build GRCh38
rs925110534 Chr.7: 144377102-144377102 on Build GRCh38
rs781628887 Chr.7: 144377206-144377206 on Build GRCh38
rs975856326 Chr.7: 144377446-144377446 on Build GRCh38
rs376174541 Chr.7: 144377455-144377455 on Build GRCh38
rs866493245 Chr.7: 144377426-144377426 on Build GRCh38
rs577411469 Chr.7: 144377450-144377450 on Build GRCh38
rs1024651695 Chr.7: 144377330-144377330 on Build GRCh38
rs373697388 Chr.7: 144377303-144377303 on Build GRCh38
rs760465416 Chr.7: 144377090-144377090 on Build GRCh38
rs891593225 Chr.7: 144377505-144377505 on Build GRCh38
rs758372239 Chr.7: 144377154-144377154 on Build GRCh38
rs986473691 Chr.7: 144377097-144377097 on Build GRCh38
rs912167381 Chr.7: 144377414-144377414 on Build GRCh38
rs767210479 Chr.7: 144377084-144377084 on Build GRCh38
rs139990614 Chr.7: 144377332-144377332 on Build GRCh38
rs975740380 Chr.7: 144377071-144377071 on Build GRCh38
rs1029975782 Chr.7: 144377452-144377452 on Build GRCh38
rs185706133 Chr.7: 144377293-144377293 on Build GRCh38
rs569668026 Chr.7: 144377227-144377227 on Build GRCh38
rs16882868 Chr.7: 144377466-144377466 on Build GRCh38
rs1033758566 Chr.7: 144377348-144377348 on Build GRCh38
rs929470986 Chr.7: 144377381-144377381 on Build GRCh38

View Full Product Details