Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 144379758-144380263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF5
Entrez Gene ID
7984
Gene Name
Rho guanine nucleotide exchange factor 5
Gene Aliases
GEF5, P60, TIM, TIM1
UniGene
Hs.334
Gene Chromosome Location
Chr.7: 144355341-144380632 on Build GRCh38
Transcripts
NM_005435.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6604366 c.4677C>T p.G1559G Chr.7: 144379939-144379939 on Build GRCh38
3923022 c.4684C>T p.P1562S Chr.7: 144379946-144379946 on Build GRCh38
485000 c.4720G>T p.V1574F Chr.7: 144379982-144379982 on Build GRCh38
1644402 c.4751G>A p.R1584Q Chr.7: 144380013-144380013 on Build GRCh38
6109249 c.4651G>T p.V1551L Chr.7: 144379913-144379913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573094905 Chr.7: 144380133-144380133 on Build GRCh38
rs1626337 Chr.7: 144379822-144379822 on Build GRCh38
rs760671504 Chr.7: 144379874-144379874 on Build GRCh38
rs529930936 Chr.7: 144379828-144379828 on Build GRCh38
rs745540669 Chr.7: 144379908-144379908 on Build GRCh38
rs760755618 Chr.7: 144379927-144379927 on Build GRCh38
rs749161855 Chr.7: 144379918-144379918 on Build GRCh38
rs903430105 Chr.7: 144379878-144379878 on Build GRCh38
rs750992168 Chr.7: 144379909-144379909 on Build GRCh38
rs774165136 Chr.7: 144379923-144379923 on Build GRCh38
rs754550612 Chr.7: 144380203-144380203 on Build GRCh38
rs1041680812 Chr.7: 144380143-144380143 on Build GRCh38
rs968745033 Chr.7: 144380114-144380114 on Build GRCh38
rs758897733 Chr.7: 144379891-144379891 on Build GRCh38
rs768592777 Chr.7: 144380047-144380047 on Build GRCh38
rs774793540 Chr.7: 144379861-144379862 on Build GRCh38
rs748003193 Chr.7: 144380051-144380051 on Build GRCh38
rs890972575 Chr.7: 144379820-144379820 on Build GRCh38
rs201944428 Chr.7: 144379860-144379860 on Build GRCh38
rs148911928 Chr.7: 144380045-144380045 on Build GRCh38
rs774972038 Chr.7: 144379926-144379926 on Build GRCh38
rs893100815 Chr.7: 144379830-144379830 on Build GRCh38
rs761964227 Chr.7: 144379852-144379852 on Build GRCh38
rs200117546 Chr.7: 144380077-144380077 on Build GRCh38
rs182264437 Chr.7: 144379913-144379913 on Build GRCh38
rs755821434 Chr.7: 144380042-144380042 on Build GRCh38
rs761017255 Chr.7: 144379795-144379795 on Build GRCh38
rs780707092 Chr.7: 144379805-144379805 on Build GRCh38
rs764752693 Chr.7: 144379866-144379872 on Build GRCh38
rs558598754 Chr.7: 144380237-144380237 on Build GRCh38
rs776561180 Chr.7: 144380055-144380055 on Build GRCh38
rs948927229 Chr.7: 144379877-144379877 on Build GRCh38
rs554793002 Chr.7: 144380124-144380124 on Build GRCh38
rs759733893 Chr.7: 144379935-144379935 on Build GRCh38
rs200654855 Chr.7: 144379986-144379986 on Build GRCh38
rs74598024 Chr.7: 144380074-144380074 on Build GRCh38
rs1050929805 Chr.7: 144379818-144379818 on Build GRCh38
rs948876211 Chr.7: 144380233-144380233 on Build GRCh38
rs7806384 Chr.7: 144380078-144380078 on Build GRCh38
rs765557375 Chr.7: 144379887-144379887 on Build GRCh38
rs776522129 Chr.7: 144379839-144379839 on Build GRCh38
rs780543294 Chr.7: 144379996-144379996 on Build GRCh38
rs753112035 Chr.7: 144379889-144379889 on Build GRCh38
rs916147638 Chr.7: 144379871-144379871 on Build GRCh38
rs749882356 Chr.7: 144380099-144380099 on Build GRCh38
rs376362386 Chr.7: 144379879-144379879 on Build GRCh38
rs1036359120 Chr.7: 144380201-144380201 on Build GRCh38
rs765592848 Chr.7: 144379899-144379899 on Build GRCh38
rs750003677 Chr.7: 144380012-144380012 on Build GRCh38
rs780850417 Chr.7: 144379907-144379907 on Build GRCh38
rs778868330 Chr.7: 144380048-144380048 on Build GRCh38
rs929855653 Chr.7: 144380028-144380028 on Build GRCh38
rs766561455 Chr.7: 144379876-144379876 on Build GRCh38
rs755327494 Chr.7: 144379881-144379881 on Build GRCh38
rs772098016 Chr.7: 144380053-144380053 on Build GRCh38
rs756733119 Chr.7: 144379904-144379904 on Build GRCh38
rs929911488 Chr.7: 144379784-144379784 on Build GRCh38
rs746461759 Chr.7: 144379902-144379902 on Build GRCh38
rs141781593 Chr.7: 144379892-144379892 on Build GRCh38
rs1031747776 Chr.7: 144380195-144380195 on Build GRCh38
rs777568718 Chr.7: 144380240-144380240 on Build GRCh38
rs762517622 Chr.7: 144379862-144379863 on Build GRCh38
rs763316091 Chr.7: 144379978-144379978 on Build GRCh38
rs201303398 Chr.7: 144380104-144380104 on Build GRCh38
rs759680608 Chr.7: 144380060-144380060 on Build GRCh38
rs755698995 Chr.7: 144380100-144380100 on Build GRCh38
rs755655039 Chr.7: 144380236-144380236 on Build GRCh38
rs977492563 Chr.7: 144379782-144379782 on Build GRCh38
rs377004169 Chr.7: 144379920-144379920 on Build GRCh38
rs10229915 Chr.7: 144380117-144380117 on Build GRCh38
rs903208857 Chr.7: 144380168-144380168 on Build GRCh38
rs762332689 Chr.7: 144380076-144380076 on Build GRCh38
rs768133674 Chr.7: 144380097-144380097 on Build GRCh38
rs762960756 Chr.7: 144379853-144379853 on Build GRCh38
rs769617324 Chr.7: 144379912-144379912 on Build GRCh38
rs1036782686 Chr.7: 144379974-144379974 on Build GRCh38
rs770944676 Chr.7: 144379928-144379928 on Build GRCh38
rs776813805 Chr.7: 144379930-144379930 on Build GRCh38
rs756752956 Chr.7: 144379990-144379990 on Build GRCh38
rs977858642 Chr.7: 144380137-144380137 on Build GRCh38
rs764608154 Chr.7: 144379985-144379985 on Build GRCh38
rs752092752 Chr.7: 144380220-144380220 on Build GRCh38
rs186573263 Chr.7: 144380026-144380026 on Build GRCh38
rs569308857 Chr.7: 144380212-144380212 on Build GRCh38
rs61734507 Chr.7: 144379966-144379966 on Build GRCh38

View Full Product Details