Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149872878-149873371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0E2
Entrez Gene ID
155066
Gene Name
ATPase H+ transporting V0 subunit e2
Gene Aliases
ATP6V0E2L, C7orf32
UniGene
Hs.556998
Gene Chromosome Location
Chr.7: 149872968-149880713 on Build GRCh38
Transcripts
NM_001100592.2 NM_001289990.1 NM_145230.3 NR_110612.1 NR_027040.1

Gene Symbol
ATP6V0E2-AS1
Entrez Gene ID
401431
Gene Name
ATP6V0E2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.556998
Gene Chromosome Location
Chr.7: 149867694-149873862 on Build GRCh38
Transcripts
NM_001100592.2 NM_001289990.1 NM_145230.3 NR_110612.1 NR_027040.1

SNP Detail

SNP ID Chromosome Location
rs185874861 Chr.7: 149872951-149872951 on Build GRCh38
rs991599001 Chr.7: 149873109-149873109 on Build GRCh38
rs990439742 Chr.7: 149873202-149873202 on Build GRCh38
rs557642169 Chr.7: 149872969-149872969 on Build GRCh38
rs1032786075 Chr.7: 149873326-149873326 on Build GRCh38
rs1029758261 Chr.7: 149873142-149873142 on Build GRCh38
rs927755073 Chr.7: 149873298-149873300 on Build GRCh38
rs924905953 Chr.7: 149872993-149872993 on Build GRCh38
rs574034840 Chr.7: 149872994-149872994 on Build GRCh38
rs941333997 Chr.7: 149873116-149873116 on Build GRCh38
rs562164158 Chr.7: 149873040-149873040 on Build GRCh38
rs982644276 Chr.7: 149873308-149873308 on Build GRCh38
rs1019969961 Chr.7: 149873017-149873017 on Build GRCh38
rs757448693 Chr.7: 149872927-149872927 on Build GRCh38
rs183202835 Chr.7: 149873234-149873234 on Build GRCh38
rs1030243183 Chr.7: 149872911-149872914 on Build GRCh38
rs978573126 Chr.7: 149873048-149873048 on Build GRCh38
rs939213190 Chr.7: 149873078-149873078 on Build GRCh38
rs1032304413 Chr.7: 149873187-149873187 on Build GRCh38
rs980902000 Chr.7: 149873244-149873244 on Build GRCh38
rs113211607 Chr.7: 149872982-149872982 on Build GRCh38
rs978046403 Chr.7: 149872954-149872954 on Build GRCh38
rs60859340 Chr.7: 149873141-149873141 on Build GRCh38
rs958932695 Chr.7: 149873102-149873102 on Build GRCh38
rs559799224 Chr.7: 149873324-149873324 on Build GRCh38
rs925686548 Chr.7: 149873054-149873054 on Build GRCh38
rs941357686 Chr.7: 149873178-149873178 on Build GRCh38
rs373850218 Chr.7: 149872910-149872910 on Build GRCh38
rs1047271321 Chr.7: 149873159-149873159 on Build GRCh38
rs999033656 Chr.7: 149873178-149873179 on Build GRCh38
rs908545168 Chr.7: 149873165-149873165 on Build GRCh38
rs532757761 Chr.7: 149873134-149873136 on Build GRCh38
rs891263372 Chr.7: 149873174-149873174 on Build GRCh38
rs1050261869 Chr.7: 149873229-149873229 on Build GRCh38
rs139836525 Chr.7: 149873032-149873032 on Build GRCh38
rs571831845 Chr.7: 149872907-149872907 on Build GRCh38
rs12703054 Chr.7: 149873011-149873011 on Build GRCh38
rs111896197 Chr.7: 149873025-149873025 on Build GRCh38
rs1028936006 Chr.7: 149873000-149873000 on Build GRCh38
rs902882164 Chr.7: 149873321-149873321 on Build GRCh38
rs766109509 Chr.7: 149872965-149872965 on Build GRCh38
rs950318340 Chr.7: 149873143-149873143 on Build GRCh38
rs1008789675 Chr.7: 149873016-149873016 on Build GRCh38
rs189248446 Chr.7: 149873305-149873305 on Build GRCh38
rs751039105 Chr.7: 149873065-149873071 on Build GRCh38
rs747650779 Chr.7: 149873129-149873129 on Build GRCh38
rs1020617737 Chr.7: 149873314-149873314 on Build GRCh38
rs545105052 Chr.7: 149873105-149873105 on Build GRCh38
rs758473318 Chr.7: 149873347-149873347 on Build GRCh38
rs544038637 Chr.7: 149873197-149873197 on Build GRCh38
rs190728024 Chr.7: 149873188-149873188 on Build GRCh38
rs377044466 Chr.7: 149872968-149872968 on Build GRCh38
rs552577136 Chr.7: 149873019-149873019 on Build GRCh38
rs914945132 Chr.7: 149873027-149873027 on Build GRCh38
rs899896048 Chr.7: 149873220-149873220 on Build GRCh38
rs6957652 Chr.7: 149873224-149873224 on Build GRCh38
rs1047399213 Chr.7: 149873090-149873090 on Build GRCh38

View Full Product Details