Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149875507-149875942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0E2
Entrez Gene ID
155066
Gene Name
ATPase H+ transporting V0 subunit e2
Gene Aliases
ATP6V0E2L, C7orf32
UniGene
Hs.556998
Gene Chromosome Location
Chr.7: 149872968-149880713 on Build GRCh38
Transcripts
NM_001100592.2 NM_001289990.1 NM_145230.3 NR_110612.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
746045 c.297C>G p.L99L Chr.7: 149875643-149875643 on Build GRCh38
1150614 c.297C>G p.L99L Chr.7: 149875643-149875643 on Build GRCh38
5551405 c.288C>A p.C96* Chr.7: 149875634-149875634 on Build GRCh38
1596880 c.280G>A p.A94T Chr.7: 149875626-149875626 on Build GRCh38
1596881 c.280G>A p.A94T Chr.7: 149875626-149875626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750236781 Chr.7: 149875641-149875641 on Build GRCh38
rs915380870 Chr.7: 149875711-149875711 on Build GRCh38
rs370114996 Chr.7: 149875691-149875691 on Build GRCh38
rs761485568 Chr.7: 149875629-149875629 on Build GRCh38
rs752201135 Chr.7: 149875569-149875569 on Build GRCh38
rs762495571 Chr.7: 149875575-149875575 on Build GRCh38
rs994395225 Chr.7: 149875561-149875561 on Build GRCh38
rs967047434 Chr.7: 149875857-149875857 on Build GRCh38
rs781442627 Chr.7: 149875694-149875694 on Build GRCh38
rs988413554 Chr.7: 149875794-149875794 on Build GRCh38
rs1006785915 Chr.7: 149875576-149875576 on Build GRCh38
rs532694680 Chr.7: 149875827-149875827 on Build GRCh38
rs10240203 Chr.7: 149875753-149875753 on Build GRCh38
rs966314136 Chr.7: 149875781-149875781 on Build GRCh38
rs775151382 Chr.7: 149875579-149875579 on Build GRCh38
rs780009148 Chr.7: 149875605-149875605 on Build GRCh38
rs372713158 Chr.7: 149875720-149875720 on Build GRCh38
rs376127751 Chr.7: 149875611-149875611 on Build GRCh38
rs189122070 Chr.7: 149875706-149875706 on Build GRCh38
rs200965623 Chr.7: 149875609-149875609 on Build GRCh38
rs140812225 Chr.7: 149875617-149875617 on Build GRCh38
rs184364483 Chr.7: 149875660-149875660 on Build GRCh38
rs897021368 Chr.7: 149875544-149875544 on Build GRCh38
rs978481912 Chr.7: 149875879-149875879 on Build GRCh38
rs868484302 Chr.7: 149875734-149875734 on Build GRCh38
rs369379550 Chr.7: 149875620-149875620 on Build GRCh38
rs759988807 Chr.7: 149875551-149875551 on Build GRCh38
rs765723927 Chr.7: 149875564-149875564 on Build GRCh38
rs753885763 Chr.7: 149875671-149875674 on Build GRCh38
rs377141036 Chr.7: 149875722-149875722 on Build GRCh38
rs151277033 Chr.7: 149875583-149875583 on Build GRCh38
rs560560615 Chr.7: 149875790-149875790 on Build GRCh38
rs750156855 Chr.7: 149875662-149875676 on Build GRCh38
rs750195320 Chr.7: 149875587-149875587 on Build GRCh38
rs994870400 Chr.7: 149875684-149875684 on Build GRCh38
rs369744898 Chr.7: 149875628-149875628 on Build GRCh38
rs754896597 Chr.7: 149875666-149875666 on Build GRCh38
rs191859295 Chr.7: 149875625-149875625 on Build GRCh38
rs371535588 Chr.7: 149875607-149875607 on Build GRCh38
rs375592794 Chr.7: 149875677-149875677 on Build GRCh38
rs763806854 Chr.7: 149875578-149875578 on Build GRCh38
rs778026623 Chr.7: 149875668-149875668 on Build GRCh38
rs201500750 Chr.7: 149875627-149875627 on Build GRCh38
rs375345690 Chr.7: 149875817-149875817 on Build GRCh38
rs751203903 Chr.7: 149875580-149875580 on Build GRCh38
rs552618868 Chr.7: 149875918-149875918 on Build GRCh38
rs367724684 Chr.7: 149875593-149875593 on Build GRCh38
rs897888214 Chr.7: 149875849-149875849 on Build GRCh38
rs762511980 Chr.7: 149875626-149875626 on Build GRCh38
rs553730534 Chr.7: 149875536-149875536 on Build GRCh38
rs1019946885 Chr.7: 149875840-149875840 on Build GRCh38
rs759920372 Chr.7: 149875621-149875621 on Build GRCh38
rs747235217 Chr.7: 149875619-149875619 on Build GRCh38
rs373442819 Chr.7: 149875663-149875663 on Build GRCh38
rs376372613 Chr.7: 149875586-149875586 on Build GRCh38
rs930724103 Chr.7: 149875894-149875894 on Build GRCh38
rs1019583149 Chr.7: 149875767-149875767 on Build GRCh38
rs939341993 Chr.7: 149875785-149875785 on Build GRCh38
rs755931386 Chr.7: 149875648-149875648 on Build GRCh38

View Full Product Details