Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150626798-150627303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP6
Entrez Gene ID
474344
Gene Name
GTPase, IMAP family member 6
Gene Aliases
IAN-2, IAN-6, IAN2, IAN6
UniGene
Hs.647105
Gene Chromosome Location
Chr.7: 150625375-150632648 on Build GRCh38
Transcripts
NM_001244071.1 NM_001244072.1 NM_024711.5

SNP Detail

SNP ID Chromosome Location
rs993949991 Chr.7: 150627187-150627187 on Build GRCh38
rs1026621285 Chr.7: 150626960-150626960 on Build GRCh38
rs117532299 Chr.7: 150627040-150627040 on Build GRCh38
rs1026716072 Chr.7: 150627197-150627197 on Build GRCh38
rs371161152 Chr.7: 150627261-150627261 on Build GRCh38
rs905783716 Chr.7: 150627103-150627103 on Build GRCh38
rs551690452 Chr.7: 150627036-150627036 on Build GRCh38
rs558597334 Chr.7: 150627217-150627217 on Build GRCh38
rs369635168 Chr.7: 150627041-150627041 on Build GRCh38
rs1045065549 Chr.7: 150627101-150627101 on Build GRCh38
rs530647083 Chr.7: 150626882-150626882 on Build GRCh38
rs961424393 Chr.7: 150626924-150626924 on Build GRCh38
rs920748058 Chr.7: 150627160-150627160 on Build GRCh38
rs11762727 Chr.7: 150627069-150627069 on Build GRCh38
rs952410986 Chr.7: 150626988-150626988 on Build GRCh38
rs551294807 Chr.7: 150627030-150627030 on Build GRCh38
rs977326972 Chr.7: 150627034-150627034 on Build GRCh38
rs117148907 Chr.7: 150627039-150627039 on Build GRCh38
rs376178326 Chr.7: 150627093-150627093 on Build GRCh38
rs1043309659 Chr.7: 150627234-150627234 on Build GRCh38
rs771973448 Chr.7: 150627235-150627235 on Build GRCh38
rs905779837 Chr.7: 150626879-150626879 on Build GRCh38
rs1040931033 Chr.7: 150626969-150626969 on Build GRCh38
rs538707568 Chr.7: 150627203-150627203 on Build GRCh38
rs896960681 Chr.7: 150627185-150627185 on Build GRCh38
rs1012240106 Chr.7: 150626849-150626849 on Build GRCh38
rs778925702 Chr.7: 150627135-150627135 on Build GRCh38
rs1001607353 Chr.7: 150627071-150627071 on Build GRCh38
rs150085269 Chr.7: 150626948-150626948 on Build GRCh38
rs1006518344 Chr.7: 150627201-150627201 on Build GRCh38
rs1035497895 Chr.7: 150626915-150626915 on Build GRCh38
rs993854889 Chr.7: 150626946-150626946 on Build GRCh38
rs891674841 Chr.7: 150627005-150627005 on Build GRCh38
rs528532470 Chr.7: 150626852-150626852 on Build GRCh38
rs973998976 Chr.7: 150627144-150627144 on Build GRCh38
rs943994656 Chr.7: 150626956-150626956 on Build GRCh38
rs551385309 Chr.7: 150626854-150626854 on Build GRCh38
rs985074942 Chr.7: 150626989-150626989 on Build GRCh38
rs948076070 Chr.7: 150627095-150627095 on Build GRCh38
rs1002754719 Chr.7: 150627154-150627154 on Build GRCh38
rs901364967 Chr.7: 150627164-150627184 on Build GRCh38
rs924175439 Chr.7: 150627272-150627272 on Build GRCh38
rs545542889 Chr.7: 150626981-150626981 on Build GRCh38
rs528075626 Chr.7: 150627164-150627164 on Build GRCh38
rs952494747 Chr.7: 150627162-150627162 on Build GRCh38
rs115125549 Chr.7: 150627226-150627226 on Build GRCh38
rs945955955 Chr.7: 150627233-150627233 on Build GRCh38
rs911495989 Chr.7: 150626993-150626993 on Build GRCh38
rs1049401503 Chr.7: 150626877-150626877 on Build GRCh38
rs186991101 Chr.7: 150627079-150627079 on Build GRCh38
rs566586828 Chr.7: 150627198-150627198 on Build GRCh38
rs904337128 Chr.7: 150627253-150627253 on Build GRCh38

View Full Product Details