Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150692689-150693185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP2
Entrez Gene ID
26157
Gene Name
GTPase, IMAP family member 2
Gene Aliases
HIMAP2, IAN12, IMAP2
UniGene
Hs.647071
Gene Chromosome Location
Chr.7: 150685706-150693641 on Build GRCh38
Transcripts
NM_015660.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7134205 c.548A>G p.N183S Chr.7: 150692834-150692834 on Build GRCh38
485042 c.464C>A p.S155Y Chr.7: 150692750-150692750 on Build GRCh38
1087251 c.484G>A p.D162N Chr.7: 150692770-150692770 on Build GRCh38
3636060 c.411C>T p.A137A Chr.7: 150692697-150692697 on Build GRCh38
4964657 c.481C>T p.H161Y Chr.7: 150692767-150692767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148143220 Chr.7: 150692985-150692985 on Build GRCh38
rs139270881 Chr.7: 150692952-150692952 on Build GRCh38
rs375623579 Chr.7: 150692728-150692728 on Build GRCh38
rs776373050 Chr.7: 150692975-150692975 on Build GRCh38
rs745906034 Chr.7: 150692727-150692727 on Build GRCh38
rs979470702 Chr.7: 150693138-150693138 on Build GRCh38
rs772943762 Chr.7: 150692840-150692840 on Build GRCh38
rs765107258 Chr.7: 150693098-150693099 on Build GRCh38
rs912122284 Chr.7: 150692802-150692802 on Build GRCh38
rs986222355 Chr.7: 150693069-150693069 on Build GRCh38
rs775981582 Chr.7: 150692848-150692848 on Build GRCh38
rs550994929 Chr.7: 150692818-150692818 on Build GRCh38
rs752650056 Chr.7: 150692860-150692860 on Build GRCh38
rs776842070 Chr.7: 150692900-150692900 on Build GRCh38
rs961994529 Chr.7: 150693000-150693000 on Build GRCh38
rs376750400 Chr.7: 150692883-150692883 on Build GRCh38
rs752453732 Chr.7: 150692777-150692777 on Build GRCh38
rs1052411631 Chr.7: 150692825-150692825 on Build GRCh38
rs368810304 Chr.7: 150692934-150692934 on Build GRCh38
rs918789969 Chr.7: 150692764-150692764 on Build GRCh38
rs759125462 Chr.7: 150692852-150692852 on Build GRCh38
rs997879336 Chr.7: 150692937-150692937 on Build GRCh38
rs769276327 Chr.7: 150692733-150692733 on Build GRCh38
rs756154784 Chr.7: 150692807-150692807 on Build GRCh38
rs751426384 Chr.7: 150692871-150692871 on Build GRCh38
rs138926685 Chr.7: 150692891-150692891 on Build GRCh38
rs772114017 Chr.7: 150692880-150692914 on Build GRCh38
rs774847313 Chr.7: 150692734-150692734 on Build GRCh38
rs370076034 Chr.7: 150692823-150692823 on Build GRCh38
rs761466415 Chr.7: 150693146-150693146 on Build GRCh38
rs145172862 Chr.7: 150692781-150692781 on Build GRCh38
rs201183603 Chr.7: 150692990-150692990 on Build GRCh38
rs985562937 Chr.7: 150692749-150692750 on Build GRCh38
rs748484310 Chr.7: 150692826-150692826 on Build GRCh38
rs778956614 Chr.7: 150692750-150692750 on Build GRCh38
rs35516375 Chr.7: 150692731-150692732 on Build GRCh38
rs146899946 Chr.7: 150692839-150692839 on Build GRCh38
rs765269305 Chr.7: 150692772-150692772 on Build GRCh38
rs2075078 Chr.7: 150692768-150692768 on Build GRCh38
rs375868708 Chr.7: 150693126-150693126 on Build GRCh38
rs930155193 Chr.7: 150693120-150693120 on Build GRCh38
rs771345666 Chr.7: 150693044-150693044 on Build GRCh38
rs913885573 Chr.7: 150692843-150692843 on Build GRCh38
rs61730412 Chr.7: 150692748-150692748 on Build GRCh38
rs141987826 Chr.7: 150692819-150692819 on Build GRCh38
rs17173567 Chr.7: 150692741-150692741 on Build GRCh38
rs749297178 Chr.7: 150692896-150692896 on Build GRCh38
rs149013037 Chr.7: 150692925-150692925 on Build GRCh38
rs778464004 Chr.7: 150693041-150693041 on Build GRCh38
rs774970470 Chr.7: 150693114-150693114 on Build GRCh38
rs769304406 Chr.7: 150692976-150692976 on Build GRCh38
rs746101312 Chr.7: 150692795-150692795 on Build GRCh38
rs766111422 Chr.7: 150692846-150692846 on Build GRCh38
rs759437182 Chr.7: 150693008-150693009 on Build GRCh38
rs766471931 Chr.7: 150693012-150693012 on Build GRCh38
rs1030203201 Chr.7: 150692947-150692947 on Build GRCh38
rs779995737 Chr.7: 150692808-150692808 on Build GRCh38
rs752881228 Chr.7: 150693035-150693035 on Build GRCh38
rs142845786 Chr.7: 150693037-150693037 on Build GRCh38
rs146107030 Chr.7: 150693080-150693080 on Build GRCh38
rs760238269 Chr.7: 150692844-150692844 on Build GRCh38
rs959726119 Chr.7: 150693050-150693050 on Build GRCh38
rs868749339 Chr.7: 150693142-150693142 on Build GRCh38
rs763378192 Chr.7: 150692732-150692732 on Build GRCh38
rs1048532412 Chr.7: 150692771-150692771 on Build GRCh38
rs267601409 Chr.7: 150692758-150692758 on Build GRCh38
rs372417348 Chr.7: 150692861-150692861 on Build GRCh38
rs748392759 Chr.7: 150692848-150692848 on Build GRCh38
rs779140676 Chr.7: 150693030-150693030 on Build GRCh38
rs765115379 Chr.7: 150692854-150692854 on Build GRCh38
rs372579875 Chr.7: 150693025-150693025 on Build GRCh38
rs142150251 Chr.7: 150692742-150692742 on Build GRCh38
rs150902506 Chr.7: 150692785-150692785 on Build GRCh38
rs776009895 Chr.7: 150692729-150692729 on Build GRCh38
rs1055657574 Chr.7: 150692928-150692928 on Build GRCh38
rs773313268 Chr.7: 150693001-150693001 on Build GRCh38
rs770458368 Chr.7: 150692970-150692970 on Build GRCh38
rs765810038 Chr.7: 150692759-150692759 on Build GRCh38
rs866130365 Chr.7: 150693092-150693092 on Build GRCh38
rs754113066 Chr.7: 150693018-150693018 on Build GRCh38
rs762317176 Chr.7: 150692736-150692736 on Build GRCh38
rs993523333 Chr.7: 150693079-150693079 on Build GRCh38
rs918822505 Chr.7: 150693097-150693097 on Build GRCh38
rs754857410 Chr.7: 150692769-150692769 on Build GRCh38
rs776096569 Chr.7: 150693144-150693144 on Build GRCh38
rs763883328 Chr.7: 150692869-150692869 on Build GRCh38
rs774376749 Chr.7: 150693127-150693127 on Build GRCh38
rs200032624 Chr.7: 150692999-150692999 on Build GRCh38
rs751294485 Chr.7: 150692997-150692997 on Build GRCh38
rs769504928 Chr.7: 150693113-150693113 on Build GRCh38
rs561894366 Chr.7: 150693096-150693096 on Build GRCh38
rs773696604 Chr.7: 150692940-150692940 on Build GRCh38
rs149977514 Chr.7: 150693099-150693099 on Build GRCh38
rs779515506 Chr.7: 150692923-150692923 on Build GRCh38

View Full Product Details