Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150742344-150742846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP1-GIMAP5
Entrez Gene ID
100527949
Gene Name
GIMAP1-GIMAP5 readthrough
Gene Aliases
-
UniGene
Hs.647079
Gene Chromosome Location
Chr.7: 150716557-150743649 on Build GRCh38
Transcripts
NM_001199577.1 NM_001303630.1 NM_018384.4

Gene Symbol
GIMAP5
Entrez Gene ID
55340
Gene Name
GTPase, IMAP family member 5
Gene Aliases
HIMAP3, IAN-5, IAN4, IAN4L1, IAN5, IMAP3, IROD
UniGene
Hs.647079
Gene Chromosome Location
Chr.7: 150737348-150743649 on Build GRCh38
Transcripts
NM_001199577.1 NM_001303630.1 NM_018384.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5406949 c.609C>T p.L203L Chr.7: 150742748-150742748 on Build GRCh38
6671010 c.286C>A p.L96M Chr.7: 150742425-150742425 on Build GRCh38
387858 c.252C>T p.P84P Chr.7: 150742391-150742391 on Build GRCh38
5506262 c.327G>C p.P109P Chr.7: 150742466-150742466 on Build GRCh38
3260828 c.228G>A p.R76R Chr.7: 150742367-150742367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927435790 Chr.7: 150742465-150742465 on Build GRCh38
rs753052223 Chr.7: 150742496-150742496 on Build GRCh38
rs756881232 Chr.7: 150742571-150742571 on Build GRCh38
rs768342609 Chr.7: 150742520-150742520 on Build GRCh38
rs61751036 Chr.7: 150742736-150742736 on Build GRCh38
rs552913796 Chr.7: 150742753-150742753 on Build GRCh38
rs866287630 Chr.7: 150742789-150742789 on Build GRCh38
rs772761622 Chr.7: 150742376-150742376 on Build GRCh38
rs749165559 Chr.7: 150742591-150742591 on Build GRCh38
rs376880387 Chr.7: 150742770-150742770 on Build GRCh38
rs778817495 Chr.7: 150742608-150742608 on Build GRCh38
rs759323787 Chr.7: 150742814-150742814 on Build GRCh38
rs576606315 Chr.7: 150742502-150742502 on Build GRCh38
rs756243315 Chr.7: 150742775-150742775 on Build GRCh38
rs777309676 Chr.7: 150742439-150742439 on Build GRCh38
rs1051756770 Chr.7: 150742549-150742549 on Build GRCh38
rs746075907 Chr.7: 150742694-150742694 on Build GRCh38
rs755612482 Chr.7: 150742417-150742417 on Build GRCh38
rs770097013 Chr.7: 150742711-150742711 on Build GRCh38
rs375326276 Chr.7: 150742554-150742554 on Build GRCh38
rs745789935 Chr.7: 150742448-150742448 on Build GRCh38
rs918681913 Chr.7: 150742767-150742767 on Build GRCh38
rs749909322 Chr.7: 150742577-150742577 on Build GRCh38
rs180974779 Chr.7: 150742786-150742786 on Build GRCh38
rs776560370 Chr.7: 150742387-150742387 on Build GRCh38
rs753161728 Chr.7: 150742559-150742559 on Build GRCh38
rs765713700 Chr.7: 150742558-150742558 on Build GRCh38
rs770802947 Chr.7: 150742381-150742381 on Build GRCh38
rs746191943 Chr.7: 150742759-150742759 on Build GRCh38
rs765212870 Chr.7: 150742760-150742760 on Build GRCh38
rs61751040 Chr.7: 150742466-150742466 on Build GRCh38
rs754403916 Chr.7: 150742414-150742414 on Build GRCh38
rs765904238 Chr.7: 150742799-150742799 on Build GRCh38
rs774356807 Chr.7: 150742530-150742530 on Build GRCh38
rs72650695 Chr.7: 150742750-150742750 on Build GRCh38
rs746872238 Chr.7: 150742773-150742773 on Build GRCh38
rs146646600 Chr.7: 150742388-150742388 on Build GRCh38
rs199678719 Chr.7: 150742741-150742741 on Build GRCh38
rs569476446 Chr.7: 150742433-150742433 on Build GRCh38
rs773424993 Chr.7: 150742787-150742787 on Build GRCh38
rs761120317 Chr.7: 150742798-150742798 on Build GRCh38
rs141446982 Chr.7: 150742392-150742392 on Build GRCh38
rs776346628 Chr.7: 150742801-150742801 on Build GRCh38
rs746455496 Chr.7: 150742440-150742440 on Build GRCh38
rs752671075 Chr.7: 150742769-150742769 on Build GRCh38
rs373837681 Chr.7: 150742459-150742459 on Build GRCh38
rs777685680 Chr.7: 150742693-150742693 on Build GRCh38
rs756285302 Chr.7: 150742413-150742413 on Build GRCh38
rs1021223904 Chr.7: 150742746-150742746 on Build GRCh38
rs763339553 Chr.7: 150742643-150742643 on Build GRCh38
rs779012038 Chr.7: 150742678-150742678 on Build GRCh38
rs779878978 Chr.7: 150742507-150742507 on Build GRCh38
rs765075238 Chr.7: 150742391-150742391 on Build GRCh38
rs150441443 Chr.7: 150742504-150742504 on Build GRCh38
rs768736186 Chr.7: 150742780-150742780 on Build GRCh38
rs200235816 Chr.7: 150742537-150742537 on Build GRCh38
rs74162071 Chr.7: 150742400-150742400 on Build GRCh38
rs765503398 Chr.7: 150742431-150742431 on Build GRCh38
rs780845085 Chr.7: 150742574-150742574 on Build GRCh38
rs370323356 Chr.7: 150742379-150742379 on Build GRCh38
rs367554935 Chr.7: 150742782-150742782 on Build GRCh38
rs201011395 Chr.7: 150742476-150742476 on Build GRCh38
rs543877528 Chr.7: 150742562-150742562 on Build GRCh38
rs368965016 Chr.7: 150742548-150742548 on Build GRCh38
rs149571417 Chr.7: 150742566-150742566 on Build GRCh38
rs183707481 Chr.7: 150742618-150742618 on Build GRCh38
rs773691352 Chr.7: 150742745-150742745 on Build GRCh38
rs769731120 Chr.7: 150742506-150742506 on Build GRCh38
rs758285735 Chr.7: 150742686-150742686 on Build GRCh38
rs776825281 Chr.7: 150742553-150742553 on Build GRCh38
rs751872887 Chr.7: 150742654-150742654 on Build GRCh38
rs749055048 Chr.7: 150742517-150742517 on Build GRCh38
rs562399062 Chr.7: 150742612-150742612 on Build GRCh38
rs147182394 Chr.7: 150742672-150742672 on Build GRCh38
rs932844423 Chr.7: 150742518-150742518 on Build GRCh38
rs569763678 Chr.7: 150742712-150742712 on Build GRCh38
rs572573174 Chr.7: 150742377-150742377 on Build GRCh38
rs767730558 Chr.7: 150742470-150742470 on Build GRCh38
rs753235405 Chr.7: 150742756-150742756 on Build GRCh38
rs759011 Chr.7: 150742412-150742412 on Build GRCh38
rs769667762 Chr.7: 150742457-150742457 on Build GRCh38
rs758470990 Chr.7: 150742500-150742500 on Build GRCh38
rs748514363 Chr.7: 150742428-150742428 on Build GRCh38
rs138178760 Chr.7: 150742611-150742611 on Build GRCh38
rs775709068 Chr.7: 150742642-150742642 on Build GRCh38
rs747907400 Chr.7: 150742683-150742683 on Build GRCh38
rs767673563 Chr.7: 150742409-150742409 on Build GRCh38
rs770429254 Chr.7: 150742445-150742445 on Build GRCh38
rs769112260 Chr.7: 150742732-150742732 on Build GRCh38
rs980646452 Chr.7: 150742633-150742633 on Build GRCh38
rs61751028 Chr.7: 150742649-150742649 on Build GRCh38
rs755491294 Chr.7: 150742477-150742477 on Build GRCh38
rs140492772 Chr.7: 150742472-150742472 on Build GRCh38
rs372355155 Chr.7: 150742671-150742671 on Build GRCh38
rs760773018 Chr.7: 150742475-150742475 on Build GRCh38
rs769752170 Chr.7: 150742638-150742638 on Build GRCh38
rs61750977 Chr.7: 150742581-150742581 on Build GRCh38
rs552901583 Chr.7: 150742424-150742424 on Build GRCh38
rs760969277 Chr.7: 150742546-150742546 on Build GRCh38
rs574994840 Chr.7: 150742809-150742809 on Build GRCh38
rs755852343 Chr.7: 150742666-150742666 on Build GRCh38
rs768703802 Chr.7: 150742600-150742600 on Build GRCh38
rs375450342 Chr.7: 150742757-150742757 on Build GRCh38
rs748105968 Chr.7: 150742824-150742824 on Build GRCh38
rs750055689 Chr.7: 150742665-150742665 on Build GRCh38
rs749408342 Chr.7: 150742714-150742714 on Build GRCh38
rs765800153 Chr.7: 150742670-150742670 on Build GRCh38
rs780355836 Chr.7: 150742779-150742779 on Build GRCh38
rs368627191 Chr.7: 150742569-150742569 on Build GRCh38
rs202237347 Chr.7: 150742503-150742503 on Build GRCh38
rs765707928 Chr.7: 150742491-150742491 on Build GRCh38

View Full Product Details