Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151014076-151014581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG9B
Entrez Gene ID
285973
Gene Name
autophagy related 9B
Gene Aliases
APG9L2, NOS3AS, SONE
UniGene
Hs.707300
Gene Chromosome Location
Chr.7: 151012209-151024499 on Build GRCh38
Transcripts
NR_073169.1 NR_133652.1 XM_011516065.1 XM_011516066.2 NM_000603.4 XM_017012232.1 XM_017012233.1

Gene Symbol
NOS3
Entrez Gene ID
4846
Gene Name
nitric oxide synthase 3
Gene Aliases
ECNOS, eNOS
UniGene
Hs.647092
Gene Chromosome Location
Chr.7: 150991056-151014599 on Build GRCh38
Transcripts
NR_073169.1 NR_133652.1 XM_011516065.1 XM_011516066.2 NM_000603.4 XM_017012232.1 XM_017012233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1622555 c.3526C>G p.Q1176E Chr.7: 151014083-151014083 on Build GRCh38
4587251 c.3593C>T p.S1198L Chr.7: 151014150-151014150 on Build GRCh38
6331558 c.3552G>T p.Q1184H Chr.7: 151014109-151014109 on Build GRCh38
6388751 c.3580G>A p.D1194N Chr.7: 151014137-151014137 on Build GRCh38
4587250 c.3562G>T p.A1188S Chr.7: 151014119-151014119 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1022609920 Chr.7: 151014444-151014450 on Build GRCh38
rs909456789 Chr.7: 151014548-151014548 on Build GRCh38
rs145823011 Chr.7: 151014104-151014104 on Build GRCh38
rs919189931 Chr.7: 151014279-151014279 on Build GRCh38
rs1001840124 Chr.7: 151014450-151014450 on Build GRCh38
rs915666995 Chr.7: 151014136-151014136 on Build GRCh38
rs1026128952 Chr.7: 151014482-151014482 on Build GRCh38
rs892696687 Chr.7: 151014141-151014141 on Build GRCh38
rs373133695 Chr.7: 151014145-151014145 on Build GRCh38
rs1052575056 Chr.7: 151014391-151014391 on Build GRCh38
rs550680447 Chr.7: 151014468-151014468 on Build GRCh38
rs964393220 Chr.7: 151014522-151014522 on Build GRCh38
rs564937301 Chr.7: 151014140-151014140 on Build GRCh38
rs746325833 Chr.7: 151014150-151014150 on Build GRCh38
rs998819295 Chr.7: 151014385-151014385 on Build GRCh38
rs770226032 Chr.7: 151014113-151014113 on Build GRCh38
rs942708871 Chr.7: 151014352-151014352 on Build GRCh38
rs1043978131 Chr.7: 151014438-151014438 on Build GRCh38
rs556568807 Chr.7: 151014490-151014490 on Build GRCh38
rs528444796 Chr.7: 151014342-151014342 on Build GRCh38
rs770846565 Chr.7: 151014147-151014147 on Build GRCh38
rs571644639 Chr.7: 151014423-151014423 on Build GRCh38
rs567097552 Chr.7: 151014543-151014543 on Build GRCh38
rs761225044 Chr.7: 151014118-151014118 on Build GRCh38
rs537135591 Chr.7: 151014428-151014428 on Build GRCh38
rs770455028 Chr.7: 151014156-151014156 on Build GRCh38
rs747320181 Chr.7: 151014146-151014146 on Build GRCh38
rs983759152 Chr.7: 151014506-151014506 on Build GRCh38
rs980965727 Chr.7: 151014178-151014178 on Build GRCh38
rs149818096 Chr.7: 151014101-151014101 on Build GRCh38
rs909247933 Chr.7: 151014224-151014224 on Build GRCh38
rs760123467 Chr.7: 151014108-151014108 on Build GRCh38
rs377732577 Chr.7: 151014457-151014457 on Build GRCh38
rs781344479 Chr.7: 151014148-151014148 on Build GRCh38
rs773809579 Chr.7: 151014181-151014181 on Build GRCh38
rs904866778 Chr.7: 151014446-151014446 on Build GRCh38
rs769133349 Chr.7: 151014177-151014177 on Build GRCh38
rs779787527 Chr.7: 151014123-151014123 on Build GRCh38
rs568163242 Chr.7: 151014305-151014305 on Build GRCh38
rs757520710 Chr.7: 151014215-151014215 on Build GRCh38
rs761543450 Chr.7: 151014192-151014192 on Build GRCh38
rs1022405149 Chr.7: 151014164-151014164 on Build GRCh38
rs1032005929 Chr.7: 151014395-151014395 on Build GRCh38
rs765016429 Chr.7: 151014556-151014557 on Build GRCh38
rs151197128 Chr.7: 151014175-151014175 on Build GRCh38
rs531094458 Chr.7: 151014557-151014559 on Build GRCh38
rs753827185 Chr.7: 151014210-151014210 on Build GRCh38
rs934164867 Chr.7: 151014371-151014371 on Build GRCh38
rs957273758 Chr.7: 151014411-151014411 on Build GRCh38
rs368256841 Chr.7: 151014206-151014206 on Build GRCh38
rs376063899 Chr.7: 151014516-151014516 on Build GRCh38
rs1010583238 Chr.7: 151014157-151014157 on Build GRCh38
rs984706873 Chr.7: 151014336-151014336 on Build GRCh38
rs781380889 Chr.7: 151014218-151014218 on Build GRCh38
rs890943593 Chr.7: 151014383-151014383 on Build GRCh38
rs549668191 Chr.7: 151014262-151014262 on Build GRCh38
rs376955666 Chr.7: 151014198-151014198 on Build GRCh38
rs749398455 Chr.7: 151014117-151014117 on Build GRCh38
rs762499907 Chr.7: 151014556-151014556 on Build GRCh38
rs560089145 Chr.7: 151014335-151014335 on Build GRCh38
rs142886510 Chr.7: 151014119-151014119 on Build GRCh38
rs901217390 Chr.7: 151014364-151014364 on Build GRCh38
rs117078328 Chr.7: 151014417-151014417 on Build GRCh38
rs761085505 Chr.7: 151014467-151014467 on Build GRCh38
rs985145510 Chr.7: 151014497-151014497 on Build GRCh38
rs753603291 Chr.7: 151014124-151014124 on Build GRCh38
rs529527080 Chr.7: 151014251-151014251 on Build GRCh38
rs766891052 Chr.7: 151014194-151014194 on Build GRCh38
rs148530358 Chr.7: 151014115-151014115 on Build GRCh38
rs769399425 Chr.7: 151014481-151014481 on Build GRCh38
rs777840587 Chr.7: 151014143-151014143 on Build GRCh38
rs541336826 Chr.7: 151014116-151014116 on Build GRCh38
rs754722607 Chr.7: 151014211-151014211 on Build GRCh38
rs776189124 Chr.7: 151014174-151014174 on Build GRCh38
rs892623024 Chr.7: 151014404-151014404 on Build GRCh38
rs910434175 Chr.7: 151014508-151014508 on Build GRCh38
rs1040199580 Chr.7: 151014361-151014361 on Build GRCh38
rs775291992 Chr.7: 151014162-151014162 on Build GRCh38
rs199762416 Chr.7: 151014213-151014213 on Build GRCh38

View Full Product Details