Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151015804-151016306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG9B
Entrez Gene ID
285973
Gene Name
autophagy related 9B
Gene Aliases
APG9L2, NOS3AS, SONE
UniGene
Hs.707300
Gene Chromosome Location
Chr.7: 151012209-151024499 on Build GRCh38
Transcripts
NM_001317056.1 NR_073169.1 NR_133652.1 XM_011516065.1 XM_011516066.2

SNP Detail

SNP ID Chromosome Location
rs143473622 Chr.7: 151016280-151016280 on Build GRCh38
rs771728228 Chr.7: 151015921-151015921 on Build GRCh38
rs1056493677 Chr.7: 151016232-151016232 on Build GRCh38
rs933082402 Chr.7: 151015836-151015836 on Build GRCh38
rs756199054 Chr.7: 151016057-151016057 on Build GRCh38
rs937279026 Chr.7: 151016031-151016031 on Build GRCh38
rs761367878 Chr.7: 151016212-151016214 on Build GRCh38
rs552138381 Chr.7: 151015872-151015872 on Build GRCh38
rs954364255 Chr.7: 151016278-151016278 on Build GRCh38
rs768205713 Chr.7: 151016119-151016119 on Build GRCh38
rs574506942 Chr.7: 151016199-151016199 on Build GRCh38
rs753925921 Chr.7: 151016227-151016227 on Build GRCh38
rs780295669 Chr.7: 151016061-151016061 on Build GRCh38
rs749713065 Chr.7: 151016074-151016074 on Build GRCh38
rs769253185 Chr.7: 151015941-151015941 on Build GRCh38
rs760161109 Chr.7: 151015931-151015931 on Build GRCh38
rs749819056 Chr.7: 151015877-151015877 on Build GRCh38
rs61910690 Chr.7: 151015902-151015902 on Build GRCh38
rs573312706 Chr.7: 151016093-151016093 on Build GRCh38
rs545593603 Chr.7: 151016110-151016110 on Build GRCh38
rs945919586 Chr.7: 151015992-151015992 on Build GRCh38
rs575892388 Chr.7: 151016144-151016144 on Build GRCh38
rs992690069 Chr.7: 151015844-151015844 on Build GRCh38
rs961044110 Chr.7: 151015826-151015826 on Build GRCh38
rs755066813 Chr.7: 151016256-151016256 on Build GRCh38
rs188063500 Chr.7: 151016207-151016207 on Build GRCh38
rs3918217 Chr.7: 151015893-151015893 on Build GRCh38
rs913104639 Chr.7: 151015841-151015841 on Build GRCh38
rs963395736 Chr.7: 151016098-151016098 on Build GRCh38
rs374704787 Chr.7: 151015972-151015972 on Build GRCh38
rs753207105 Chr.7: 151016267-151016267 on Build GRCh38
rs3918218 Chr.7: 151016143-151016143 on Build GRCh38
rs953262731 Chr.7: 151016171-151016171 on Build GRCh38
rs895130238 Chr.7: 151015887-151015887 on Build GRCh38
rs926878118 Chr.7: 151016211-151016212 on Build GRCh38
rs772375266 Chr.7: 151016135-151016135 on Build GRCh38
rs745911616 Chr.7: 151015866-151015866 on Build GRCh38
rs748527527 Chr.7: 151016134-151016134 on Build GRCh38
rs963997424 Chr.7: 151015951-151015951 on Build GRCh38
rs761002527 Chr.7: 151016206-151016206 on Build GRCh38
rs746726088 Chr.7: 151015929-151015929 on Build GRCh38
rs1029214680 Chr.7: 151016260-151016260 on Build GRCh38
rs752565788 Chr.7: 151015973-151015973 on Build GRCh38
rs752889707 Chr.7: 151015942-151015942 on Build GRCh38
rs904414827 Chr.7: 151015875-151015875 on Build GRCh38
rs372822402 Chr.7: 151016172-151016172 on Build GRCh38
rs1014789329 Chr.7: 151016087-151016087 on Build GRCh38
rs765936005 Chr.7: 151015949-151015949 on Build GRCh38
rs549932119 Chr.7: 151016244-151016244 on Build GRCh38
rs770366000 Chr.7: 151016147-151016147 on Build GRCh38
rs750618079 Chr.7: 151016205-151016205 on Build GRCh38
rs376001984 Chr.7: 151016202-151016202 on Build GRCh38
rs750741009 Chr.7: 151016008-151016008 on Build GRCh38
rs3918239 Chr.7: 151015878-151015878 on Build GRCh38
rs575751217 Chr.7: 151016131-151016133 on Build GRCh38
rs759349248 Chr.7: 151016175-151016175 on Build GRCh38
rs767840666 Chr.7: 151015994-151015994 on Build GRCh38
rs3918219 Chr.7: 151016182-151016182 on Build GRCh38
rs962179187 Chr.7: 151016095-151016095 on Build GRCh38
rs950890773 Chr.7: 151015915-151015915 on Build GRCh38
rs533839618 Chr.7: 151015923-151015923 on Build GRCh38
rs758167636 Chr.7: 151015976-151015976 on Build GRCh38
rs778191409 Chr.7: 151016170-151016170 on Build GRCh38
rs946979120 Chr.7: 151015970-151015970 on Build GRCh38
rs187407160 Chr.7: 151015954-151015954 on Build GRCh38
rs767186137 Chr.7: 151016246-151016248 on Build GRCh38
rs1004688252 Chr.7: 151016050-151016050 on Build GRCh38
rs978973382 Chr.7: 151016001-151016001 on Build GRCh38
rs35921696 Chr.7: 151015998-151015999 on Build GRCh38
rs759528412 Chr.7: 151015959-151015959 on Build GRCh38
rs1037521238 Chr.7: 151016274-151016274 on Build GRCh38
rs78171488 Chr.7: 151015876-151015876 on Build GRCh38
rs779465284 Chr.7: 151016262-151016262 on Build GRCh38
rs1051508069 Chr.7: 151015840-151015840 on Build GRCh38

View Full Product Details