Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151076817-151077321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTK
Entrez Gene ID
10922
Gene Name
Fas activated serine/threonine kinase
Gene Aliases
FAST
UniGene
Hs.647094
Gene Chromosome Location
Chr.7: 151076621-151080883 on Build GRCh38
Transcripts
NM_001258461.1 NM_006712.4 NM_033015.3 XM_005249932.1 XM_005249933.3 XM_011515761.2 XM_011515762.2 XM_011515763.1 XM_017011704.1 XM_017011705.1 XM_017011706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7321146 c.1545A>G p.L515L Chr.7: 151076830-151076830 on Build GRCh38
6570300 c.1390G>T p.A464S Chr.7: 151077138-151077138 on Build GRCh38
1488373 c.1491G>C p.S497S Chr.7: 151076964-151076964 on Build GRCh38
4952893 c.1283A>C p.Y428S Chr.7: 151077318-151077318 on Build GRCh38
7245666 c.1475G>A p.R492Q Chr.7: 151076980-151076980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749421055 Chr.7: 151077046-151077046 on Build GRCh38
rs747990923 Chr.7: 151076953-151076953 on Build GRCh38
rs958108560 Chr.7: 151077191-151077191 on Build GRCh38
rs201888609 Chr.7: 151077143-151077143 on Build GRCh38
rs532948471 Chr.7: 151077083-151077083 on Build GRCh38
rs752924153 Chr.7: 151077254-151077254 on Build GRCh38
rs754383437 Chr.7: 151076943-151076943 on Build GRCh38
rs906773220 Chr.7: 151077185-151077185 on Build GRCh38
rs184536752 Chr.7: 151076860-151076860 on Build GRCh38
rs535070707 Chr.7: 151077079-151077079 on Build GRCh38
rs746665750 Chr.7: 151077293-151077294 on Build GRCh38
rs924056895 Chr.7: 151077113-151077113 on Build GRCh38
rs768336858 Chr.7: 151077283-151077283 on Build GRCh38
rs753396971 Chr.7: 151077010-151077010 on Build GRCh38
rs950707927 Chr.7: 151077267-151077267 on Build GRCh38
rs762393572 Chr.7: 151077100-151077100 on Build GRCh38
rs755794955 Chr.7: 151076916-151076916 on Build GRCh38
rs778769725 Chr.7: 151076866-151076866 on Build GRCh38
rs768577800 Chr.7: 151076985-151076985 on Build GRCh38
rs758330052 Chr.7: 151077036-151077036 on Build GRCh38
rs368509732 Chr.7: 151076890-151076890 on Build GRCh38
rs906204673 Chr.7: 151076981-151076981 on Build GRCh38
rs750055870 Chr.7: 151076852-151076852 on Build GRCh38
rs781684166 Chr.7: 151077272-151077272 on Build GRCh38
rs375441194 Chr.7: 151077255-151077255 on Build GRCh38
rs750271006 Chr.7: 151077126-151077126 on Build GRCh38
rs373219100 Chr.7: 151077276-151077276 on Build GRCh38
rs375095180 Chr.7: 151076870-151076870 on Build GRCh38
rs774003292 Chr.7: 151077044-151077044 on Build GRCh38
rs557896050 Chr.7: 151077184-151077184 on Build GRCh38
rs941805088 Chr.7: 151077037-151077037 on Build GRCh38
rs751117999 Chr.7: 151077215-151077215 on Build GRCh38
rs759082855 Chr.7: 151077080-151077080 on Build GRCh38
rs868369726 Chr.7: 151077093-151077093 on Build GRCh38
rs201230993 Chr.7: 151077034-151077034 on Build GRCh38
rs761176338 Chr.7: 151077005-151077005 on Build GRCh38
rs200625890 Chr.7: 151077107-151077107 on Build GRCh38
rs1053952911 Chr.7: 151076889-151076889 on Build GRCh38
rs751459242 Chr.7: 151077112-151077112 on Build GRCh38
rs966898658 Chr.7: 151076887-151076887 on Build GRCh38
rs758134278 Chr.7: 151077257-151077258 on Build GRCh38
rs773130885 Chr.7: 151077292-151077292 on Build GRCh38
rs534704710 Chr.7: 151077149-151077149 on Build GRCh38
rs199875725 Chr.7: 151077110-151077110 on Build GRCh38
rs777346432 Chr.7: 151077276-151077277 on Build GRCh38
rs1053984090 Chr.7: 151077122-151077122 on Build GRCh38
rs35031898 Chr.7: 151076853-151076853 on Build GRCh38
rs781623037 Chr.7: 151076845-151076845 on Build GRCh38
rs371534765 Chr.7: 151077124-151077124 on Build GRCh38
rs750169376 Chr.7: 151077223-151077223 on Build GRCh38
rs768971500 Chr.7: 151077060-151077060 on Build GRCh38
rs775545148 Chr.7: 151076885-151076885 on Build GRCh38
rs748916422 Chr.7: 151077282-151077282 on Build GRCh38
rs748125112 Chr.7: 151076868-151076868 on Build GRCh38
rs766859417 Chr.7: 151077008-151077008 on Build GRCh38
rs755148604 Chr.7: 151077127-151077127 on Build GRCh38
rs370215160 Chr.7: 151077297-151077297 on Build GRCh38
rs149426387 Chr.7: 151077196-151077196 on Build GRCh38
rs765009890 Chr.7: 151077084-151077084 on Build GRCh38
rs150140464 Chr.7: 151077148-151077148 on Build GRCh38
rs370608397 Chr.7: 151076865-151076865 on Build GRCh38
rs772689989 Chr.7: 151077067-151077067 on Build GRCh38
rs754684863 Chr.7: 151077030-151077030 on Build GRCh38
rs771377198 Chr.7: 151077078-151077078 on Build GRCh38
rs755877080 Chr.7: 151077240-151077240 on Build GRCh38
rs773016707 Chr.7: 151076878-151076878 on Build GRCh38
rs200663684 Chr.7: 151077286-151077286 on Build GRCh38
rs371380337 Chr.7: 151076990-151076990 on Build GRCh38
rs374560885 Chr.7: 151076914-151076914 on Build GRCh38
rs576660474 Chr.7: 151077222-151077222 on Build GRCh38
rs778026436 Chr.7: 151077176-151077176 on Build GRCh38
rs753371057 Chr.7: 151076940-151076940 on Build GRCh38
rs748390462 Chr.7: 151077138-151077138 on Build GRCh38
rs775142551 Chr.7: 151077195-151077195 on Build GRCh38
rs200553873 Chr.7: 151076891-151076891 on Build GRCh38
rs140806409 Chr.7: 151077136-151077136 on Build GRCh38
rs145106916 Chr.7: 151076863-151076863 on Build GRCh38
rs779249636 Chr.7: 151077131-151077131 on Build GRCh38
rs986078441 Chr.7: 151077052-151077052 on Build GRCh38
rs757695276 Chr.7: 151077263-151077263 on Build GRCh38
rs747155740 Chr.7: 151077258-151077258 on Build GRCh38
rs200153094 Chr.7: 151076984-151076984 on Build GRCh38
rs367561144 Chr.7: 151076965-151076965 on Build GRCh38
rs35223428 Chr.7: 151077003-151077003 on Build GRCh38
rs752137739 Chr.7: 151077087-151077087 on Build GRCh38
rs369209406 Chr.7: 151077068-151077068 on Build GRCh38
rs769991300 Chr.7: 151076980-151076980 on Build GRCh38
rs201972478 Chr.7: 151077033-151077033 on Build GRCh38
rs746070762 Chr.7: 151076884-151076884 on Build GRCh38
rs771609832 Chr.7: 151077182-151077182 on Build GRCh38
rs139030865 Chr.7: 151077061-151077061 on Build GRCh38
rs756979216 Chr.7: 151077043-151077043 on Build GRCh38
rs776857942 Chr.7: 151077183-151077183 on Build GRCh38
rs746368037 Chr.7: 151077275-151077275 on Build GRCh38
rs761368870 Chr.7: 151077284-151077284 on Build GRCh38
rs35919349 Chr.7: 151077253-151077253 on Build GRCh38
rs761264423 Chr.7: 151077069-151077069 on Build GRCh38
rs747500824 Chr.7: 151077177-151077177 on Build GRCh38
rs768176226 Chr.7: 151076994-151076994 on Build GRCh38
rs2288648 Chr.7: 151077221-151077221 on Build GRCh38
rs775740504 Chr.7: 151077280-151077280 on Build GRCh38
rs748844181 Chr.7: 151076864-151076864 on Build GRCh38
rs1130560 Chr.7: 151077103-151077103 on Build GRCh38
rs758413763 Chr.7: 151076964-151076964 on Build GRCh38
rs778520792 Chr.7: 151076948-151076948 on Build GRCh38
rs1018902616 Chr.7: 151077288-151077288 on Build GRCh38
rs749945491 Chr.7: 151077293-151077293 on Build GRCh38
rs777747992 Chr.7: 151077039-151077039 on Build GRCh38
rs757207922 Chr.7: 151077116-151077116 on Build GRCh38
rs766198899 Chr.7: 151076934-151076934 on Build GRCh38
rs774510924 Chr.7: 151076989-151076989 on Build GRCh38
rs752472683 Chr.7: 151077176-151077177 on Build GRCh38
rs577321027 Chr.7: 151077190-151077190 on Build GRCh38
rs746874695 Chr.7: 151076967-151076967 on Build GRCh38

View Full Product Details