Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27734413-27734917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC25
Entrez Gene ID
55246
Gene Name
coiled-coil domain containing 25
Gene Aliases
-
UniGene
Hs.445512
Gene Chromosome Location
Chr.8: 27733316-27772656 on Build GRCh38
Transcripts
NM_001304529.1 NM_001304530.1 NM_001304532.1 NM_018246.2 NR_130761.1 NR_130762.1 XM_011544570.2 XM_011544571.2

SNP Detail

SNP ID Chromosome Location
rs1028946512 Chr.8: 27734787-27734787 on Build GRCh38
rs6150519 Chr.8: 27734589-27734590 on Build GRCh38
rs886087952 Chr.8: 27734608-27734608 on Build GRCh38
rs34032448 Chr.8: 27734629-27734630 on Build GRCh38
rs776739679 Chr.8: 27734454-27734454 on Build GRCh38
rs573787814 Chr.8: 27734609-27734609 on Build GRCh38
rs949148391 Chr.8: 27734584-27734584 on Build GRCh38
rs866563465 Chr.8: 27734594-27734594 on Build GRCh38
rs900437302 Chr.8: 27734654-27734654 on Build GRCh38
rs1016746304 Chr.8: 27734738-27734738 on Build GRCh38
rs185982050 Chr.8: 27734810-27734810 on Build GRCh38
rs73231085 Chr.8: 27734485-27734485 on Build GRCh38
rs869136133 Chr.8: 27734594-27734595 on Build GRCh38
rs1004019129 Chr.8: 27734731-27734731 on Build GRCh38
rs927580480 Chr.8: 27734518-27734518 on Build GRCh38
rs915480284 Chr.8: 27734577-27734577 on Build GRCh38
rs993405932 Chr.8: 27734572-27734572 on Build GRCh38
rs968680092 Chr.8: 27734776-27734776 on Build GRCh38
rs181606484 Chr.8: 27734732-27734732 on Build GRCh38
rs867548828 Chr.8: 27734589-27734589 on Build GRCh38
rs978747219 Chr.8: 27734777-27734777 on Build GRCh38
rs911991535 Chr.8: 27734890-27734890 on Build GRCh38
rs147645561 Chr.8: 27734642-27734642 on Build GRCh38
rs869272237 Chr.8: 27734590-27734591 on Build GRCh38
rs1020393969 Chr.8: 27734771-27734771 on Build GRCh38
rs35919665 Chr.8: 27734509-27734510 on Build GRCh38
rs375755206 Chr.8: 27734872-27734872 on Build GRCh38
rs1003665862 Chr.8: 27734644-27734644 on Build GRCh38
rs886914442 Chr.8: 27734693-27734693 on Build GRCh38
rs890330313 Chr.8: 27734727-27734727 on Build GRCh38
rs935520331 Chr.8: 27734536-27734536 on Build GRCh38
rs995972297 Chr.8: 27734675-27734675 on Build GRCh38
rs1037593640 Chr.8: 27734651-27734651 on Build GRCh38
rs1030588358 Chr.8: 27734698-27734698 on Build GRCh38
rs1010734971 Chr.8: 27734730-27734730 on Build GRCh38
rs958953786 Chr.8: 27734840-27734840 on Build GRCh38
rs1056225572 Chr.8: 27734647-27734647 on Build GRCh38
rs28362501 Chr.8: 27734593-27734594 on Build GRCh38
rs917640028 Chr.8: 27734578-27734578 on Build GRCh38
rs765455780 Chr.8: 27734486-27734486 on Build GRCh38
rs968729339 Chr.8: 27734468-27734468 on Build GRCh38
rs953405884 Chr.8: 27734793-27734793 on Build GRCh38
rs937483885 Chr.8: 27734540-27734540 on Build GRCh38
rs904898347 Chr.8: 27734621-27734621 on Build GRCh38

View Full Product Details