Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28028845-28029378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUGGC
Entrez Gene ID
389643
Gene Name
nuclear GTPase, germinal center associated
Gene Aliases
C8orf80, HMFN0672, SLIP-GC
UniGene
Hs.370129
Gene Chromosome Location
Chr.8: 28021964-28084068 on Build GRCh38
Transcripts
NM_001010906.1 XM_011544523.2 XM_011544524.2 XM_011544525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4668058 c.2045C>T p.A682V Chr.8: 28029375-28029375 on Build GRCh38
1098810 c.2087G>A p.R696Q Chr.8: 28029333-28029333 on Build GRCh38
6861093 c.2093T>G p.V698G Chr.8: 28029327-28029327 on Build GRCh38
1098814 c.2062G>T p.D688Y Chr.8: 28029358-28029358 on Build GRCh38
4825552 c.2131C>A p.Q711K Chr.8: 28029289-28029289 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055761495 Chr.8: 28029133-28029133 on Build GRCh38
rs201861945 Chr.8: 28029231-28029231 on Build GRCh38
rs749369973 Chr.8: 28029137-28029137 on Build GRCh38
rs952861960 Chr.8: 28028951-28028951 on Build GRCh38
rs529660308 Chr.8: 28029205-28029205 on Build GRCh38
rs1045933283 Chr.8: 28029056-28029056 on Build GRCh38
rs546939680 Chr.8: 28029138-28029138 on Build GRCh38
rs763484760 Chr.8: 28029352-28029352 on Build GRCh38
rs747956681 Chr.8: 28029029-28029029 on Build GRCh38
rs984249214 Chr.8: 28029194-28029194 on Build GRCh38
rs935812803 Chr.8: 28029100-28029100 on Build GRCh38
rs375483912 Chr.8: 28029232-28029232 on Build GRCh38
rs977379299 Chr.8: 28029204-28029204 on Build GRCh38
rs911739961 Chr.8: 28028893-28028893 on Build GRCh38
rs576293533 Chr.8: 28028950-28028950 on Build GRCh38
rs922226302 Chr.8: 28029007-28029007 on Build GRCh38
rs571666276 Chr.8: 28029291-28029291 on Build GRCh38
rs1045984864 Chr.8: 28029280-28029280 on Build GRCh38
rs945843784 Chr.8: 28028895-28028895 on Build GRCh38
rs564046332 Chr.8: 28029151-28029151 on Build GRCh38
rs1046044851 Chr.8: 28029289-28029289 on Build GRCh38
rs574481213 Chr.8: 28029175-28029175 on Build GRCh38
rs977533862 Chr.8: 28028936-28028936 on Build GRCh38
rs994289947 Chr.8: 28029182-28029182 on Build GRCh38
rs117499371 Chr.8: 28028871-28028871 on Build GRCh38
rs778570883 Chr.8: 28029265-28029265 on Build GRCh38
rs752683112 Chr.8: 28029234-28029234 on Build GRCh38
rs1055718977 Chr.8: 28029015-28029015 on Build GRCh38
rs9644134 Chr.8: 28029183-28029183 on Build GRCh38
rs115562142 Chr.8: 28028945-28028945 on Build GRCh38
rs751358753 Chr.8: 28029226-28029226 on Build GRCh38
rs377099582 Chr.8: 28029333-28029333 on Build GRCh38
rs144775340 Chr.8: 28029206-28029206 on Build GRCh38
rs776811590 Chr.8: 28029310-28029310 on Build GRCh38
rs747619885 Chr.8: 28029282-28029282 on Build GRCh38
rs868153265 Chr.8: 28029321-28029327 on Build GRCh38
rs906506373 Chr.8: 28029160-28029160 on Build GRCh38
rs199698334 Chr.8: 28029327-28029327 on Build GRCh38
rs200971147 Chr.8: 28029334-28029334 on Build GRCh38
rs114256518 Chr.8: 28029213-28029213 on Build GRCh38
rs753352506 Chr.8: 28029353-28029353 on Build GRCh38
rs563236354 Chr.8: 28028873-28028873 on Build GRCh38
rs768604595 Chr.8: 28029303-28029303 on Build GRCh38
rs148595255 Chr.8: 28029248-28029248 on Build GRCh38
rs561693149 Chr.8: 28028892-28028892 on Build GRCh38
rs373935705 Chr.8: 28029337-28029337 on Build GRCh38
rs994546250 Chr.8: 28029159-28029159 on Build GRCh38
rs894753765 Chr.8: 28029035-28029035 on Build GRCh38
rs976446673 Chr.8: 28029001-28029001 on Build GRCh38
rs113558485 Chr.8: 28029221-28029221 on Build GRCh38
rs761829847 Chr.8: 28029316-28029316 on Build GRCh38
rs755572114 Chr.8: 28029237-28029237 on Build GRCh38
rs1004750377 Chr.8: 28029147-28029147 on Build GRCh38
rs756182890 Chr.8: 28029042-28029042 on Build GRCh38
rs775910905 Chr.8: 28029275-28029275 on Build GRCh38
rs952914471 Chr.8: 28029186-28029186 on Build GRCh38
rs1041835793 Chr.8: 28029046-28029046 on Build GRCh38
rs1024989670 Chr.8: 28029153-28029153 on Build GRCh38
rs770091256 Chr.8: 28029320-28029320 on Build GRCh38
rs533007680 Chr.8: 28029177-28029177 on Build GRCh38
rs772441783 Chr.8: 28029274-28029274 on Build GRCh38
rs138862929 Chr.8: 28028957-28028957 on Build GRCh38
rs898903982 Chr.8: 28029157-28029157 on Build GRCh38
rs773579924 Chr.8: 28029321-28029321 on Build GRCh38
rs527284260 Chr.8: 28029059-28029059 on Build GRCh38
rs759335784 Chr.8: 28029325-28029325 on Build GRCh38
rs911465434 Chr.8: 28028999-28028999 on Build GRCh38
rs984363724 Chr.8: 28028990-28028990 on Build GRCh38
rs776714468 Chr.8: 28029217-28029217 on Build GRCh38
rs777833545 Chr.8: 28029065-28029065 on Build GRCh38
rs373161383 Chr.8: 28029256-28029256 on Build GRCh38
rs115069806 Chr.8: 28029202-28029202 on Build GRCh38
rs950289210 Chr.8: 28029049-28029049 on Build GRCh38
rs560247922 Chr.8: 28029222-28029222 on Build GRCh38
rs866336686 Chr.8: 28029338-28029338 on Build GRCh38
rs746183956 Chr.8: 28029267-28029267 on Build GRCh38
rs936036867 Chr.8: 28029000-28029000 on Build GRCh38
rs142195858 Chr.8: 28029040-28029040 on Build GRCh38

View Full Product Details