Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28030232-28030740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUGGC
Entrez Gene ID
389643
Gene Name
nuclear GTPase, germinal center associated
Gene Aliases
C8orf80, HMFN0672, SLIP-GC
UniGene
Hs.370129
Gene Chromosome Location
Chr.8: 28021964-28084068 on Build GRCh38
Transcripts
NM_001010906.1 XM_011544523.2 XM_011544524.2 XM_011544525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1456443 c.2017G>T p.E673* Chr.8: 28030310-28030310 on Build GRCh38
86281 c.1955A>T p.K652M Chr.8: 28030372-28030372 on Build GRCh38
41105 c.1936G>T p.D646Y Chr.8: 28030391-28030391 on Build GRCh38
1456445 c.1936G>A p.D646N Chr.8: 28030391-28030391 on Build GRCh38
5897681 c.1927G>A p.G643S Chr.8: 28030400-28030400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866248621 Chr.8: 28030396-28030396 on Build GRCh38
rs914296745 Chr.8: 28030525-28030525 on Build GRCh38
rs367607568 Chr.8: 28030395-28030395 on Build GRCh38
rs573676758 Chr.8: 28030679-28030679 on Build GRCh38
rs371363580 Chr.8: 28030403-28030403 on Build GRCh38
rs756724417 Chr.8: 28030461-28030461 on Build GRCh38
rs878905829 Chr.8: 28030574-28030574 on Build GRCh38
rs1054633595 Chr.8: 28030431-28030431 on Build GRCh38
rs942701494 Chr.8: 28030648-28030648 on Build GRCh38
rs571533689 Chr.8: 28030609-28030609 on Build GRCh38
rs772750037 Chr.8: 28030418-28030418 on Build GRCh38
rs144982138 Chr.8: 28030457-28030457 on Build GRCh38
rs539114647 Chr.8: 28030688-28030688 on Build GRCh38
rs878983258 Chr.8: 28030486-28030486 on Build GRCh38
rs556963225 Chr.8: 28030260-28030260 on Build GRCh38
rs775593869 Chr.8: 28030577-28030577 on Build GRCh38
rs956123207 Chr.8: 28030637-28030637 on Build GRCh38
rs202103381 Chr.8: 28030314-28030314 on Build GRCh38
rs769978070 Chr.8: 28030348-28030348 on Build GRCh38
rs560036453 Chr.8: 28030298-28030298 on Build GRCh38
rs974664756 Chr.8: 28030385-28030385 on Build GRCh38
rs901455299 Chr.8: 28030667-28030667 on Build GRCh38
rs928900824 Chr.8: 28030707-28030707 on Build GRCh38
rs560923166 Chr.8: 28030408-28030408 on Build GRCh38
rs530177452 Chr.8: 28030452-28030452 on Build GRCh38
rs573468521 Chr.8: 28030299-28030299 on Build GRCh38
rs914768618 Chr.8: 28030659-28030659 on Build GRCh38
rs1047346399 Chr.8: 28030494-28030494 on Build GRCh38
rs765998334 Chr.8: 28030442-28030442 on Build GRCh38
rs767827058 Chr.8: 28030456-28030456 on Build GRCh38
rs531085859 Chr.8: 28030369-28030369 on Build GRCh38
rs763763612 Chr.8: 28030712-28030712 on Build GRCh38
rs758097023 Chr.8: 28030303-28030303 on Build GRCh38
rs753735823 Chr.8: 28030271-28030271 on Build GRCh38
rs753387168 Chr.8: 28030460-28030473 on Build GRCh38
rs990210864 Chr.8: 28030640-28030640 on Build GRCh38
rs374292249 Chr.8: 28030313-28030313 on Build GRCh38
rs1036866086 Chr.8: 28030386-28030386 on Build GRCh38
rs765851203 Chr.8: 28030303-28030303 on Build GRCh38
rs928559180 Chr.8: 28030352-28030352 on Build GRCh38
rs549953041 Chr.8: 28030685-28030685 on Build GRCh38
rs779602831 Chr.8: 28030306-28030306 on Build GRCh38
rs149117776 Chr.8: 28030686-28030686 on Build GRCh38
rs551257783 Chr.8: 28030559-28030559 on Build GRCh38
rs955595513 Chr.8: 28030604-28030604 on Build GRCh38
rs771242077 Chr.8: 28030365-28030365 on Build GRCh38
rs924689487 Chr.8: 28030402-28030402 on Build GRCh38
rs201021494 Chr.8: 28030426-28030426 on Build GRCh38
rs918430744 Chr.8: 28030595-28030595 on Build GRCh38
rs1031159753 Chr.8: 28030463-28030463 on Build GRCh38
rs751902053 Chr.8: 28030312-28030312 on Build GRCh38
rs755364340 Chr.8: 28030445-28030445 on Build GRCh38
rs79628515 Chr.8: 28030543-28030543 on Build GRCh38
rs536931160 Chr.8: 28030611-28030611 on Build GRCh38
rs979722062 Chr.8: 28030555-28030555 on Build GRCh38
rs572806986 Chr.8: 28030711-28030711 on Build GRCh38
rs7817068 Chr.8: 28030296-28030296 on Build GRCh38
rs958164567 Chr.8: 28030475-28030475 on Build GRCh38
rs751174152 Chr.8: 28030444-28030444 on Build GRCh38
rs573814693 Chr.8: 28030267-28030267 on Build GRCh38
rs778044873 Chr.8: 28030358-28030358 on Build GRCh38
rs566585285 Chr.8: 28030517-28030517 on Build GRCh38
rs553115850 Chr.8: 28030710-28030710 on Build GRCh38
rs1030238619 Chr.8: 28030598-28030598 on Build GRCh38
rs764704854 Chr.8: 28030413-28030413 on Build GRCh38
rs200096438 Chr.8: 28030404-28030404 on Build GRCh38
rs921289062 Chr.8: 28030390-28030390 on Build GRCh38
rs749202731 Chr.8: 28030691-28030691 on Build GRCh38
rs949004348 Chr.8: 28030458-28030458 on Build GRCh38
rs748667608 Chr.8: 28030324-28030324 on Build GRCh38
rs557026017 Chr.8: 28030664-28030664 on Build GRCh38
rs1002535487 Chr.8: 28030472-28030472 on Build GRCh38
rs374100228 Chr.8: 28030393-28030393 on Build GRCh38
rs964557008 Chr.8: 28030555-28030555 on Build GRCh38
rs200709366 Chr.8: 28030292-28030292 on Build GRCh38
rs889861056 Chr.8: 28030624-28030624 on Build GRCh38
rs1041048277 Chr.8: 28030649-28030649 on Build GRCh38
rs149939816 Chr.8: 28030359-28030359 on Build GRCh38

View Full Product Details