Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28030966-28031466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUGGC
Entrez Gene ID
389643
Gene Name
nuclear GTPase, germinal center associated
Gene Aliases
C8orf80, HMFN0672, SLIP-GC
UniGene
Hs.370129
Gene Chromosome Location
Chr.8: 28021964-28084068 on Build GRCh38
Transcripts
NM_001010906.1 XM_011544523.2 XM_011544524.2 XM_011544525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5119993 c.1891A>C p.N631H Chr.8: 28031260-28031260 on Build GRCh38
3647892 c.1834G>A p.E612K Chr.8: 28031317-28031317 on Build GRCh38
4668060 c.1851T>C p.I617I Chr.8: 28031300-28031300 on Build GRCh38
3647890 c.1854G>A p.G618G Chr.8: 28031297-28031297 on Build GRCh38
6197206 c.1773G>A p.T591T Chr.8: 28031378-28031378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs58252544 Chr.8: 28031327-28031327 on Build GRCh38
rs532464714 Chr.8: 28031048-28031048 on Build GRCh38
rs750435784 Chr.8: 28031064-28031064 on Build GRCh38
rs911348187 Chr.8: 28031016-28031016 on Build GRCh38
rs267601886 Chr.8: 28031255-28031255 on Build GRCh38
rs773772348 Chr.8: 28031380-28031380 on Build GRCh38
rs754385600 Chr.8: 28031421-28031421 on Build GRCh38
rs368713059 Chr.8: 28031278-28031278 on Build GRCh38
rs755498396 Chr.8: 28031259-28031259 on Build GRCh38
rs901174967 Chr.8: 28031049-28031049 on Build GRCh38
rs559150872 Chr.8: 28031239-28031239 on Build GRCh38
rs114856066 Chr.8: 28031081-28031081 on Build GRCh38
rs900250973 Chr.8: 28031140-28031140 on Build GRCh38
rs761056062 Chr.8: 28031413-28031413 on Build GRCh38
rs938964244 Chr.8: 28031078-28031078 on Build GRCh38
rs201027118 Chr.8: 28031270-28031270 on Build GRCh38
rs758588752 Chr.8: 28031352-28031352 on Build GRCh38
rs375019295 Chr.8: 28031102-28031102 on Build GRCh38
rs777668889 Chr.8: 28031184-28031184 on Build GRCh38
rs778459817 Chr.8: 28031194-28031194 on Build GRCh38
rs538331274 Chr.8: 28031177-28031177 on Build GRCh38
rs540247086 Chr.8: 28031440-28031440 on Build GRCh38
rs775299483 Chr.8: 28031408-28031408 on Build GRCh38
rs952603766 Chr.8: 28031187-28031187 on Build GRCh38
rs990263118 Chr.8: 28031006-28031006 on Build GRCh38
rs2305456 Chr.8: 28031109-28031109 on Build GRCh38
rs911064804 Chr.8: 28031259-28031259 on Build GRCh38
rs765871389 Chr.8: 28031276-28031276 on Build GRCh38
rs536258342 Chr.8: 28031138-28031138 on Build GRCh38
rs749336850 Chr.8: 28031203-28031203 on Build GRCh38
rs780286891 Chr.8: 28031360-28031360 on Build GRCh38
rs377585931 Chr.8: 28031273-28031273 on Build GRCh38
rs746082643 Chr.8: 28031344-28031344 on Build GRCh38
rs769581405 Chr.8: 28031373-28031373 on Build GRCh38
rs748047866 Chr.8: 28031269-28031269 on Build GRCh38
rs369886793 Chr.8: 28031249-28031249 on Build GRCh38
rs762381336 Chr.8: 28031274-28031274 on Build GRCh38
rs762277736 Chr.8: 28031422-28031422 on Build GRCh38
rs905337088 Chr.8: 28031111-28031111 on Build GRCh38
rs2305455 Chr.8: 28031112-28031112 on Build GRCh38
rs375520230 Chr.8: 28031238-28031238 on Build GRCh38
rs753067539 Chr.8: 28031289-28031289 on Build GRCh38
rs756573428 Chr.8: 28031315-28031315 on Build GRCh38
rs1037451003 Chr.8: 28031116-28031116 on Build GRCh38
rs779403391 Chr.8: 28031405-28031405 on Build GRCh38
rs138224837 Chr.8: 28031091-28031091 on Build GRCh38
rs756794412 Chr.8: 28031282-28031282 on Build GRCh38
rs1035824964 Chr.8: 28031164-28031164 on Build GRCh38
rs767789820 Chr.8: 28031030-28031030 on Build GRCh38
rs905389379 Chr.8: 28031445-28031445 on Build GRCh38
rs983936228 Chr.8: 28031012-28031012 on Build GRCh38
rs143160215 Chr.8: 28031033-28031033 on Build GRCh38
rs551588021 Chr.8: 28031076-28031076 on Build GRCh38
rs554844302 Chr.8: 28031324-28031324 on Build GRCh38
rs779133944 Chr.8: 28031245-28031245 on Build GRCh38
rs371097445 Chr.8: 28031425-28031425 on Build GRCh38
rs935303626 Chr.8: 28031072-28031072 on Build GRCh38
rs574764382 Chr.8: 28031375-28031375 on Build GRCh38
rs747879736 Chr.8: 28031372-28031372 on Build GRCh38
rs757356436 Chr.8: 28031326-28031326 on Build GRCh38
rs749240160 Chr.8: 28031378-28031378 on Build GRCh38
rs759113213 Chr.8: 28031385-28031385 on Build GRCh38
rs894045469 Chr.8: 28031077-28031077 on Build GRCh38
rs144985793 Chr.8: 28031176-28031176 on Build GRCh38
rs771578530 Chr.8: 28031395-28031395 on Build GRCh38
rs141259939 Chr.8: 28031174-28031174 on Build GRCh38
rs373284363 Chr.8: 28031342-28031342 on Build GRCh38
rs764623914 Chr.8: 28031415-28031415 on Build GRCh38
rs757299660 Chr.8: 28031220-28031220 on Build GRCh38
rs544358490 Chr.8: 28031321-28031321 on Build GRCh38
rs377427046 Chr.8: 28031379-28031379 on Build GRCh38
rs759158439 Chr.8: 28031281-28031281 on Build GRCh38

View Full Product Details