Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28096901-28097446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP3
Entrez Gene ID
55140
Gene Name
elongator acetyltransferase complex subunit 3
Gene Aliases
KAT9
UniGene
Hs.491336
Gene Chromosome Location
Chr.8: 28090231-28191153 on Build GRCh38
Transcripts
NM_001284220.1 NM_001284222.1 NM_001284224.1 NM_001284225.1 NM_001284226.1 NM_018091.5 XM_006716354.2 XM_017013604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3715912 c.85G>A p.A29T Chr.8: 28097284-28097284 on Build GRCh38
7296992 c.91G>A p.E31K Chr.8: 28097290-28097290 on Build GRCh38
3412965 c.48G>A p.M16I Chr.8: 28097247-28097247 on Build GRCh38
3982502 c.90C>T p.H30H Chr.8: 28097289-28097289 on Build GRCh38
7111291 c.78G>C p.L26L Chr.8: 28097277-28097277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182385725 Chr.8: 28097388-28097388 on Build GRCh38
rs764296715 Chr.8: 28097278-28097278 on Build GRCh38
rs747637581 Chr.8: 28097238-28097238 on Build GRCh38
rs1006848044 Chr.8: 28097090-28097090 on Build GRCh38
rs530503900 Chr.8: 28096938-28096938 on Build GRCh38
rs78313841 Chr.8: 28097108-28097108 on Build GRCh38
rs1048302401 Chr.8: 28097051-28097051 on Build GRCh38
rs989112735 Chr.8: 28097052-28097052 on Build GRCh38
rs560647549 Chr.8: 28097262-28097262 on Build GRCh38
rs781220298 Chr.8: 28097063-28097063 on Build GRCh38
rs780715744 Chr.8: 28097204-28097204 on Build GRCh38
rs781009387 Chr.8: 28097313-28097313 on Build GRCh38
rs770018555 Chr.8: 28097185-28097185 on Build GRCh38
rs748138953 Chr.8: 28097366-28097367 on Build GRCh38
rs772563558 Chr.8: 28097351-28097351 on Build GRCh38
rs754417860 Chr.8: 28097196-28097196 on Build GRCh38
rs115640995 Chr.8: 28097116-28097116 on Build GRCh38
rs751566242 Chr.8: 28097187-28097187 on Build GRCh38
rs777631204 Chr.8: 28097250-28097250 on Build GRCh38
rs780260717 Chr.8: 28097333-28097334 on Build GRCh38
rs1019687786 Chr.8: 28097106-28097106 on Build GRCh38
rs1036465792 Chr.8: 28097032-28097032 on Build GRCh38
rs575255302 Chr.8: 28097071-28097071 on Build GRCh38
rs368354209 Chr.8: 28097307-28097307 on Build GRCh38
rs200551475 Chr.8: 28097365-28097365 on Build GRCh38
rs372533677 Chr.8: 28097327-28097327 on Build GRCh38
rs753320384 Chr.8: 28097048-28097048 on Build GRCh38
rs113326698 Chr.8: 28097398-28097398 on Build GRCh38
rs1563055 Chr.8: 28096948-28096948 on Build GRCh38
rs771468946 Chr.8: 28097254-28097254 on Build GRCh38
rs930755604 Chr.8: 28097348-28097348 on Build GRCh38
rs762715756 Chr.8: 28097290-28097290 on Build GRCh38
rs889653724 Chr.8: 28097373-28097373 on Build GRCh38
rs779734365 Chr.8: 28097256-28097256 on Build GRCh38
rs748558306 Chr.8: 28097127-28097127 on Build GRCh38
rs568099904 Chr.8: 28097405-28097405 on Build GRCh38
rs748985249 Chr.8: 28097325-28097325 on Build GRCh38
rs770737109 Chr.8: 28097326-28097326 on Build GRCh38
rs772133351 Chr.8: 28097367-28097368 on Build GRCh38
rs982620212 Chr.8: 28097311-28097311 on Build GRCh38
rs754899290 Chr.8: 28097308-28097308 on Build GRCh38
rs10282902 Chr.8: 28097134-28097134 on Build GRCh38
rs756310209 Chr.8: 28097175-28097175 on Build GRCh38
rs769518295 Chr.8: 28097244-28097244 on Build GRCh38
rs758256682 Chr.8: 28097185-28097185 on Build GRCh38
rs766204278 Chr.8: 28097298-28097298 on Build GRCh38
rs760796319 Chr.8: 28097370-28097370 on Build GRCh38
rs567343859 Chr.8: 28097056-28097056 on Build GRCh38
rs368407461 Chr.8: 28097182-28097182 on Build GRCh38
rs558589069 Chr.8: 28097017-28097017 on Build GRCh38
rs777543022 Chr.8: 28097321-28097321 on Build GRCh38
rs749653454 Chr.8: 28097253-28097253 on Build GRCh38
rs1048251088 Chr.8: 28097355-28097355 on Build GRCh38
rs534818177 Chr.8: 28097280-28097280 on Build GRCh38
rs989603992 Chr.8: 28097135-28097135 on Build GRCh38
rs146779290 Chr.8: 28097289-28097289 on Build GRCh38
rs999660974 Chr.8: 28097112-28097112 on Build GRCh38
rs758719237 Chr.8: 28097093-28097093 on Build GRCh38
rs372777216 Chr.8: 28097186-28097186 on Build GRCh38
rs540910515 Chr.8: 28097139-28097139 on Build GRCh38
rs755739854 Chr.8: 28097319-28097319 on Build GRCh38
rs775580720 Chr.8: 28097277-28097277 on Build GRCh38
rs879079696 Chr.8: 28097147-28097147 on Build GRCh38
rs746231197 Chr.8: 28097329-28097329 on Build GRCh38
rs942595026 Chr.8: 28097380-28097380 on Build GRCh38
rs1046821361 Chr.8: 28096961-28096961 on Build GRCh38
rs779548212 Chr.8: 28097008-28097008 on Build GRCh38
rs747024523 Chr.8: 28097035-28097035 on Build GRCh38
rs772476943 Chr.8: 28097266-28097266 on Build GRCh38
rs752242709 Chr.8: 28097315-28097315 on Build GRCh38

View Full Product Details