Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28132098-28132498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP3
Entrez Gene ID
55140
Gene Name
elongator acetyltransferase complex subunit 3
Gene Aliases
KAT9
UniGene
Hs.491336
Gene Chromosome Location
Chr.8: 28090231-28191153 on Build GRCh38
Transcripts
NM_001284220.1 NM_001284222.1 NM_001284224.1 NM_001284225.1 NM_001284226.1 NM_018091.5 XM_006716354.2 XM_017013604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
454392 c.903C>T p.F301F Chr.8: 28132401-28132401 on Build GRCh38
5359033 c.860C>A p.P287H Chr.8: 28132358-28132358 on Build GRCh38
5590744 c.891C>T p.D297D Chr.8: 28132389-28132389 on Build GRCh38
5786245 c.850C>T p.H284Y Chr.8: 28132348-28132348 on Build GRCh38
3647909 c.795G>A p.K265K Chr.8: 28132293-28132293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550236608 Chr.8: 28132350-28132350 on Build GRCh38
rs569153927 Chr.8: 28132371-28132371 on Build GRCh38
rs985381492 Chr.8: 28132174-28132174 on Build GRCh38
rs945328548 Chr.8: 28132437-28132437 on Build GRCh38
rs866987228 Chr.8: 28132300-28132300 on Build GRCh38
rs763810644 Chr.8: 28132251-28132251 on Build GRCh38
rs80066224 Chr.8: 28132223-28132223 on Build GRCh38
rs764954376 Chr.8: 28132391-28132391 on Build GRCh38
rs757726055 Chr.8: 28132206-28132206 on Build GRCh38
rs781450721 Chr.8: 28132316-28132316 on Build GRCh38
rs1053943345 Chr.8: 28132309-28132309 on Build GRCh38
rs763648850 Chr.8: 28132353-28132353 on Build GRCh38
rs772135398 Chr.8: 28132345-28132345 on Build GRCh38
rs757811822 Chr.8: 28132426-28132426 on Build GRCh38
rs1055684926 Chr.8: 28132165-28132165 on Build GRCh38
rs372074866 Chr.8: 28132351-28132351 on Build GRCh38
rs368068287 Chr.8: 28132427-28132427 on Build GRCh38
rs912219800 Chr.8: 28132181-28132181 on Build GRCh38
rs752418673 Chr.8: 28132411-28132411 on Build GRCh38
rs376651415 Chr.8: 28132124-28132124 on Build GRCh38
rs746248780 Chr.8: 28132274-28132274 on Build GRCh38
rs779432524 Chr.8: 28132390-28132390 on Build GRCh38
rs148935021 Chr.8: 28132281-28132281 on Build GRCh38
rs547890457 Chr.8: 28132199-28132199 on Build GRCh38
rs779107100 Chr.8: 28132273-28132273 on Build GRCh38
rs878928401 Chr.8: 28132196-28132196 on Build GRCh38
rs751945671 Chr.8: 28132262-28132262 on Build GRCh38
rs753484781 Chr.8: 28132256-28132256 on Build GRCh38
rs368317768 Chr.8: 28132329-28132329 on Build GRCh38
rs993653398 Chr.8: 28132225-28132225 on Build GRCh38
rs773962755 Chr.8: 28132331-28132331 on Build GRCh38
rs759464706 Chr.8: 28132339-28132339 on Build GRCh38
rs780595629 Chr.8: 28132280-28132280 on Build GRCh38
rs373954614 Chr.8: 28132416-28132416 on Build GRCh38
rs867667800 Chr.8: 28132318-28132318 on Build GRCh38
rs1014893868 Chr.8: 28132191-28132191 on Build GRCh38
rs770082832 Chr.8: 28132330-28132330 on Build GRCh38
rs761306790 Chr.8: 28132372-28132372 on Build GRCh38
rs752245799 Chr.8: 28132228-28132228 on Build GRCh38
rs943707167 Chr.8: 28132204-28132204 on Build GRCh38
rs79959029 Chr.8: 28132129-28132129 on Build GRCh38
rs756959039 Chr.8: 28132262-28132262 on Build GRCh38
rs976175317 Chr.8: 28132384-28132384 on Build GRCh38
rs894485952 Chr.8: 28132187-28132187 on Build GRCh38
rs758844650 Chr.8: 28132279-28132279 on Build GRCh38
rs768762720 Chr.8: 28132284-28132284 on Build GRCh38
rs755754385 Chr.8: 28132245-28132245 on Build GRCh38

View Full Product Details