Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28717476-28717980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL3
Entrez Gene ID
2137
Gene Name
exostosin like glycosyltransferase 3
Gene Aliases
BOTV, EXTL1L, EXTR1, REGR, RPR
UniGene
Hs.491354
Gene Chromosome Location
Chr.8: 28701473-28753690 on Build GRCh38
Transcripts
NM_001440.3 NR_073468.1 NR_073469.1 XM_011544440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6213304 c.1848C>T p.H616H Chr.8: 28717907-28717907 on Build GRCh38
7255720 c.1758A>T p.S586S Chr.8: 28717817-28717817 on Build GRCh38
6180716 c.1570G>A p.D524N Chr.8: 28717629-28717629 on Build GRCh38
4654374 c.1610G>T p.R537L Chr.8: 28717669-28717669 on Build GRCh38
5351506 c.1853C>T p.P618L Chr.8: 28717912-28717912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200367738 Chr.8: 28717578-28717578 on Build GRCh38
rs779253231 Chr.8: 28717517-28717517 on Build GRCh38
rs912993523 Chr.8: 28717814-28717814 on Build GRCh38
rs530699484 Chr.8: 28717812-28717812 on Build GRCh38
rs753586923 Chr.8: 28717803-28717803 on Build GRCh38
rs371346996 Chr.8: 28717625-28717625 on Build GRCh38
rs145382567 Chr.8: 28717588-28717588 on Build GRCh38
rs765007284 Chr.8: 28717931-28717931 on Build GRCh38
rs758179582 Chr.8: 28717784-28717786 on Build GRCh38
rs905519299 Chr.8: 28717655-28717655 on Build GRCh38
rs775024686 Chr.8: 28717668-28717668 on Build GRCh38
rs770098386 Chr.8: 28717863-28717863 on Build GRCh38
rs763058199 Chr.8: 28717780-28717780 on Build GRCh38
rs994319491 Chr.8: 28717718-28717718 on Build GRCh38
rs759701104 Chr.8: 28717799-28717799 on Build GRCh38
rs764122764 Chr.8: 28717501-28717501 on Build GRCh38
rs1011249482 Chr.8: 28717640-28717640 on Build GRCh38
rs367805992 Chr.8: 28717559-28717559 on Build GRCh38
rs753651394 Chr.8: 28717735-28717735 on Build GRCh38
rs891789506 Chr.8: 28717920-28717920 on Build GRCh38
rs766423907 Chr.8: 28717953-28717953 on Build GRCh38
rs770287048 Chr.8: 28717775-28717775 on Build GRCh38
rs150064380 Chr.8: 28717562-28717562 on Build GRCh38
rs756512091 Chr.8: 28717737-28717737 on Build GRCh38
rs545077269 Chr.8: 28717580-28717580 on Build GRCh38
rs937010739 Chr.8: 28717923-28717923 on Build GRCh38
rs200923391 Chr.8: 28717520-28717520 on Build GRCh38
rs781097911 Chr.8: 28717525-28717525 on Build GRCh38
rs576797149 Chr.8: 28717632-28717632 on Build GRCh38
rs746530379 Chr.8: 28717835-28717835 on Build GRCh38
rs143705855 Chr.8: 28717696-28717696 on Build GRCh38
rs1018265838 Chr.8: 28717748-28717748 on Build GRCh38
rs752490010 Chr.8: 28717621-28717621 on Build GRCh38
rs752528930 Chr.8: 28717747-28717761 on Build GRCh38
rs200375528 Chr.8: 28717739-28717739 on Build GRCh38
rs762741027 Chr.8: 28717947-28717947 on Build GRCh38
rs771639784 Chr.8: 28717572-28717572 on Build GRCh38
rs781014999 Chr.8: 28717852-28717852 on Build GRCh38
rs35781576 Chr.8: 28717708-28717708 on Build GRCh38
rs769679440 Chr.8: 28717536-28717536 on Build GRCh38
rs761635451 Chr.8: 28717925-28717925 on Build GRCh38
rs907521015 Chr.8: 28717759-28717759 on Build GRCh38
rs765968684 Chr.8: 28717497-28717497 on Build GRCh38
rs755902308 Chr.8: 28717725-28717725 on Build GRCh38
rs759698175 Chr.8: 28717695-28717695 on Build GRCh38
rs777711529 Chr.8: 28717541-28717541 on Build GRCh38
rs891526676 Chr.8: 28717633-28717633 on Build GRCh38
rs750596299 Chr.8: 28717604-28717604 on Build GRCh38
rs771738641 Chr.8: 28717914-28717914 on Build GRCh38
rs996983305 Chr.8: 28717850-28717850 on Build GRCh38
rs749822351 Chr.8: 28717893-28717893 on Build GRCh38
rs141360609 Chr.8: 28717598-28717598 on Build GRCh38
rs201109508 Chr.8: 28717587-28717587 on Build GRCh38
rs748555675 Chr.8: 28717763-28717763 on Build GRCh38
rs376869552 Chr.8: 28717831-28717831 on Build GRCh38
rs754585925 Chr.8: 28717958-28717958 on Build GRCh38
rs751064521 Chr.8: 28717957-28717957 on Build GRCh38
rs769181445 Chr.8: 28717669-28717669 on Build GRCh38
rs201439713 Chr.8: 28717828-28717828 on Build GRCh38
rs138950489 Chr.8: 28717647-28717647 on Build GRCh38
rs1050016206 Chr.8: 28717881-28717881 on Build GRCh38
rs923182549 Chr.8: 28717798-28717798 on Build GRCh38
rs759791895 Chr.8: 28717502-28717502 on Build GRCh38
rs775735115 Chr.8: 28717919-28717919 on Build GRCh38
rs367666905 Chr.8: 28717624-28717624 on Build GRCh38
rs780170557 Chr.8: 28717614-28717614 on Build GRCh38
rs376394802 Chr.8: 28717719-28717719 on Build GRCh38
rs956170870 Chr.8: 28717679-28717679 on Build GRCh38
rs747900653 Chr.8: 28717532-28717532 on Build GRCh38
rs891358223 Chr.8: 28717886-28717886 on Build GRCh38
rs777077240 Chr.8: 28717758-28717758 on Build GRCh38
rs200563165 Chr.8: 28717896-28717896 on Build GRCh38
rs761951214 Chr.8: 28717594-28717594 on Build GRCh38
rs147333542 Chr.8: 28717829-28717829 on Build GRCh38
rs140625555 Chr.8: 28717685-28717685 on Build GRCh38
rs201787949 Chr.8: 28717706-28717706 on Build GRCh38
rs771711597 Chr.8: 28717661-28717661 on Build GRCh38
rs141817681 Chr.8: 28717756-28717756 on Build GRCh38
rs943257939 Chr.8: 28717577-28717577 on Build GRCh38
rs898650432 Chr.8: 28717535-28717535 on Build GRCh38
rs757432629 Chr.8: 28717512-28717512 on Build GRCh38
rs754610555 Chr.8: 28717840-28717840 on Build GRCh38
rs747753901 Chr.8: 28717909-28717909 on Build GRCh38
rs746724112 Chr.8: 28717743-28717743 on Build GRCh38
rs767996682 Chr.8: 28717503-28717503 on Build GRCh38
rs369743761 Chr.8: 28717726-28717726 on Build GRCh38
rs750476711 Chr.8: 28717705-28717705 on Build GRCh38
rs143652582 Chr.8: 28717934-28717934 on Build GRCh38
rs766439584 Chr.8: 28717782-28717782 on Build GRCh38
rs771484952 Chr.8: 28717894-28717894 on Build GRCh38
rs750365532 Chr.8: 28717811-28717811 on Build GRCh38
rs759295414 Chr.8: 28717569-28717569 on Build GRCh38
rs773872905 Chr.8: 28717563-28717563 on Build GRCh38
rs201980808 Chr.8: 28717509-28717509 on Build GRCh38
rs779725016 Chr.8: 28717740-28717740 on Build GRCh38
rs1042826033 Chr.8: 28717842-28717842 on Build GRCh38
rs759647888 Chr.8: 28717901-28717901 on Build GRCh38
rs758580194 Chr.8: 28717613-28717613 on Build GRCh38
rs774545299 Chr.8: 28717788-28717788 on Build GRCh38
rs928526657 Chr.8: 28717728-28717728 on Build GRCh38
rs145090231 Chr.8: 28717641-28717641 on Build GRCh38
rs200494893 Chr.8: 28717738-28717738 on Build GRCh38
rs563208430 Chr.8: 28717656-28717656 on Build GRCh38
rs754724657 Chr.8: 28717524-28717524 on Build GRCh38
rs756582203 Chr.8: 28717506-28717506 on Build GRCh38
rs773766888 Chr.8: 28717864-28717864 on Build GRCh38
rs773642035 Chr.8: 28717779-28717779 on Build GRCh38
rs372319537 Chr.8: 28717804-28717804 on Build GRCh38
rs112724129 Chr.8: 28717646-28717646 on Build GRCh38
rs576766032 Chr.8: 28717686-28717686 on Build GRCh38
rs748502441 Chr.8: 28717862-28717862 on Build GRCh38

View Full Product Details