Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 29335442-29335952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP4
Entrez Gene ID
1846
Gene Name
dual specificity phosphatase 4
Gene Aliases
HVH2, MKP-2, MKP2, TYP
UniGene
Hs.417962
Gene Chromosome Location
Chr.8: 29333062-29350750 on Build GRCh38
Transcripts
NM_001394.6 NM_057158.3 XM_011544428.2

SNP Detail

SNP ID Chromosome Location
rs530905358 Chr.8: 29335587-29335587 on Build GRCh38
rs750269875 Chr.8: 29335683-29335683 on Build GRCh38
rs142857847 Chr.8: 29335832-29335832 on Build GRCh38
rs371352573 Chr.8: 29335532-29335532 on Build GRCh38
rs1006040249 Chr.8: 29335866-29335866 on Build GRCh38
rs755962094 Chr.8: 29335726-29335726 on Build GRCh38
rs976054794 Chr.8: 29335576-29335576 on Build GRCh38
rs923612998 Chr.8: 29335652-29335652 on Build GRCh38
rs921944716 Chr.8: 29335815-29335815 on Build GRCh38
rs1014920988 Chr.8: 29335728-29335728 on Build GRCh38
rs187828370 Chr.8: 29335468-29335468 on Build GRCh38
rs1044937339 Chr.8: 29335789-29335789 on Build GRCh38
rs28575768 Chr.8: 29335545-29335545 on Build GRCh38
rs999824543 Chr.8: 29335574-29335574 on Build GRCh38
rs151091682 Chr.8: 29335881-29335881 on Build GRCh38
rs187715778 Chr.8: 29335824-29335824 on Build GRCh38
rs748086063 Chr.8: 29335654-29335654 on Build GRCh38
rs963446899 Chr.8: 29335764-29335764 on Build GRCh38
rs934260612 Chr.8: 29335597-29335597 on Build GRCh38
rs36006276 Chr.8: 29335782-29335783 on Build GRCh38
rs894162230 Chr.8: 29335643-29335643 on Build GRCh38
rs192896325 Chr.8: 29335852-29335852 on Build GRCh38
rs540431845 Chr.8: 29335487-29335487 on Build GRCh38
rs761470586 Chr.8: 29335792-29335792 on Build GRCh38
rs761727746 Chr.8: 29335524-29335524 on Build GRCh38
rs912165620 Chr.8: 29335851-29335851 on Build GRCh38
rs539349019 Chr.8: 29335889-29335889 on Build GRCh38
rs112604209 Chr.8: 29335504-29335504 on Build GRCh38
rs75976495 Chr.8: 29335582-29335582 on Build GRCh38
rs542956615 Chr.8: 29335722-29335722 on Build GRCh38
rs951408208 Chr.8: 29335540-29335540 on Build GRCh38
rs1014153628 Chr.8: 29335670-29335670 on Build GRCh38
rs904281974 Chr.8: 29335554-29335554 on Build GRCh38
rs182743701 Chr.8: 29335823-29335823 on Build GRCh38
rs890612374 Chr.8: 29335530-29335530 on Build GRCh38
rs904250653 Chr.8: 29335923-29335923 on Build GRCh38
rs951451265 Chr.8: 29335849-29335849 on Build GRCh38
rs1012569952 Chr.8: 29335478-29335478 on Build GRCh38
rs935073563 Chr.8: 29335703-29335703 on Build GRCh38
rs1024681237 Chr.8: 29335682-29335682 on Build GRCh38
rs906419919 Chr.8: 29335806-29335806 on Build GRCh38
rs951237816 Chr.8: 29335712-29335712 on Build GRCh38
rs975581108 Chr.8: 29335909-29335909 on Build GRCh38
rs766238007 Chr.8: 29335787-29335787 on Build GRCh38
rs1041480706 Chr.8: 29335553-29335553 on Build GRCh38
rs964275352 Chr.8: 29335569-29335569 on Build GRCh38
rs192223343 Chr.8: 29335535-29335535 on Build GRCh38
rs766372308 Chr.8: 29335530-29335531 on Build GRCh38
rs964540091 Chr.8: 29335905-29335905 on Build GRCh38

View Full Product Details