Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30179328-30179840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN6
Entrez Gene ID
10671
Gene Name
dynactin subunit 6
Gene Aliases
WS-3, WS3, p27
UniGene
Hs.158427
Gene Chromosome Location
Chr.8: 30156297-30183639 on Build GRCh38
Transcripts
NM_006571.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203174 c.314A>C p.N105T Chr.8: 30179438-30179438 on Build GRCh38
5791750 c.300G>T p.K100N Chr.8: 30179424-30179424 on Build GRCh38
1623911 c.314A>G p.N105S Chr.8: 30179438-30179438 on Build GRCh38
5955179 c.318C>T p.V106V Chr.8: 30179442-30179442 on Build GRCh38
5358782 c.331+2T>C p.? Chr.8: 30179457-30179457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780375048 Chr.8: 30179773-30179773 on Build GRCh38
rs768942114 Chr.8: 30179503-30179503 on Build GRCh38
rs373600254 Chr.8: 30179362-30179362 on Build GRCh38
rs117333940 Chr.8: 30179512-30179512 on Build GRCh38
rs558001181 Chr.8: 30179572-30179572 on Build GRCh38
rs183063028 Chr.8: 30179580-30179580 on Build GRCh38
rs112774219 Chr.8: 30179573-30179573 on Build GRCh38
rs772163498 Chr.8: 30179390-30179390 on Build GRCh38
rs1031123270 Chr.8: 30179590-30179590 on Build GRCh38
rs78669402 Chr.8: 30179413-30179413 on Build GRCh38
rs142703056 Chr.8: 30179780-30179781 on Build GRCh38
rs987718962 Chr.8: 30179769-30179769 on Build GRCh38
rs370710105 Chr.8: 30179389-30179389 on Build GRCh38
rs531521186 Chr.8: 30179781-30179781 on Build GRCh38
rs982623988 Chr.8: 30179650-30179650 on Build GRCh38
rs138392089 Chr.8: 30179757-30179757 on Build GRCh38
rs946206702 Chr.8: 30179781-30179781 on Build GRCh38
rs762997333 Chr.8: 30179415-30179415 on Build GRCh38
rs756517627 Chr.8: 30179360-30179360 on Build GRCh38
rs746366976 Chr.8: 30179482-30179482 on Build GRCh38
rs145932152 Chr.8: 30179629-30179629 on Build GRCh38
rs543568823 Chr.8: 30179583-30179583 on Build GRCh38
rs374010509 Chr.8: 30179418-30179418 on Build GRCh38
rs778111151 Chr.8: 30179436-30179436 on Build GRCh38
rs191556638 Chr.8: 30179396-30179396 on Build GRCh38
rs747239552 Chr.8: 30179496-30179496 on Build GRCh38
rs765155808 Chr.8: 30179397-30179397 on Build GRCh38
rs756444797 Chr.8: 30179672-30179672 on Build GRCh38
rs946237864 Chr.8: 30179365-30179365 on Build GRCh38
rs1041755795 Chr.8: 30179380-30179380 on Build GRCh38
rs899386422 Chr.8: 30179543-30179543 on Build GRCh38
rs527460618 Chr.8: 30179562-30179562 on Build GRCh38
rs755720070 Chr.8: 30179427-30179427 on Build GRCh38
rs920908047 Chr.8: 30179751-30179751 on Build GRCh38
rs1016678190 Chr.8: 30179664-30179664 on Build GRCh38
rs369018320 Chr.8: 30179382-30179382 on Build GRCh38
rs752311748 Chr.8: 30179426-30179426 on Build GRCh38
rs570690354 Chr.8: 30179531-30179531 on Build GRCh38
rs750509782 Chr.8: 30179671-30179671 on Build GRCh38
rs779332192 Chr.8: 30179480-30179480 on Build GRCh38
rs138634227 Chr.8: 30179425-30179425 on Build GRCh38
rs772069009 Chr.8: 30179491-30179491 on Build GRCh38
rs975479309 Chr.8: 30179721-30179721 on Build GRCh38
rs754139130 Chr.8: 30179454-30179454 on Build GRCh38
rs757617028 Chr.8: 30179471-30179471 on Build GRCh38
rs781481164 Chr.8: 30179631-30179631 on Build GRCh38
rs1014137267 Chr.8: 30179530-30179530 on Build GRCh38
rs142488069 Chr.8: 30179374-30179374 on Build GRCh38
rs894375012 Chr.8: 30179508-30179508 on Build GRCh38
rs200264812 Chr.8: 30179420-30179420 on Build GRCh38
rs904644766 Chr.8: 30179391-30179391 on Build GRCh38
rs370211567 Chr.8: 30179493-30179493 on Build GRCh38
rs751508980 Chr.8: 30179419-30179419 on Build GRCh38
rs988680279 Chr.8: 30179645-30179645 on Build GRCh38
rs1024643738 Chr.8: 30179618-30179618 on Build GRCh38

View Full Product Details