Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30182636-30183160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN6
Entrez Gene ID
10671
Gene Name
dynactin subunit 6
Gene Aliases
WS-3, WS3, p27
UniGene
Hs.158427
Gene Chromosome Location
Chr.8: 30156297-30183639 on Build GRCh38
Transcripts
NM_006571.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5677958 c.499T>G p.L167V Chr.8: 30183099-30183099 on Build GRCh38
1099087 c.534G>T p.K178N Chr.8: 30183134-30183134 on Build GRCh38
750703 c.529C>T p.L177L Chr.8: 30183129-30183129 on Build GRCh38
3899530 c.490C>A p.L164M Chr.8: 30183090-30183090 on Build GRCh38
3942639 c.498C>G p.F166L Chr.8: 30183098-30183098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893807597 Chr.8: 30182925-30182925 on Build GRCh38
rs191140272 Chr.8: 30182870-30182870 on Build GRCh38
rs56302321 Chr.8: 30183018-30183018 on Build GRCh38
rs943211683 Chr.8: 30182841-30182841 on Build GRCh38
rs550907496 Chr.8: 30182902-30182902 on Build GRCh38
rs951803258 Chr.8: 30183007-30183007 on Build GRCh38
rs756993241 Chr.8: 30182877-30182877 on Build GRCh38
rs897373234 Chr.8: 30183034-30183034 on Build GRCh38
rs529884392 Chr.8: 30182693-30182693 on Build GRCh38
rs924952213 Chr.8: 30182910-30182910 on Build GRCh38
rs779787423 Chr.8: 30183128-30183128 on Build GRCh38
rs889920485 Chr.8: 30182831-30182831 on Build GRCh38
rs1013775532 Chr.8: 30182705-30182705 on Build GRCh38
rs185174634 Chr.8: 30182971-30182971 on Build GRCh38
rs545120032 Chr.8: 30182851-30182851 on Build GRCh38
rs180801074 Chr.8: 30182914-30182914 on Build GRCh38
rs961829214 Chr.8: 30182995-30182995 on Build GRCh38
rs1025210575 Chr.8: 30182782-30182782 on Build GRCh38
rs1010784802 Chr.8: 30182961-30182961 on Build GRCh38
rs961353244 Chr.8: 30182801-30182801 on Build GRCh38
rs190864998 Chr.8: 30183001-30183001 on Build GRCh38
rs895324783 Chr.8: 30182697-30182697 on Build GRCh38
rs952793414 Chr.8: 30182821-30182821 on Build GRCh38
rs936412678 Chr.8: 30182956-30182956 on Build GRCh38
rs1034749824 Chr.8: 30182982-30182982 on Build GRCh38
rs942865801 Chr.8: 30183100-30183100 on Build GRCh38
rs756290370 Chr.8: 30182745-30182745 on Build GRCh38
rs781116443 Chr.8: 30182888-30182888 on Build GRCh38
rs749453554 Chr.8: 30183083-30183083 on Build GRCh38
rs145462992 Chr.8: 30182816-30182816 on Build GRCh38
rs749405078 Chr.8: 30182828-30182828 on Build GRCh38
rs931613303 Chr.8: 30182815-30182815 on Build GRCh38
rs752133349 Chr.8: 30183037-30183037 on Build GRCh38
rs778120146 Chr.8: 30183071-30183071 on Build GRCh38
rs565007861 Chr.8: 30182860-30182860 on Build GRCh38
rs910367058 Chr.8: 30182836-30182836 on Build GRCh38
rs568220835 Chr.8: 30182683-30182683 on Build GRCh38
rs767707593 Chr.8: 30183049-30183049 on Build GRCh38
rs529214996 Chr.8: 30182793-30182793 on Build GRCh38
rs567089003 Chr.8: 30183005-30183005 on Build GRCh38
rs758239607 Chr.8: 30183090-30183090 on Build GRCh38
rs745875674 Chr.8: 30182998-30182998 on Build GRCh38
rs576606767 Chr.8: 30182850-30182850 on Build GRCh38
rs969219305 Chr.8: 30182963-30182963 on Build GRCh38
rs918837539 Chr.8: 30182734-30182734 on Build GRCh38
rs1042039952 Chr.8: 30182908-30182908 on Build GRCh38
rs763414787 Chr.8: 30183025-30183025 on Build GRCh38
rs766630230 Chr.8: 30183032-30183032 on Build GRCh38
rs757801441 Chr.8: 30182962-30182962 on Build GRCh38
rs756130915 Chr.8: 30183054-30183054 on Build GRCh38
rs755500284 Chr.8: 30183047-30183047 on Build GRCh38

View Full Product Details