Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 37736487-37736987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERLIN2
Entrez Gene ID
11160
Gene Name
ER lipid raft associated 2
Gene Aliases
C8orf2, Erlin-2, NET32, SPFH2, SPG18
UniGene
Hs.705490
Gene Chromosome Location
Chr.8: 37736579-37757801 on Build GRCh38
Transcripts
NM_001003790.3 NM_001003791.2 NM_007175.6 XM_005273392.2 XM_006716280.2 NR_125821.1

Gene Symbol
LOC102723701
Entrez Gene ID
102723701
Gene Name
uncharacterized LOC102723701
Gene Aliases
-
UniGene
Hs.588388
Gene Chromosome Location
Chr.8: 37734761-37737426 on Build GRCh38
Transcripts
NM_001003790.3 NM_001003791.2 NM_007175.6 XM_005273392.2 XM_006716280.2 NR_125821.1

SNP Detail

SNP ID Chromosome Location
rs928557758 Chr.8: 37736723-37736723 on Build GRCh38
rs550515267 Chr.8: 37736617-37736617 on Build GRCh38
rs35071288 Chr.8: 37736895-37736896 on Build GRCh38
rs550446832 Chr.8: 37736925-37736925 on Build GRCh38
rs763411561 Chr.8: 37736505-37736505 on Build GRCh38
rs958811109 Chr.8: 37736740-37736740 on Build GRCh38
rs547852240 Chr.8: 37736817-37736817 on Build GRCh38
rs975466703 Chr.8: 37736611-37736611 on Build GRCh38
rs566059959 Chr.8: 37736832-37736832 on Build GRCh38
rs1040600366 Chr.8: 37736672-37736672 on Build GRCh38
rs763248113 Chr.8: 37736519-37736536 on Build GRCh38
rs779206026 Chr.8: 37736528-37736528 on Build GRCh38
rs375491607 Chr.8: 37736882-37736882 on Build GRCh38
rs1044817024 Chr.8: 37736748-37736748 on Build GRCh38
rs899826854 Chr.8: 37736820-37736820 on Build GRCh38
rs766002950 Chr.8: 37736521-37736521 on Build GRCh38
rs2407540 Chr.8: 37736529-37736529 on Build GRCh38
rs1019533836 Chr.8: 37736561-37736561 on Build GRCh38
rs372875705 Chr.8: 37736552-37736552 on Build GRCh38
rs907829844 Chr.8: 37736593-37736593 on Build GRCh38
rs377607740 Chr.8: 37736624-37736624 on Build GRCh38
rs1031226877 Chr.8: 37736574-37736574 on Build GRCh38
rs376401158 Chr.8: 37736616-37736616 on Build GRCh38
rs751918955 Chr.8: 37736768-37736769 on Build GRCh38
rs1008888915 Chr.8: 37736815-37736815 on Build GRCh38
rs146112468 Chr.8: 37736834-37736834 on Build GRCh38
rs1045289892 Chr.8: 37736541-37736541 on Build GRCh38
rs866478706 Chr.8: 37736599-37736599 on Build GRCh38
rs1025913795 Chr.8: 37736549-37736549 on Build GRCh38
rs1028807597 Chr.8: 37736623-37736623 on Build GRCh38
rs996781932 Chr.8: 37736955-37736955 on Build GRCh38
rs907759783 Chr.8: 37736683-37736683 on Build GRCh38
rs980537514 Chr.8: 37736919-37736919 on Build GRCh38
rs984600567 Chr.8: 37736863-37736863 on Build GRCh38
rs974911078 Chr.8: 37736569-37736569 on Build GRCh38
rs1041048617 Chr.8: 37736928-37736928 on Build GRCh38
rs369977791 Chr.8: 37736935-37736935 on Build GRCh38
rs948207849 Chr.8: 37736506-37736506 on Build GRCh38
rs996915570 Chr.8: 37736827-37736827 on Build GRCh38
rs992073117 Chr.8: 37736885-37736885 on Build GRCh38
rs902089589 Chr.8: 37736947-37736947 on Build GRCh38
rs12679022 Chr.8: 37736685-37736685 on Build GRCh38
rs951989990 Chr.8: 37736842-37736842 on Build GRCh38
rs902037555 Chr.8: 37736674-37736674 on Build GRCh38
rs569668464 Chr.8: 37736566-37736566 on Build GRCh38
rs924987010 Chr.8: 37736929-37736929 on Build GRCh38
rs999796975 Chr.8: 37736662-37736662 on Build GRCh38
rs1019665380 Chr.8: 37736595-37736595 on Build GRCh38
rs892686087 Chr.8: 37736804-37736804 on Build GRCh38
rs935332425 Chr.8: 37736855-37736855 on Build GRCh38
rs937162799 Chr.8: 37736608-37736608 on Build GRCh38
rs372674481 Chr.8: 37736631-37736631 on Build GRCh38
rs1005890477 Chr.8: 37736696-37736696 on Build GRCh38
rs981781056 Chr.8: 37736715-37736715 on Build GRCh38
rs758980426 Chr.8: 37736554-37736554 on Build GRCh38
rs991801910 Chr.8: 37736698-37736698 on Build GRCh38
rs1055649292 Chr.8: 37736609-37736609 on Build GRCh38
rs902476347 Chr.8: 37736631-37736631 on Build GRCh38
rs947822691 Chr.8: 37736714-37736714 on Build GRCh38
rs1010899619 Chr.8: 37736519-37736519 on Build GRCh38
rs532559942 Chr.8: 37736799-37736799 on Build GRCh38
rs555029237 Chr.8: 37736934-37736934 on Build GRCh38
rs796367282 Chr.8: 37736905-37736906 on Build GRCh38
rs112956462 Chr.8: 37736607-37736607 on Build GRCh38
rs904037021 Chr.8: 37736752-37736752 on Build GRCh38
rs916652544 Chr.8: 37736613-37736614 on Build GRCh38
rs1008191332 Chr.8: 37736594-37736594 on Build GRCh38
rs985126286 Chr.8: 37736643-37736643 on Build GRCh38
rs1028727910 Chr.8: 37736570-37736570 on Build GRCh38
rs951788142 Chr.8: 37736690-37736692 on Build GRCh38
rs895152583 Chr.8: 37736509-37736509 on Build GRCh38
rs936832718 Chr.8: 37736790-37736790 on Build GRCh38
rs529801446 Chr.8: 37736551-37736551 on Build GRCh38
rs1015263481 Chr.8: 37736866-37736866 on Build GRCh38
rs892481548 Chr.8: 37736585-37736585 on Build GRCh38
rs984416159 Chr.8: 37736591-37736591 on Build GRCh38
rs1053121024 Chr.8: 37736792-37736792 on Build GRCh38
rs969016615 Chr.8: 37736918-37736918 on Build GRCh38
rs916701124 Chr.8: 37736795-37736795 on Build GRCh38
rs951867889 Chr.8: 37736626-37736626 on Build GRCh38
rs917611077 Chr.8: 37736907-37736907 on Build GRCh38
rs917580078 Chr.8: 37736702-37736702 on Build GRCh38

View Full Product Details