Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38210440-38210969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG4
Entrez Gene ID
9530
Gene Name
BCL2 associated athanogene 4
Gene Aliases
BAG-4, SODD
UniGene
Hs.194726
Gene Chromosome Location
Chr.8: 38176588-38213301 on Build GRCh38
Transcripts
NM_001204878.1 NM_004874.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5559595 c.1345C>G p.L449V Chr.8: 38210464-38210464 on Build GRCh38
217430 c.1365_1367delAGG p.G456delG Chr.8: 38210484-38210486 on Build GRCh38
5063181 c.1340C>T p.A447V Chr.8: 38210459-38210459 on Build GRCh38
1456885 c.1357_1358insA p.G456fs*>3 Chr.8: 38210476-38210477 on Build GRCh38
1456886 c.1358delA p.G456fs*>2 Chr.8: 38210477-38210477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962471207 Chr.8: 38210863-38210864 on Build GRCh38
rs1029470127 Chr.8: 38210804-38210804 on Build GRCh38
rs774440063 Chr.8: 38210494-38210494 on Build GRCh38
rs910867234 Chr.8: 38210538-38210538 on Build GRCh38
rs757324408 Chr.8: 38210520-38210520 on Build GRCh38
rs187571848 Chr.8: 38210500-38210500 on Build GRCh38
rs572774512 Chr.8: 38210530-38210530 on Build GRCh38
rs1043782 Chr.8: 38210806-38210806 on Build GRCh38
rs752844806 Chr.8: 38210564-38210564 on Build GRCh38
rs193259612 Chr.8: 38210688-38210688 on Build GRCh38
rs377003478 Chr.8: 38210582-38210582 on Build GRCh38
rs565399420 Chr.8: 38210587-38210587 on Build GRCh38
rs765067904 Chr.8: 38210529-38210529 on Build GRCh38
rs3313 Chr.8: 38210902-38210902 on Build GRCh38
rs555103516 Chr.8: 38210666-38210666 on Build GRCh38
rs910972600 Chr.8: 38210833-38210833 on Build GRCh38
rs80041165 Chr.8: 38210606-38210606 on Build GRCh38
rs985189379 Chr.8: 38210830-38210830 on Build GRCh38
rs758597761 Chr.8: 38210667-38210667 on Build GRCh38
rs763091825 Chr.8: 38210486-38210486 on Build GRCh38
rs758029217 Chr.8: 38210544-38210544 on Build GRCh38
rs541002014 Chr.8: 38210905-38210905 on Build GRCh38
rs532501974 Chr.8: 38210807-38210807 on Build GRCh38
rs764235854 Chr.8: 38210744-38210744 on Build GRCh38
rs895999960 Chr.8: 38210687-38210687 on Build GRCh38
rs543869203 Chr.8: 38210759-38210759 on Build GRCh38
rs553034109 Chr.8: 38210513-38210513 on Build GRCh38
rs1033044511 Chr.8: 38210753-38210753 on Build GRCh38
rs534585314 Chr.8: 38210508-38210508 on Build GRCh38
rs1037886147 Chr.8: 38210638-38210638 on Build GRCh38
rs1010187305 Chr.8: 38210802-38210802 on Build GRCh38
rs905823870 Chr.8: 38210717-38210717 on Build GRCh38
rs111385150 Chr.8: 38210585-38210585 on Build GRCh38
rs918273493 Chr.8: 38210904-38210904 on Build GRCh38
rs766536483 Chr.8: 38210488-38210488 on Build GRCh38
rs1044891697 Chr.8: 38210692-38210692 on Build GRCh38
rs1000204658 Chr.8: 38210743-38210743 on Build GRCh38
rs941035021 Chr.8: 38210615-38210615 on Build GRCh38
rs1047870247 Chr.8: 38210674-38210674 on Build GRCh38
rs758890670 Chr.8: 38210580-38210580 on Build GRCh38
rs929597728 Chr.8: 38210671-38210671 on Build GRCh38
rs780857982 Chr.8: 38210897-38210897 on Build GRCh38
rs929545276 Chr.8: 38210925-38210925 on Build GRCh38
rs899419132 Chr.8: 38210652-38210652 on Build GRCh38
rs766558104 Chr.8: 38210509-38210510 on Build GRCh38
rs751610425 Chr.8: 38210757-38210757 on Build GRCh38
rs973709672 Chr.8: 38210881-38210881 on Build GRCh38

View Full Product Details