Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72160036-72160537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ44635
Entrez Gene ID
392490
Gene Name
TPT1-like protein
Gene Aliases
-
UniGene
Hs.660859
Gene Chromosome Location
Chr.X: 72144184-72161750 on Build GRCh38
Transcripts
NM_207422.2

SNP Detail

SNP ID Chromosome Location
rs879086310 Chr.X: 72160081-72160081 on Build GRCh38
rs1055768765 Chr.X: 72160339-72160339 on Build GRCh38
rs772941211 Chr.X: 72160181-72160181 on Build GRCh38
rs771518122 Chr.X: 72160094-72160094 on Build GRCh38
rs766239159 Chr.X: 72160194-72160194 on Build GRCh38
rs777947699 Chr.X: 72160157-72160157 on Build GRCh38
rs747731775 Chr.X: 72160073-72160073 on Build GRCh38
rs7881332 Chr.X: 72160302-72160302 on Build GRCh38
rs377560939 Chr.X: 72160089-72160089 on Build GRCh38
rs751904169 Chr.X: 72160252-72160252 on Build GRCh38
rs201184731 Chr.X: 72160231-72160231 on Build GRCh38
rs879191642 Chr.X: 72160195-72160195 on Build GRCh38
rs761636700 Chr.X: 72160166-72160166 on Build GRCh38
rs747820208 Chr.X: 72160284-72160284 on Build GRCh38
rs778468064 Chr.X: 72160076-72160076 on Build GRCh38
rs11552476 Chr.X: 72160373-72160373 on Build GRCh38
rs753968072 Chr.X: 72160340-72160340 on Build GRCh38
rs145418226 Chr.X: 72160153-72160153 on Build GRCh38
rs879013865 Chr.X: 72160090-72160090 on Build GRCh38
rs760847684 Chr.X: 72160406-72160406 on Build GRCh38
rs1030535064 Chr.X: 72160082-72160082 on Build GRCh38
rs879244715 Chr.X: 72160116-72160116 on Build GRCh38
rs373347597 Chr.X: 72160277-72160277 on Build GRCh38
rs779431152 Chr.X: 72160121-72160121 on Build GRCh38
rs758657075 Chr.X: 72160218-72160218 on Build GRCh38
rs748794247 Chr.X: 72160276-72160276 on Build GRCh38
rs778244589 Chr.X: 72160280-72160280 on Build GRCh38
rs775553252 Chr.X: 72160315-72160315 on Build GRCh38
rs772998268 Chr.X: 72160298-72160298 on Build GRCh38
rs942064728 Chr.X: 72160374-72160374 on Build GRCh38
rs748383695 Chr.X: 72160402-72160402 on Build GRCh38
rs985775164 Chr.X: 72160126-72160126 on Build GRCh38
rs746097898 Chr.X: 72160118-72160138 on Build GRCh38
rs779033843 Chr.X: 72160365-72160365 on Build GRCh38
rs753934430 Chr.X: 72160207-72160207 on Build GRCh38
rs768891308 Chr.X: 72160453-72160453 on Build GRCh38
rs1017706728 Chr.X: 72160139-72160139 on Build GRCh38
rs937941749 Chr.X: 72160307-72160307 on Build GRCh38
rs760617068 Chr.X: 72160189-72160189 on Build GRCh38
rs879207436 Chr.X: 72160147-72160147 on Build GRCh38
rs558711983 Chr.X: 72160266-72160266 on Build GRCh38
rs781014416 Chr.X: 72160436-72160436 on Build GRCh38
rs771884513 Chr.X: 72160175-72160175 on Build GRCh38
rs756737071 Chr.X: 72160303-72160303 on Build GRCh38
rs754952504 Chr.X: 72160355-72160355 on Build GRCh38
rs950735181 Chr.X: 72160391-72160391 on Build GRCh38
rs772304523 Chr.X: 72160415-72160415 on Build GRCh38
rs186016133 Chr.X: 72160300-72160300 on Build GRCh38
rs749639480 Chr.X: 72160238-72160238 on Build GRCh38
rs878940992 Chr.X: 72160111-72160111 on Build GRCh38
rs983646903 Chr.X: 72160433-72160433 on Build GRCh38
rs752935788 Chr.X: 72160215-72160215 on Build GRCh38
rs774919908 Chr.X: 72160294-72160294 on Build GRCh38
rs11552481 Chr.X: 72160349-72160349 on Build GRCh38
rs770230402 Chr.X: 72160279-72160281 on Build GRCh38
rs878863009 Chr.X: 72160169-72160169 on Build GRCh38
rs748824035 Chr.X: 72160127-72160127 on Build GRCh38
rs759648916 Chr.X: 72160210-72160210 on Build GRCh38
rs959718950 Chr.X: 72160293-72160293 on Build GRCh38
rs780663439 Chr.X: 72160309-72160309 on Build GRCh38
rs993594358 Chr.X: 72160505-72160505 on Build GRCh38

View Full Product Details